Gene Transcripts
- Gene Symbol
- TPTE2
- Entrez Gene ID
- 93492
- Gene Name
- transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
- Gene Aliases
- TPIP
- Chromosome Location
- Chr. 13: 19422877 - 19561574 on Build GRCh38
- UniGene ID
- Hs.377488
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001141968.1 |
NP_001135440.1
|
3, 4 |
420 |
|
NM_130785.3 |
NP_570141.3
|
2, 3 |
228 |
|
NM_199254.2 |
NP_954863.2
|
2, 3 |
228 |
|
NR_073485.1 |
|
4, 5 |
340 |
|
NR_073486.1 |
|
1, 2 |
60 |
|
NR_073487.1 |
|
2, 3 |
228 |
|
GenBank mRNA |
AJ421032.1 |
- |
2, 3 |
228 |
|
AJ421033.1 |
- |
|
17 |
|
AK301741.1 |
- |
4, 5 |
340 |
|
AY219890.1 |
- |
2, 3 |
228 |
|
BC128146.1 |
- |
3, 4 |
402 |
40112547 |
BC128147.1 |
- |
2, 3 |
210 |
|
View Full Product Details