Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70108275-70108771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
UniGene
Hs.503074
Gene Chromosome Location
Chr.11: 70078302-70189545 on Build GRCh38
Transcripts
NM_018043.5 NR_030691.1 XM_006718602.2 XM_006718604.2 XM_006718605.2 XM_011545121.2 XM_011545123.2 XM_011545124.2 XM_011545125.2 XM_011545126.2 XM_011545127.2 XM_011545128.2 XM_011545129.2 XM_011545131.2 XM_017017956.1 XM_017017957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5827031 c.686C>T p.T229M Chr.11: 70108375-70108375 on Build GRCh38
4200321 c.664-1G>A p.? Chr.11: 70108352-70108352 on Build GRCh38
2043625 c.748-1G>A p.? Chr.11: 70108352-70108352 on Build GRCh38
5360381 c.799+10delT p.? Chr.11: 70108414-70108414 on Build GRCh38
4662005 c.798G>A p.M266I Chr.11: 70108403-70108403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770676842 Chr.11: 70108563-70108563 on Build GRCh38
rs1050584686 Chr.11: 70108435-70108435 on Build GRCh38
rs991347840 Chr.11: 70108652-70108652 on Build GRCh38
rs914777695 Chr.11: 70108682-70108682 on Build GRCh38
rs747968568 Chr.11: 70108378-70108378 on Build GRCh38
rs1005698765 Chr.11: 70108564-70108564 on Build GRCh38
rs114536292 Chr.11: 70108334-70108334 on Build GRCh38
rs753641865 Chr.11: 70108412-70108412 on Build GRCh38
rs774376698 Chr.11: 70108376-70108376 on Build GRCh38
rs764330705 Chr.11: 70108401-70108401 on Build GRCh38
rs1047032430 Chr.11: 70108712-70108712 on Build GRCh38
rs1041828326 Chr.11: 70108722-70108722 on Build GRCh38
rs541910902 Chr.11: 70108342-70108342 on Build GRCh38
rs74621510 Chr.11: 70108581-70108581 on Build GRCh38
rs752115819 Chr.11: 70108402-70108402 on Build GRCh38
rs543857789 Chr.11: 70108332-70108332 on Build GRCh38
rs138501952 Chr.11: 70108739-70108739 on Build GRCh38
rs777634453 Chr.11: 70108492-70108492 on Build GRCh38
rs749901776 Chr.11: 70108422-70108422 on Build GRCh38
rs368162367 Chr.11: 70108530-70108530 on Build GRCh38
rs1018386547 Chr.11: 70108571-70108571 on Build GRCh38
rs776837992 Chr.11: 70108380-70108380 on Build GRCh38
rs567007748 Chr.11: 70108671-70108671 on Build GRCh38
rs771149930 Chr.11: 70108379-70108379 on Build GRCh38
rs1011698550 Chr.11: 70108461-70108461 on Build GRCh38
rs754807123 Chr.11: 70108449-70108449 on Build GRCh38
rs755953496 Chr.11: 70108329-70108329 on Build GRCh38
rs2509182 Chr.11: 70108306-70108306 on Build GRCh38
rs1026112495 Chr.11: 70108315-70108315 on Build GRCh38
rs371160863 Chr.11: 70108318-70108318 on Build GRCh38
rs1026859574 Chr.11: 70108565-70108565 on Build GRCh38
rs888560707 Chr.11: 70108738-70108738 on Build GRCh38
rs527498587 Chr.11: 70108463-70108463 on Build GRCh38
rs919163160 Chr.11: 70108702-70108702 on Build GRCh38
rs1051112932 Chr.11: 70108676-70108676 on Build GRCh38
rs772216899 Chr.11: 70108662-70108662 on Build GRCh38
rs925730212 Chr.11: 70108639-70108639 on Build GRCh38
rs1045661132 Chr.11: 70108690-70108690 on Build GRCh38
rs759906575 Chr.11: 70108384-70108384 on Build GRCh38
rs759774513 Chr.11: 70108329-70108329 on Build GRCh38
rs371034128 Chr.11: 70108450-70108450 on Build GRCh38
rs559239268 Chr.11: 70108707-70108707 on Build GRCh38
rs1039739214 Chr.11: 70108747-70108747 on Build GRCh38
rs140582554 Chr.11: 70108431-70108431 on Build GRCh38
rs765526243 Chr.11: 70108389-70108389 on Build GRCh38
rs998465793 Chr.11: 70108583-70108583 on Build GRCh38
rs372786273 Chr.11: 70108674-70108674 on Build GRCh38
rs1002856080 Chr.11: 70108513-70108513 on Build GRCh38
rs775769327 Chr.11: 70108391-70108391 on Build GRCh38
rs1047179532 Chr.11: 70108557-70108557 on Build GRCh38
rs745574939 Chr.11: 70108349-70108349 on Build GRCh38
rs768285449 Chr.11: 70108375-70108375 on Build GRCh38
rs894419474 Chr.11: 70108464-70108464 on Build GRCh38
rs78268055 Chr.11: 70108640-70108640 on Build GRCh38
rs755810028 Chr.11: 70108424-70108424 on Build GRCh38
rs185890353 Chr.11: 70108554-70108554 on Build GRCh38
rs569239780 Chr.11: 70108587-70108587 on Build GRCh38
rs779861334 Chr.11: 70108362-70108362 on Build GRCh38
rs755754834 Chr.11: 70108358-70108358 on Build GRCh38
rs977483830 Chr.11: 70108619-70108619 on Build GRCh38
rs1046038529 Chr.11: 70108499-70108499 on Build GRCh38
rs767077417 Chr.11: 70108413-70108413 on Build GRCh38
rs368436856 Chr.11: 70108433-70108433 on Build GRCh38
rs967078109 Chr.11: 70108591-70108591 on Build GRCh38
rs534462014 Chr.11: 70108302-70108302 on Build GRCh38
rs573502709 Chr.11: 70108301-70108301 on Build GRCh38
rs150480281 Chr.11: 70108586-70108586 on Build GRCh38
rs977201213 Chr.11: 70108426-70108426 on Build GRCh38
rs749084555 Chr.11: 70108374-70108374 on Build GRCh38
rs551986355 Chr.11: 70108516-70108516 on Build GRCh38
rs536322986 Chr.11: 70108598-70108598 on Build GRCh38
rs181250366 Chr.11: 70108333-70108333 on Build GRCh38

View Full Product Details