Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70116378-70116883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
UniGene
Hs.503074
Gene Chromosome Location
Chr.11: 70078302-70189545 on Build GRCh38
Transcripts
NM_018043.5 NR_030691.1 XM_006718602.2 XM_006718604.2 XM_006718605.2 XM_011545121.2 XM_011545123.2 XM_011545124.2 XM_011545125.2 XM_011545126.2 XM_011545127.2 XM_011545128.2 XM_011545129.2 XM_011545131.2 XM_017017956.1 XM_017017957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2043637 c.879C>T p.V293V Chr.11: 70116481-70116481 on Build GRCh38
5823204 c.805G>C p.D269H Chr.11: 70116491-70116491 on Build GRCh38
7105596 c.801C>T p.F267F Chr.11: 70116487-70116487 on Build GRCh38
7188938 c.880G>T p.E294* Chr.11: 70116482-70116482 on Build GRCh38
7188940 c.796G>T p.E266* Chr.11: 70116482-70116482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043832742 Chr.11: 70116592-70116592 on Build GRCh38
rs762641258 Chr.11: 70116418-70116418 on Build GRCh38
rs371730153 Chr.11: 70116614-70116614 on Build GRCh38
rs762764354 Chr.11: 70116526-70116526 on Build GRCh38
rs367638256 Chr.11: 70116549-70116549 on Build GRCh38
rs765366737 Chr.11: 70116546-70116546 on Build GRCh38
rs962318381 Chr.11: 70116639-70116639 on Build GRCh38
rs750332523 Chr.11: 70116467-70116467 on Build GRCh38
rs958722556 Chr.11: 70116466-70116466 on Build GRCh38
rs758991143 Chr.11: 70116413-70116413 on Build GRCh38
rs541635940 Chr.11: 70116558-70116558 on Build GRCh38
rs746980476 Chr.11: 70116515-70116515 on Build GRCh38
rs1024703685 Chr.11: 70116423-70116423 on Build GRCh38
rs2515245 Chr.11: 70116536-70116536 on Build GRCh38
rs903735468 Chr.11: 70116601-70116601 on Build GRCh38
rs966302286 Chr.11: 70116832-70116832 on Build GRCh38
rs903646526 Chr.11: 70116426-70116426 on Build GRCh38
rs972206769 Chr.11: 70116683-70116683 on Build GRCh38
rs952215612 Chr.11: 70116746-70116746 on Build GRCh38
rs770025376 Chr.11: 70116638-70116638 on Build GRCh38
rs148980879 Chr.11: 70116775-70116775 on Build GRCh38
rs925498159 Chr.11: 70116693-70116693 on Build GRCh38
rs978469653 Chr.11: 70116563-70116563 on Build GRCh38
rs951636007 Chr.11: 70116649-70116649 on Build GRCh38
rs761572447 Chr.11: 70116566-70116566 on Build GRCh38
rs767375305 Chr.11: 70116465-70116465 on Build GRCh38
rs775129554 Chr.11: 70116524-70116524 on Build GRCh38
rs1001974749 Chr.11: 70116611-70116611 on Build GRCh38
rs763812474 Chr.11: 70116528-70116528 on Build GRCh38
rs1028153923 Chr.11: 70116698-70116698 on Build GRCh38
rs892104138 Chr.11: 70116580-70116580 on Build GRCh38
rs559789770 Chr.11: 70116559-70116559 on Build GRCh38
rs949245027 Chr.11: 70116749-70116749 on Build GRCh38
rs377438416 Chr.11: 70116531-70116531 on Build GRCh38
rs539071245 Chr.11: 70116507-70116507 on Build GRCh38
rs896419230 Chr.11: 70116615-70116615 on Build GRCh38
rs746014586 Chr.11: 70116428-70116430 on Build GRCh38
rs375801362 Chr.11: 70116478-70116478 on Build GRCh38
rs767272832 Chr.11: 70116535-70116535 on Build GRCh38
rs529353250 Chr.11: 70116779-70116779 on Build GRCh38
rs773353499 Chr.11: 70116674-70116674 on Build GRCh38
rs975483368 Chr.11: 70116679-70116681 on Build GRCh38
rs373500767 Chr.11: 70116514-70116514 on Build GRCh38
rs1019789225 Chr.11: 70116663-70116663 on Build GRCh38
rs778151682 Chr.11: 70116482-70116482 on Build GRCh38
rs747581961 Chr.11: 70116409-70116409 on Build GRCh38
rs757264330 Chr.11: 70116435-70116435 on Build GRCh38
rs986946315 Chr.11: 70116654-70116654 on Build GRCh38
rs988476729 Chr.11: 70116739-70116739 on Build GRCh38
rs1013895835 Chr.11: 70116619-70116619 on Build GRCh38
rs1013392232 Chr.11: 70116555-70116555 on Build GRCh38
rs200387111 Chr.11: 70116533-70116533 on Build GRCh38
rs573879283 Chr.11: 70116557-70116557 on Build GRCh38
rs772422314 Chr.11: 70116445-70116446 on Build GRCh38
rs969040316 Chr.11: 70116424-70116424 on Build GRCh38
rs879335268 Chr.11: 70116446-70116446 on Build GRCh38
rs765521159 Chr.11: 70116407-70116407 on Build GRCh38
rs368871647 Chr.11: 70116479-70116479 on Build GRCh38
rs369323555 Chr.11: 70116430-70116430 on Build GRCh38
rs913576689 Chr.11: 70116863-70116863 on Build GRCh38
rs910696633 Chr.11: 70116810-70116810 on Build GRCh38
rs974336769 Chr.11: 70116437-70116437 on Build GRCh38
rs375300283 Chr.11: 70116416-70116416 on Build GRCh38
rs372328002 Chr.11: 70116404-70116404 on Build GRCh38
rs756064260 Chr.11: 70116541-70116541 on Build GRCh38
rs189386669 Chr.11: 70116425-70116425 on Build GRCh38
rs879830448 Chr.11: 70116581-70116581 on Build GRCh38
rs372837155 Chr.11: 70116433-70116433 on Build GRCh38
rs755015798 Chr.11: 70116470-70116470 on Build GRCh38
rs912614155 Chr.11: 70116481-70116481 on Build GRCh38
rs892315231 Chr.11: 70116553-70116553 on Build GRCh38
rs936664099 Chr.11: 70116713-70116713 on Build GRCh38
rs747352529 Chr.11: 70116489-70116489 on Build GRCh38
rs771280216 Chr.11: 70116490-70116490 on Build GRCh38
rs764766877 Chr.11: 70116415-70116415 on Build GRCh38
rs777037795 Chr.11: 70116491-70116491 on Build GRCh38
rs976702561 Chr.11: 70116857-70116857 on Build GRCh38
rs776426117 Chr.11: 70116562-70116562 on Build GRCh38
rs373727703 Chr.11: 70116403-70116403 on Build GRCh38

View Full Product Details