Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70206799-70207304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADD
Entrez Gene ID
8772
Gene Name
Fas associated via death domain
Gene Aliases
GIG3, MORT1
UniGene
Hs.86131
Gene Chromosome Location
Chr.11: 70203163-70207402 on Build GRCh38
Transcripts
NM_003824.3

SNP Detail

SNP ID Chromosome Location
rs925024659 Chr.11: 70206880-70206880 on Build GRCh38
rs891710474 Chr.11: 70207274-70207274 on Build GRCh38
rs767813006 Chr.11: 70207221-70207221 on Build GRCh38
rs368539929 Chr.11: 70206884-70206884 on Build GRCh38
rs1057128980 Chr.11: 70206980-70206980 on Build GRCh38
rs968141004 Chr.11: 70207133-70207133 on Build GRCh38
rs565588896 Chr.11: 70207198-70207199 on Build GRCh38
rs1051422 Chr.11: 70207029-70207029 on Build GRCh38
rs985019405 Chr.11: 70207044-70207044 on Build GRCh38
rs397720848 Chr.11: 70207020-70207021 on Build GRCh38
rs542661900 Chr.11: 70207078-70207078 on Build GRCh38
rs180955808 Chr.11: 70206848-70206848 on Build GRCh38
rs981709757 Chr.11: 70207151-70207151 on Build GRCh38
rs940291106 Chr.11: 70207195-70207195 on Build GRCh38
rs910813820 Chr.11: 70207050-70207050 on Build GRCh38
rs149373917 Chr.11: 70207118-70207118 on Build GRCh38
rs898640527 Chr.11: 70207193-70207193 on Build GRCh38
rs532769136 Chr.11: 70207149-70207149 on Build GRCh38
rs374051992 Chr.11: 70207162-70207162 on Build GRCh38
rs561218421 Chr.11: 70207085-70207085 on Build GRCh38
rs575194795 Chr.11: 70207049-70207049 on Build GRCh38
rs1027972047 Chr.11: 70207040-70207040 on Build GRCh38
rs558881172 Chr.11: 70207124-70207124 on Build GRCh38
rs41269123 Chr.11: 70206994-70206994 on Build GRCh38
rs190056190 Chr.11: 70206952-70206952 on Build GRCh38
rs1025666558 Chr.11: 70207032-70207032 on Build GRCh38
rs78879392 Chr.11: 70207021-70207021 on Build GRCh38
rs551248887 Chr.11: 70207177-70207177 on Build GRCh38
rs753526348 Chr.11: 70207251-70207251 on Build GRCh38
rs766907631 Chr.11: 70207076-70207076 on Build GRCh38
rs551318384 Chr.11: 70206844-70206844 on Build GRCh38
rs988279155 Chr.11: 70207148-70207148 on Build GRCh38
rs890816007 Chr.11: 70207042-70207042 on Build GRCh38
rs41269125 Chr.11: 70207011-70207011 on Build GRCh38
rs4197 Chr.11: 70207017-70207018 on Build GRCh38
rs1019548422 Chr.11: 70207080-70207080 on Build GRCh38
rs539369302 Chr.11: 70206843-70206843 on Build GRCh38
rs915333577 Chr.11: 70206853-70206853 on Build GRCh38
rs756872185 Chr.11: 70207259-70207259 on Build GRCh38
rs61740747 Chr.11: 70206885-70206885 on Build GRCh38
rs1004389539 Chr.11: 70207006-70207006 on Build GRCh38
rs45542838 Chr.11: 70207019-70207020 on Build GRCh38
rs1031568671 Chr.11: 70207079-70207079 on Build GRCh38
rs946344587 Chr.11: 70206982-70206982 on Build GRCh38
rs1031333063 Chr.11: 70207255-70207255 on Build GRCh38
rs1055887329 Chr.11: 70207010-70207010 on Build GRCh38
rs548202059 Chr.11: 70207236-70207236 on Build GRCh38
rs562455931 Chr.11: 70207045-70207045 on Build GRCh38
rs955838146 Chr.11: 70207115-70207115 on Build GRCh38
rs111538026 Chr.11: 70207242-70207242 on Build GRCh38
rs569616488 Chr.11: 70207185-70207185 on Build GRCh38
rs908805287 Chr.11: 70207186-70207186 on Build GRCh38
rs1049956608 Chr.11: 70207230-70207230 on Build GRCh38
rs1045982058 Chr.11: 70206992-70206992 on Build GRCh38
rs533730679 Chr.11: 70206962-70206962 on Build GRCh38
rs528622608 Chr.11: 70207112-70207112 on Build GRCh38
rs112972843 Chr.11: 70207172-70207172 on Build GRCh38
rs926638632 Chr.11: 70206888-70206888 on Build GRCh38
rs544063886 Chr.11: 70207019-70207019 on Build GRCh38
rs887247714 Chr.11: 70206998-70206998 on Build GRCh38
rs138762178 Chr.11: 70207068-70207068 on Build GRCh38
rs1038546675 Chr.11: 70207212-70207212 on Build GRCh38
rs144756684 Chr.11: 70207215-70207215 on Build GRCh38

View Full Product Details