Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72106090-72106592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC15
Entrez Gene ID
25906
Gene Name
anaphase promoting complex subunit 15
Gene Aliases
APC15, C11orf51, HSPC020
UniGene
Hs.38044
Gene Chromosome Location
Chr.11: 72106254-72112793 on Build GRCh38
Transcripts
XM_017017488.1 XM_017017495.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017356.1 XM_017017357.1 XM_017017358.1

Gene Symbol
LRTOMT
Entrez Gene ID
220074
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases
CFAP111, DFNB63, LRRC51
UniGene
Hs.317243
Gene Chromosome Location
Chr.11: 72079982-72110782 on Build GRCh38
Transcripts
XM_017017488.1 XM_017017495.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017356.1 XM_017017357.1 XM_017017358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5429783 c.336G>C p.L112F Chr.11: 72106188-72106188 on Build GRCh38
7201280 c.287G>A p.G96D Chr.11: 72106139-72106139 on Build GRCh38
6042579 c.319A>G p.S107G Chr.11: 72106171-72106171 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767713899 Chr.11: 72106192-72106192 on Build GRCh38
rs999863107 Chr.11: 72106175-72106175 on Build GRCh38
rs1012740858 Chr.11: 72106443-72106443 on Build GRCh38
rs1031174889 Chr.11: 72106414-72106414 on Build GRCh38
rs886048629 Chr.11: 72106128-72106128 on Build GRCh38
rs561660987 Chr.11: 72106300-72106300 on Build GRCh38
rs902083155 Chr.11: 72106319-72106319 on Build GRCh38
rs991831788 Chr.11: 72106195-72106195 on Build GRCh38
rs137853187 Chr.11: 72106180-72106180 on Build GRCh38
rs564390967 Chr.11: 72106236-72106236 on Build GRCh38
rs774544844 Chr.11: 72106179-72106179 on Build GRCh38
rs547157797 Chr.11: 72106455-72106455 on Build GRCh38
rs777091947 Chr.11: 72106367-72106367 on Build GRCh38
rs550941923 Chr.11: 72106376-72106376 on Build GRCh38
rs867427331 Chr.11: 72106527-72106527 on Build GRCh38
rs767941990 Chr.11: 72106445-72106445 on Build GRCh38
rs919376046 Chr.11: 72106220-72106220 on Build GRCh38
rs545947177 Chr.11: 72106214-72106214 on Build GRCh38
rs137853188 Chr.11: 72106185-72106185 on Build GRCh38
rs916499499 Chr.11: 72106128-72106128 on Build GRCh38
rs989238802 Chr.11: 72106119-72106119 on Build GRCh38
rs776559425 Chr.11: 72106139-72106139 on Build GRCh38
rs764464540 Chr.11: 72106172-72106172 on Build GRCh38
rs947949971 Chr.11: 72106176-72106176 on Build GRCh38
rs527896965 Chr.11: 72106551-72106551 on Build GRCh38
rs367802080 Chr.11: 72106550-72106550 on Build GRCh38
rs900829173 Chr.11: 72106385-72106385 on Build GRCh38
rs1007526596 Chr.11: 72106330-72106331 on Build GRCh38
rs560775281 Chr.11: 72106531-72106531 on Build GRCh38
rs958556554 Chr.11: 72106423-72106423 on Build GRCh38
rs971336687 Chr.11: 72106479-72106479 on Build GRCh38
rs763075211 Chr.11: 72106143-72106143 on Build GRCh38
rs931478705 Chr.11: 72106264-72106264 on Build GRCh38
rs181092713 Chr.11: 72106204-72106204 on Build GRCh38
rs529034429 Chr.11: 72106393-72106393 on Build GRCh38
rs12576752 Chr.11: 72106245-72106245 on Build GRCh38
rs997847456 Chr.11: 72106126-72106126 on Build GRCh38
rs770566557 Chr.11: 72106108-72106108 on Build GRCh38
rs910737723 Chr.11: 72106243-72106243 on Build GRCh38
rs1032698535 Chr.11: 72106407-72106407 on Build GRCh38
rs773009362 Chr.11: 72106197-72106197 on Build GRCh38
rs773358488 Chr.11: 72106163-72106164 on Build GRCh38
rs890062285 Chr.11: 72106306-72106306 on Build GRCh38
rs967095444 Chr.11: 72106154-72106154 on Build GRCh38
rs1023395484 Chr.11: 72106549-72106549 on Build GRCh38
rs998296595 Chr.11: 72106353-72106353 on Build GRCh38
rs137853186 Chr.11: 72106165-72106165 on Build GRCh38
rs760043225 Chr.11: 72106395-72106395 on Build GRCh38
rs957857044 Chr.11: 72106118-72106118 on Build GRCh38
rs1020485331 Chr.11: 72106129-72106129 on Build GRCh38
rs930719591 Chr.11: 72106223-72106223 on Build GRCh38

View Full Product Details