Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75435298-75435785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD5
Entrez Gene ID
81544
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
GDE2, PP1665
UniGene
Hs.503297
Gene Chromosome Location
Chr.11: 75434640-75525916 on Build GRCh38
Transcripts
NM_030792.6 XM_006718697.3 XM_011545276.2 XM_011545277.2 XM_011545278.2 XM_011545279.2 XM_011545280.1 XM_011545281.2 XM_011545285.2 XM_011545286.1 XM_011545287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670402 c.1707C>T p.S569S Chr.11: 75435618-75435618 on Build GRCh38
931988 c.1763G>A p.G588D Chr.11: 75435562-75435562 on Build GRCh38
4036735 c.1768C>T p.R590* Chr.11: 75435557-75435557 on Build GRCh38
4686753 c.1813C>T p.R605C Chr.11: 75435512-75435512 on Build GRCh38
211801 c.1787C>A p.T596N Chr.11: 75435538-75435538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983737566 Chr.11: 75435648-75435648 on Build GRCh38
rs376398826 Chr.11: 75435641-75435641 on Build GRCh38
rs12790806 Chr.11: 75435746-75435746 on Build GRCh38
rs754023687 Chr.11: 75435663-75435663 on Build GRCh38
rs749608394 Chr.11: 75435629-75435629 on Build GRCh38
rs1010187248 Chr.11: 75435393-75435393 on Build GRCh38
rs946323401 Chr.11: 75435739-75435739 on Build GRCh38
rs778938510 Chr.11: 75435591-75435591 on Build GRCh38
rs760729755 Chr.11: 75435660-75435660 on Build GRCh38
rs569209623 Chr.11: 75435717-75435717 on Build GRCh38
rs540766381 Chr.11: 75435760-75435760 on Build GRCh38
rs1046191051 Chr.11: 75435436-75435436 on Build GRCh38
rs12790617 Chr.11: 75435723-75435723 on Build GRCh38
rs750661900 Chr.11: 75435497-75435497 on Build GRCh38
rs148544864 Chr.11: 75435618-75435618 on Build GRCh38
rs768958934 Chr.11: 75435698-75435698 on Build GRCh38
rs964493099 Chr.11: 75435346-75435346 on Build GRCh38
rs752634222 Chr.11: 75435671-75435671 on Build GRCh38
rs569699868 Chr.11: 75435575-75435575 on Build GRCh38
rs758390363 Chr.11: 75435680-75435680 on Build GRCh38
rs574893404 Chr.11: 75435396-75435396 on Build GRCh38
rs535938113 Chr.11: 75435332-75435332 on Build GRCh38
rs914692763 Chr.11: 75435728-75435728 on Build GRCh38
rs376454689 Chr.11: 75435687-75435687 on Build GRCh38
rs974646219 Chr.11: 75435347-75435347 on Build GRCh38
rs779528500 Chr.11: 75435345-75435345 on Build GRCh38
rs750559361 Chr.11: 75435557-75435557 on Build GRCh38
rs906047811 Chr.11: 75435480-75435480 on Build GRCh38
rs1018622681 Chr.11: 75435567-75435567 on Build GRCh38
rs775901461 Chr.11: 75435537-75435537 on Build GRCh38
rs527424229 Chr.11: 75435354-75435354 on Build GRCh38
rs183534412 Chr.11: 75435458-75435458 on Build GRCh38
rs12792174 Chr.11: 75435744-75435744 on Build GRCh38
rs765157828 Chr.11: 75435670-75435670 on Build GRCh38
rs1054620434 Chr.11: 75435725-75435725 on Build GRCh38
rs752073631 Chr.11: 75435547-75435547 on Build GRCh38
rs770587553 Chr.11: 75435617-75435617 on Build GRCh38
rs187459718 Chr.11: 75435624-75435624 on Build GRCh38
rs537176106 Chr.11: 75435630-75435630 on Build GRCh38
rs558683671 Chr.11: 75435712-75435712 on Build GRCh38
rs779850795 Chr.11: 75435603-75435604 on Build GRCh38
rs575520727 Chr.11: 75435368-75435368 on Build GRCh38
rs751633617 Chr.11: 75435485-75435485 on Build GRCh38
rs540408478 Chr.11: 75435448-75435448 on Build GRCh38
rs764635820 Chr.11: 75435481-75435481 on Build GRCh38
rs747662962 Chr.11: 75435688-75435688 on Build GRCh38
rs991810972 Chr.11: 75435483-75435483 on Build GRCh38
rs747652235 Chr.11: 75435513-75435513 on Build GRCh38
rs758166903 Chr.11: 75435470-75435471 on Build GRCh38
rs1043638 Chr.11: 75435339-75435339 on Build GRCh38
rs887730578 Chr.11: 75435321-75435321 on Build GRCh38
rs561078577 Chr.11: 75435394-75435394 on Build GRCh38
rs780830160 Chr.11: 75435693-75435693 on Build GRCh38
rs748104045 Chr.11: 75435593-75435593 on Build GRCh38
rs964545542 Chr.11: 75435568-75435568 on Build GRCh38
rs759730108 Chr.11: 75435669-75435669 on Build GRCh38
rs746550127 Chr.11: 75435609-75435609 on Build GRCh38
rs528344017 Chr.11: 75435490-75435490 on Build GRCh38
rs754882301 Chr.11: 75435588-75435588 on Build GRCh38
rs781609091 Chr.11: 75435551-75435551 on Build GRCh38
rs968172808 Chr.11: 75435429-75435429 on Build GRCh38
rs955475133 Chr.11: 75435369-75435369 on Build GRCh38
rs193179784 Chr.11: 75435703-75435703 on Build GRCh38
rs538127273 Chr.11: 75435761-75435761 on Build GRCh38
rs551593132 Chr.11: 75435511-75435511 on Build GRCh38
rs529675945 Chr.11: 75435683-75435683 on Build GRCh38
rs372960103 Chr.11: 75435542-75435542 on Build GRCh38
rs768603832 Chr.11: 75435512-75435512 on Build GRCh38
rs926838927 Chr.11: 75435450-75435450 on Build GRCh38
rs756334817 Chr.11: 75435561-75435561 on Build GRCh38
rs538826350 Chr.11: 75435358-75435358 on Build GRCh38
rs1027564886 Chr.11: 75435598-75435598 on Build GRCh38
rs2155166 Chr.11: 75435700-75435700 on Build GRCh38
rs558866419 Chr.11: 75435363-75435363 on Build GRCh38
rs761812654 Chr.11: 75435549-75435549 on Build GRCh38
rs770006144 Chr.11: 75435526-75435526 on Build GRCh38
rs777367424 Chr.11: 75435608-75435608 on Build GRCh38
rs917594274 Chr.11: 75435748-75435748 on Build GRCh38
rs746911586 Chr.11: 75435676-75435677 on Build GRCh38
rs1042339550 Chr.11: 75435747-75435747 on Build GRCh38
rs144731330 Chr.11: 75435576-75435576 on Build GRCh38
rs757385377 Chr.11: 75435489-75435489 on Build GRCh38
rs764570484 Chr.11: 75435544-75435544 on Build GRCh38
rs779919308 Chr.11: 75435509-75435509 on Build GRCh38
rs917693526 Chr.11: 75435515-75435515 on Build GRCh38
rs138154091 Chr.11: 75435702-75435702 on Build GRCh38
rs745910762 Chr.11: 75435520-75435520 on Build GRCh38
rs781332651 Chr.11: 75435496-75435496 on Build GRCh38
rs544601341 Chr.11: 75435370-75435370 on Build GRCh38
rs766614363 Chr.11: 75435662-75435662 on Build GRCh38
rs757942561 Chr.11: 75435597-75435597 on Build GRCh38
rs767614808 Chr.11: 75435556-75435556 on Build GRCh38
rs373412770 Chr.11: 75435649-75435649 on Build GRCh38
rs201881925 Chr.11: 75435521-75435521 on Build GRCh38
rs369743472 Chr.11: 75435647-75435647 on Build GRCh38

View Full Product Details