Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75436852-75437357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD5
Entrez Gene ID
81544
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
GDE2, PP1665
UniGene
Hs.503297
Gene Chromosome Location
Chr.11: 75434640-75525916 on Build GRCh38
Transcripts
NM_030792.6 XM_006718697.3 XM_011545276.2 XM_011545277.2 XM_011545278.2 XM_011545279.2 XM_011545280.1 XM_011545281.2 XM_011545285.2 XM_011545286.1 XM_011545287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369370 c.1631G>A p.R544Q Chr.11: 75436974-75436974 on Build GRCh38
6670395 c.1577G>A p.R526Q Chr.11: 75437028-75437028 on Build GRCh38
6841594 c.1618C>T p.R540C Chr.11: 75436987-75436987 on Build GRCh38
931989 c.1613C>T p.A538V Chr.11: 75436992-75436992 on Build GRCh38
4686754 c.1592A>T p.E531V Chr.11: 75437013-75437013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370697890 Chr.11: 75437060-75437060 on Build GRCh38
rs144938037 Chr.11: 75437027-75437027 on Build GRCh38
rs761633624 Chr.11: 75437028-75437028 on Build GRCh38
rs183462310 Chr.11: 75437067-75437067 on Build GRCh38
rs748495456 Chr.11: 75436983-75436983 on Build GRCh38
rs762909557 Chr.11: 75436927-75436927 on Build GRCh38
rs991195569 Chr.11: 75437326-75437326 on Build GRCh38
rs749926326 Chr.11: 75437037-75437037 on Build GRCh38
rs188334441 Chr.11: 75437132-75437132 on Build GRCh38
rs139459790 Chr.11: 75437161-75437161 on Build GRCh38
rs1012850067 Chr.11: 75437246-75437246 on Build GRCh38
rs982119676 Chr.11: 75437306-75437306 on Build GRCh38
rs962037143 Chr.11: 75437235-75437235 on Build GRCh38
rs1019630720 Chr.11: 75436885-75436885 on Build GRCh38
rs147755302 Chr.11: 75436986-75436986 on Build GRCh38
rs770136380 Chr.11: 75436994-75436994 on Build GRCh38
rs144535854 Chr.11: 75436974-75436974 on Build GRCh38
rs151185777 Chr.11: 75436979-75436979 on Build GRCh38
rs947297554 Chr.11: 75437099-75437099 on Build GRCh38
rs1041135441 Chr.11: 75437148-75437148 on Build GRCh38
rs556738736 Chr.11: 75437303-75437303 on Build GRCh38
rs755676909 Chr.11: 75437043-75437043 on Build GRCh38
rs920883934 Chr.11: 75437062-75437062 on Build GRCh38
rs762768246 Chr.11: 75437009-75437009 on Build GRCh38
rs971752283 Chr.11: 75437237-75437237 on Build GRCh38
rs192729036 Chr.11: 75436901-75436901 on Build GRCh38
rs539818516 Chr.11: 75437273-75437273 on Build GRCh38
rs996953039 Chr.11: 75436964-75436964 on Build GRCh38
rs952887699 Chr.11: 75437219-75437219 on Build GRCh38
rs776831478 Chr.11: 75436988-75436988 on Build GRCh38
rs753496541 Chr.11: 75437045-75437045 on Build GRCh38
rs554306014 Chr.11: 75437165-75437165 on Build GRCh38
rs1050665647 Chr.11: 75437080-75437082 on Build GRCh38
rs570893949 Chr.11: 75437177-75437177 on Build GRCh38
rs778025538 Chr.11: 75437064-75437064 on Build GRCh38
rs1024199766 Chr.11: 75437257-75437257 on Build GRCh38
rs775912902 Chr.11: 75437000-75437000 on Build GRCh38
rs745930501 Chr.11: 75437096-75437096 on Build GRCh38
rs201429141 Chr.11: 75436975-75436975 on Build GRCh38
rs1028455945 Chr.11: 75437195-75437195 on Build GRCh38
rs776680526 Chr.11: 75436950-75436950 on Build GRCh38
rs75763080 Chr.11: 75436969-75436969 on Build GRCh38
rs145342602 Chr.11: 75436943-75436943 on Build GRCh38
rs772340698 Chr.11: 75436984-75436984 on Build GRCh38
rs781781936 Chr.11: 75437071-75437071 on Build GRCh38
rs760753595 Chr.11: 75436905-75436905 on Build GRCh38
rs756885247 Chr.11: 75437274-75437274 on Build GRCh38
rs764033083 Chr.11: 75436952-75436952 on Build GRCh38
rs902923607 Chr.11: 75437136-75437136 on Build GRCh38
rs149941214 Chr.11: 75437023-75437023 on Build GRCh38
rs751456136 Chr.11: 75436963-75436963 on Build GRCh38
rs1000066289 Chr.11: 75437204-75437204 on Build GRCh38
rs140932082 Chr.11: 75437029-75437029 on Build GRCh38
rs562241573 Chr.11: 75437012-75437012 on Build GRCh38
rs770996558 Chr.11: 75436987-75436987 on Build GRCh38
rs771312458 Chr.11: 75437068-75437068 on Build GRCh38
rs753096399 Chr.11: 75437133-75437133 on Build GRCh38
rs959783539 Chr.11: 75437295-75437295 on Build GRCh38
rs529959632 Chr.11: 75436891-75436891 on Build GRCh38
rs747252431 Chr.11: 75436909-75436909 on Build GRCh38
rs568666238 Chr.11: 75437116-75437116 on Build GRCh38
rs997005371 Chr.11: 75437181-75437181 on Build GRCh38
rs1042957501 Chr.11: 75437120-75437120 on Build GRCh38
rs901264208 Chr.11: 75437155-75437155 on Build GRCh38
rs372590239 Chr.11: 75436908-75436908 on Build GRCh38
rs778279799 Chr.11: 75436985-75436985 on Build GRCh38
rs759731505 Chr.11: 75436991-75436991 on Build GRCh38
rs766776502 Chr.11: 75436883-75436883 on Build GRCh38
rs752509447 Chr.11: 75436920-75436920 on Build GRCh38
rs1006229950 Chr.11: 75437222-75437222 on Build GRCh38
rs772996205 Chr.11: 75437272-75437272 on Build GRCh38
rs369220088 Chr.11: 75437288-75437288 on Build GRCh38
rs777194804 Chr.11: 75437231-75437231 on Build GRCh38
rs140583703 Chr.11: 75436970-75436970 on Build GRCh38
rs1015832700 Chr.11: 75437223-75437223 on Build GRCh38
rs747451006 Chr.11: 75437065-75437065 on Build GRCh38
rs973533058 Chr.11: 75437059-75437059 on Build GRCh38
rs559690888 Chr.11: 75436914-75436914 on Build GRCh38
rs779621330 Chr.11: 75437044-75437044 on Build GRCh38
rs759861566 Chr.11: 75436910-75436910 on Build GRCh38

View Full Product Details