Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75442122-75442658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD5
Entrez Gene ID
81544
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
GDE2, PP1665
UniGene
Hs.503297
Gene Chromosome Location
Chr.11: 75434640-75525916 on Build GRCh38
Transcripts
NM_030792.6 XM_006718697.3 XM_011545276.2 XM_011545277.2 XM_011545278.2 XM_011545279.2 XM_011545280.1 XM_011545281.2 XM_011545285.2 XM_011545286.1 XM_011545287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117061 c.1091G>A p.R364Q Chr.11: 75442439-75442439 on Build GRCh38
4886460 c.972C>A p.S324R Chr.11: 75442558-75442558 on Build GRCh38
3453354 c.1014C>T p.S338S Chr.11: 75442516-75442516 on Build GRCh38
5739378 c.1113T>A p.S371R Chr.11: 75442417-75442417 on Build GRCh38
931991 c.1019G>A p.C340Y Chr.11: 75442511-75442511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774346065 Chr.11: 75442381-75442381 on Build GRCh38
rs546074207 Chr.11: 75442347-75442347 on Build GRCh38
rs941604022 Chr.11: 75442565-75442565 on Build GRCh38
rs369818036 Chr.11: 75442350-75442350 on Build GRCh38
rs961791595 Chr.11: 75442186-75442186 on Build GRCh38
rs757811407 Chr.11: 75442542-75442542 on Build GRCh38
rs763876729 Chr.11: 75442380-75442380 on Build GRCh38
rs753534413 Chr.11: 75442501-75442501 on Build GRCh38
rs991443978 Chr.11: 75442211-75442211 on Build GRCh38
rs922826182 Chr.11: 75442299-75442299 on Build GRCh38
rs562373998 Chr.11: 75442312-75442312 on Build GRCh38
rs771143854 Chr.11: 75442334-75442334 on Build GRCh38
rs1015901880 Chr.11: 75442175-75442175 on Build GRCh38
rs775118711 Chr.11: 75442357-75442357 on Build GRCh38
rs761716355 Chr.11: 75442451-75442451 on Build GRCh38
rs140233225 Chr.11: 75442425-75442425 on Build GRCh38
rs553628735 Chr.11: 75442627-75442627 on Build GRCh38
rs143278799 Chr.11: 75442409-75442409 on Build GRCh38
rs986033127 Chr.11: 75442537-75442537 on Build GRCh38
rs1041500785 Chr.11: 75442282-75442282 on Build GRCh38
rs375711308 Chr.11: 75442439-75442439 on Build GRCh38
rs965755753 Chr.11: 75442280-75442280 on Build GRCh38
rs779599639 Chr.11: 75442338-75442338 on Build GRCh38
rs542403440 Chr.11: 75442232-75442232 on Build GRCh38
rs768057862 Chr.11: 75442576-75442579 on Build GRCh38
rs533870538 Chr.11: 75442546-75442546 on Build GRCh38
rs930625626 Chr.11: 75442426-75442426 on Build GRCh38
rs745913078 Chr.11: 75442562-75442562 on Build GRCh38
rs575574610 Chr.11: 75442272-75442272 on Build GRCh38
rs1035540777 Chr.11: 75442199-75442199 on Build GRCh38
rs749983548 Chr.11: 75442390-75442390 on Build GRCh38
rs750470860 Chr.11: 75442569-75442569 on Build GRCh38
rs367673696 Chr.11: 75442620-75442620 on Build GRCh38
rs960129675 Chr.11: 75442202-75442202 on Build GRCh38
rs1007825926 Chr.11: 75442448-75442448 on Build GRCh38
rs978740681 Chr.11: 75442257-75442257 on Build GRCh38
rs774859387 Chr.11: 75442368-75442373 on Build GRCh38
rs759900921 Chr.11: 75442399-75442400 on Build GRCh38
rs772703046 Chr.11: 75442473-75442473 on Build GRCh38
rs999068208 Chr.11: 75442456-75442456 on Build GRCh38
rs993629699 Chr.11: 75442194-75442194 on Build GRCh38
rs570889737 Chr.11: 75442348-75442348 on Build GRCh38
rs146651187 Chr.11: 75442424-75442424 on Build GRCh38
rs368361611 Chr.11: 75442452-75442452 on Build GRCh38
rs554296536 Chr.11: 75442442-75442442 on Build GRCh38
rs748582968 Chr.11: 75442319-75442319 on Build GRCh38
rs763187804 Chr.11: 75442506-75442506 on Build GRCh38
rs756292537 Chr.11: 75442223-75442223 on Build GRCh38
rs758804960 Chr.11: 75442400-75442400 on Build GRCh38
rs531974668 Chr.11: 75442165-75442165 on Build GRCh38
rs371189017 Chr.11: 75442531-75442531 on Build GRCh38
rs778076504 Chr.11: 75442407-75442407 on Build GRCh38
rs994886053 Chr.11: 75442210-75442210 on Build GRCh38
rs372653666 Chr.11: 75442356-75442356 on Build GRCh38
rs1031826542 Chr.11: 75442465-75442465 on Build GRCh38
rs749672358 Chr.11: 75442277-75442277 on Build GRCh38
rs533150675 Chr.11: 75442371-75442371 on Build GRCh38
rs376352073 Chr.11: 75442611-75442611 on Build GRCh38
rs889399266 Chr.11: 75442236-75442236 on Build GRCh38
rs771982431 Chr.11: 75442621-75442621 on Build GRCh38
rs141005869 Chr.11: 75442408-75442408 on Build GRCh38
rs748966109 Chr.11: 75442444-75442444 on Build GRCh38
rs761268895 Chr.11: 75442384-75442384 on Build GRCh38
rs780094934 Chr.11: 75442594-75442594 on Build GRCh38
rs377131289 Chr.11: 75442392-75442392 on Build GRCh38
rs562440951 Chr.11: 75442160-75442160 on Build GRCh38
rs768808844 Chr.11: 75442602-75442602 on Build GRCh38
rs778404533 Chr.11: 75442331-75442331 on Build GRCh38
rs752992362 Chr.11: 75442395-75442395 on Build GRCh38
rs752022999 Chr.11: 75442535-75442535 on Build GRCh38
rs61737932 Chr.11: 75442492-75442492 on Build GRCh38
rs760292424 Chr.11: 75442484-75442484 on Build GRCh38
rs34354475 Chr.11: 75442378-75442379 on Build GRCh38
rs139846951 Chr.11: 75442393-75442393 on Build GRCh38
rs774223933 Chr.11: 75442445-75442445 on Build GRCh38
rs1050419682 Chr.11: 75442153-75442153 on Build GRCh38
rs961186201 Chr.11: 75442625-75442625 on Build GRCh38
rs1031648870 Chr.11: 75442431-75442431 on Build GRCh38
rs956223976 Chr.11: 75442440-75442440 on Build GRCh38
rs934766768 Chr.11: 75442276-75442276 on Build GRCh38
rs199811868 Chr.11: 75442333-75442333 on Build GRCh38
rs767036518 Chr.11: 75442386-75442386 on Build GRCh38
rs1007711715 Chr.11: 75442250-75442251 on Build GRCh38
rs999017956 Chr.11: 75442290-75442290 on Build GRCh38
rs897823854 Chr.11: 75442208-75442208 on Build GRCh38
rs1013158474 Chr.11: 75442387-75442387 on Build GRCh38
rs756220201 Chr.11: 75442586-75442586 on Build GRCh38
rs749543050 Chr.11: 75442596-75442596 on Build GRCh38
rs374322974 Chr.11: 75442303-75442303 on Build GRCh38
rs758828715 Chr.11: 75442329-75442329 on Build GRCh38
rs867643865 Chr.11: 75442485-75442485 on Build GRCh38

View Full Product Details