Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77589463-77589956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP11
Entrez Gene ID
282679
Gene Name
aquaporin 11
Gene Aliases
AQPX1
UniGene
Hs.503345
Gene Chromosome Location
Chr.11: 77589635-77610356 on Build GRCh38
Transcripts
NM_173039.2 XM_005273917.4 XM_017018644.1

Gene Symbol
LOC107987163
Entrez Gene ID
107987163
Gene Name
uncharacterized LOC107987163
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 77523814-77616414 on Build GRCh38
Transcripts
NM_173039.2 XM_005273917.4 XM_017018644.1

SNP Detail

SNP ID Chromosome Location
rs142331691 Chr.11: 77589777-77589777 on Build GRCh38
rs535964160 Chr.11: 77589593-77589593 on Build GRCh38
rs1008237195 Chr.11: 77589621-77589621 on Build GRCh38
rs989877311 Chr.11: 77589828-77589835 on Build GRCh38
rs925897515 Chr.11: 77589676-77589676 on Build GRCh38
rs971379172 Chr.11: 77589764-77589764 on Build GRCh38
rs941320960 Chr.11: 77589541-77589541 on Build GRCh38
rs772196095 Chr.11: 77589862-77589862 on Build GRCh38
rs994805533 Chr.11: 77589753-77589753 on Build GRCh38
rs4301811 Chr.11: 77589696-77589696 on Build GRCh38
rs750366609 Chr.11: 77589765-77589772 on Build GRCh38
rs141363114 Chr.11: 77589776-77589776 on Build GRCh38
rs60220816 Chr.11: 77589766-77589775 on Build GRCh38
rs557532206 Chr.11: 77589666-77589666 on Build GRCh38
rs949740036 Chr.11: 77589652-77589652 on Build GRCh38
rs970278623 Chr.11: 77589809-77589809 on Build GRCh38
rs954958306 Chr.11: 77589616-77589616 on Build GRCh38
rs1016755876 Chr.11: 77589619-77589619 on Build GRCh38
rs973470509 Chr.11: 77589544-77589544 on Build GRCh38
rs1052693373 Chr.11: 77589855-77589855 on Build GRCh38
rs948636314 Chr.11: 77589668-77589668 on Build GRCh38
rs926945667 Chr.11: 77589792-77589792 on Build GRCh38
rs750460595 Chr.11: 77589548-77589548 on Build GRCh38
rs946217829 Chr.11: 77589671-77589671 on Build GRCh38
rs950808024 Chr.11: 77589762-77589762 on Build GRCh38
rs545031882 Chr.11: 77589767-77589767 on Build GRCh38
rs998360377 Chr.11: 77589685-77589685 on Build GRCh38
rs113419462 Chr.11: 77589664-77589664 on Build GRCh38
rs1051908300 Chr.11: 77589694-77589694 on Build GRCh38
rs948648871 Chr.11: 77589554-77589554 on Build GRCh38
rs535636365 Chr.11: 77589519-77589519 on Build GRCh38
rs929120948 Chr.11: 77589547-77589547 on Build GRCh38
rs187710032 Chr.11: 77589677-77589677 on Build GRCh38
rs1024809956 Chr.11: 77589931-77589931 on Build GRCh38
rs564229141 Chr.11: 77589612-77589636 on Build GRCh38
rs535046879 Chr.11: 77589608-77589608 on Build GRCh38
rs775340011 Chr.11: 77589745-77589754 on Build GRCh38
rs1007031451 Chr.11: 77589618-77589641 on Build GRCh38
rs1002931969 Chr.11: 77589635-77589635 on Build GRCh38
rs986108178 Chr.11: 77589524-77589524 on Build GRCh38
rs910572346 Chr.11: 77589538-77589538 on Build GRCh38
rs1047094903 Chr.11: 77589556-77589556 on Build GRCh38
rs528440934 Chr.11: 77589545-77589545 on Build GRCh38
rs1014283052 Chr.11: 77589929-77589929 on Build GRCh38
rs781599953 Chr.11: 77589894-77589894 on Build GRCh38
rs904212338 Chr.11: 77589573-77589573 on Build GRCh38
rs1017897657 Chr.11: 77589629-77589629 on Build GRCh38
rs566101531 Chr.11: 77589599-77589599 on Build GRCh38
rs1026321077 Chr.11: 77589754-77589754 on Build GRCh38
rs985288358 Chr.11: 77589518-77589518 on Build GRCh38
rs942064352 Chr.11: 77589542-77589542 on Build GRCh38
rs953983415 Chr.11: 77589506-77589506 on Build GRCh38
rs1010458045 Chr.11: 77589581-77589581 on Build GRCh38
rs925866562 Chr.11: 77589839-77589839 on Build GRCh38
rs530839377 Chr.11: 77589869-77589869 on Build GRCh38
rs1051125079 Chr.11: 77589925-77589925 on Build GRCh38
rs963658269 Chr.11: 77589734-77589734 on Build GRCh38
rs886426954 Chr.11: 77589927-77589927 on Build GRCh38
rs780765719 Chr.11: 77589672-77589672 on Build GRCh38
rs917371112 Chr.11: 77589791-77589791 on Build GRCh38
rs894611410 Chr.11: 77589749-77589749 on Build GRCh38
rs950871088 Chr.11: 77589660-77589660 on Build GRCh38
rs953510330 Chr.11: 77589616-77589617 on Build GRCh38
rs528064319 Chr.11: 77589705-77589705 on Build GRCh38
rs558151029 Chr.11: 77589634-77589634 on Build GRCh38
rs530611629 Chr.11: 77589502-77589502 on Build GRCh38
rs1026291444 Chr.11: 77589926-77589926 on Build GRCh38
rs1017867981 Chr.11: 77589709-77589709 on Build GRCh38
rs7115612 Chr.11: 77589500-77589500 on Build GRCh38
rs967644212 Chr.11: 77589813-77589813 on Build GRCh38
rs369175306 Chr.11: 77589909-77589909 on Build GRCh38
rs565031815 Chr.11: 77589788-77589788 on Build GRCh38
rs570007216 Chr.11: 77589555-77589555 on Build GRCh38
rs979947525 Chr.11: 77589836-77589836 on Build GRCh38
rs1011709288 Chr.11: 77589769-77589769 on Build GRCh38
rs933620236 Chr.11: 77589915-77589915 on Build GRCh38
rs933698465 Chr.11: 77589875-77589875 on Build GRCh38
rs879184389 Chr.11: 77589910-77589910 on Build GRCh38
rs867216561 Chr.11: 77589678-77589678 on Build GRCh38
rs566460883 Chr.11: 77589764-77589765 on Build GRCh38
rs899376951 Chr.11: 77589919-77589919 on Build GRCh38
rs370259820 Chr.11: 77589772-77589773 on Build GRCh38
rs550800754 Chr.11: 77589521-77589521 on Build GRCh38
rs919363899 Chr.11: 77589546-77589546 on Build GRCh38
rs995105429 Chr.11: 77589923-77589923 on Build GRCh38
rs551588971 Chr.11: 77589552-77589552 on Build GRCh38
rs1009702236 Chr.11: 77589609-77589609 on Build GRCh38
rs1044044825 Chr.11: 77589557-77589557 on Build GRCh38
rs901819915 Chr.11: 77589600-77589600 on Build GRCh38
rs935920819 Chr.11: 77589849-77589849 on Build GRCh38
rs958364506 Chr.11: 77589827-77589827 on Build GRCh38
rs1030917090 Chr.11: 77589614-77589614 on Build GRCh38
rs963353785 Chr.11: 77589631-77589631 on Build GRCh38
rs1006196074 Chr.11: 77589932-77589932 on Build GRCh38
rs1038819537 Chr.11: 77589935-77589958 on Build GRCh38
rs145090302 Chr.11: 77589763-77589764 on Build GRCh38
rs1051268346 Chr.11: 77589883-77589883 on Build GRCh38
rs919332897 Chr.11: 77589654-77589654 on Build GRCh38
rs902120562 Chr.11: 77589675-77589675 on Build GRCh38
rs891504412 Chr.11: 77589681-77589681 on Build GRCh38
rs4486654 Chr.11: 77589598-77589598 on Build GRCh38
rs539492071 Chr.11: 77589755-77589772 on Build GRCh38
rs927033292 Chr.11: 77589658-77589658 on Build GRCh38
rs578225068 Chr.11: 77589650-77589650 on Build GRCh38
rs1025966151 Chr.11: 77589714-77589714 on Build GRCh38
rs914338944 Chr.11: 77589843-77589843 on Build GRCh38

View Full Product Details