Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103113577-103114120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC2H1
Entrez Gene ID
79659
Gene Name
dynein cytoplasmic 2 heavy chain 1
Gene Aliases
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
UniGene
Hs.503721
Gene Chromosome Location
Chr.11: 103109431-103479863 on Build GRCh38
Transcripts
NM_001080463.1 NM_001377.2 XM_006718903.2 XM_017018291.1 XM_017018293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346199 c.263A>G p.D88G Chr.11: 103113604-103113604 on Build GRCh38
4017309 c.342A>C p.Q114H Chr.11: 103113683-103113683 on Build GRCh38
4017308 c.239A>G p.K80R Chr.11: 103113580-103113580 on Build GRCh38
260619 c.253G>T p.V85L Chr.11: 103113594-103113594 on Build GRCh38
3967275 c.334G>A p.V112I Chr.11: 103113675-103113675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933764802 Chr.11: 103114058-103114058 on Build GRCh38
rs961092240 Chr.11: 103114005-103114005 on Build GRCh38
rs558261659 Chr.11: 103113985-103113985 on Build GRCh38
rs996995565 Chr.11: 103113923-103113923 on Build GRCh38
rs780782483 Chr.11: 103114068-103114068 on Build GRCh38
rs889437709 Chr.11: 103113800-103113800 on Build GRCh38
rs927849195 Chr.11: 103113787-103113787 on Build GRCh38
rs759952846 Chr.11: 103113719-103113719 on Build GRCh38
rs200485999 Chr.11: 103113658-103113658 on Build GRCh38
rs1020246530 Chr.11: 103113688-103113688 on Build GRCh38
rs757693058 Chr.11: 103113727-103113727 on Build GRCh38
rs796264533 Chr.11: 103113976-103113978 on Build GRCh38
rs1041311789 Chr.11: 103113896-103113896 on Build GRCh38
rs779916760 Chr.11: 103113733-103113733 on Build GRCh38
rs763066231 Chr.11: 103113701-103113701 on Build GRCh38
rs549177489 Chr.11: 103113656-103113656 on Build GRCh38
rs765216105 Chr.11: 103113654-103113654 on Build GRCh38
rs997943933 Chr.11: 103113790-103113790 on Build GRCh38
rs543428081 Chr.11: 103114031-103114031 on Build GRCh38
rs778222081 Chr.11: 103113677-103113677 on Build GRCh38
rs536504758 Chr.11: 103113793-103113793 on Build GRCh38
rs756807806 Chr.11: 103113675-103113675 on Build GRCh38
rs769921141 Chr.11: 103113695-103113695 on Build GRCh38
rs915506622 Chr.11: 103113995-103113995 on Build GRCh38
rs748147524 Chr.11: 103114069-103114069 on Build GRCh38
rs778083906 Chr.11: 103113697-103113699 on Build GRCh38
rs373757155 Chr.11: 103113721-103113721 on Build GRCh38
rs886047558 Chr.11: 103113607-103113607 on Build GRCh38
rs762017411 Chr.11: 103113640-103113640 on Build GRCh38
rs551497110 Chr.11: 103113679-103113679 on Build GRCh38
rs1016807975 Chr.11: 103113876-103113876 on Build GRCh38
rs768760103 Chr.11: 103113628-103113628 on Build GRCh38
rs1006484042 Chr.11: 103113827-103113827 on Build GRCh38
rs982204422 Chr.11: 103113948-103113948 on Build GRCh38
rs773125998 Chr.11: 103113696-103113696 on Build GRCh38
rs528677847 Chr.11: 103113664-103113664 on Build GRCh38
rs12270130 Chr.11: 103113967-103113967 on Build GRCh38
rs114305658 Chr.11: 103113729-103113729 on Build GRCh38
rs972575271 Chr.11: 103113888-103113888 on Build GRCh38
rs574001441 Chr.11: 103114091-103114091 on Build GRCh38
rs756541802 Chr.11: 103113724-103113724 on Build GRCh38
rs1042990364 Chr.11: 103114008-103114008 on Build GRCh38
rs754831505 Chr.11: 103113755-103113755 on Build GRCh38
rs990632781 Chr.11: 103113801-103113801 on Build GRCh38
rs766629658 Chr.11: 103113706-103113706 on Build GRCh38
rs753256254 Chr.11: 103113723-103113723 on Build GRCh38
rs762193731 Chr.11: 103113789-103113794 on Build GRCh38
rs768081432 Chr.11: 103113669-103113669 on Build GRCh38
rs114066525 Chr.11: 103114070-103114070 on Build GRCh38
rs938014512 Chr.11: 103113984-103113984 on Build GRCh38
rs902297614 Chr.11: 103113762-103113762 on Build GRCh38
rs902662004 Chr.11: 103114027-103114027 on Build GRCh38
rs773169868 Chr.11: 103114072-103114072 on Build GRCh38
rs745569868 Chr.11: 103113678-103113678 on Build GRCh38
rs746836884 Chr.11: 103113684-103113684 on Build GRCh38
rs1042529479 Chr.11: 103113753-103113753 on Build GRCh38
rs571231065 Chr.11: 103113683-103113683 on Build GRCh38
rs749527696 Chr.11: 103113726-103113727 on Build GRCh38
rs367826430 Chr.11: 103113725-103113725 on Build GRCh38
rs776765795 Chr.11: 103113689-103113689 on Build GRCh38
rs116481300 Chr.11: 103113878-103113878 on Build GRCh38
rs377529350 Chr.11: 103113632-103113632 on Build GRCh38
rs573074084 Chr.11: 103114000-103114000 on Build GRCh38
rs755305630 Chr.11: 103113668-103113668 on Build GRCh38
rs768564016 Chr.11: 103113686-103113686 on Build GRCh38
rs185446871 Chr.11: 103113772-103113772 on Build GRCh38
rs200257097 Chr.11: 103113717-103113717 on Build GRCh38
rs747976646 Chr.11: 103113690-103113690 on Build GRCh38
rs773303282 Chr.11: 103114086-103114086 on Build GRCh38
rs760706534 Chr.11: 103113610-103113610 on Build GRCh38
rs901409589 Chr.11: 103113911-103113911 on Build GRCh38
rs751970677 Chr.11: 103113665-103113665 on Build GRCh38
rs772875964 Chr.11: 103113746-103113746 on Build GRCh38
rs749396289 Chr.11: 103114089-103114089 on Build GRCh38
rs573032534 Chr.11: 103113941-103113941 on Build GRCh38
rs1025471783 Chr.11: 103113899-103113899 on Build GRCh38
rs916034760 Chr.11: 103113806-103113811 on Build GRCh38
rs1043128048 Chr.11: 103113871-103113872 on Build GRCh38

View Full Product Details