Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104954662-104955168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP4
Entrez Gene ID
837
Gene Name
caspase 4
Gene Aliases
ICE(rel)II, ICEREL-II, ICH-2, Mih1, Mih1/TX, TX
UniGene
Hs.138378
Gene Chromosome Location
Chr.11: 104942866-104968598 on Build GRCh38
Transcripts
NM_001225.3 NM_033306.2 XM_011543019.1 XM_017018397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5448076 c.8-6T>G p.? Chr.11: 104955007-104955007 on Build GRCh38
685728 c.91G>C p.E31Q Chr.11: 104954918-104954918 on Build GRCh38
5371533 c.70G>A p.G24S Chr.11: 104954939-104954939 on Build GRCh38
685727 c.57A>T p.K19N Chr.11: 104954952-104954952 on Build GRCh38
3442848 c.40T>C p.L14L Chr.11: 104954969-104954969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371962072 Chr.11: 104954961-104954961 on Build GRCh38
rs925240829 Chr.11: 104954720-104954720 on Build GRCh38
rs765687371 Chr.11: 104954970-104954970 on Build GRCh38
rs947651320 Chr.11: 104955035-104955035 on Build GRCh38
rs751821857 Chr.11: 104954736-104954736 on Build GRCh38
rs540315323 Chr.11: 104955013-104955013 on Build GRCh38
rs112900780 Chr.11: 104954754-104954754 on Build GRCh38
rs746730933 Chr.11: 104954778-104954778 on Build GRCh38
rs756692795 Chr.11: 104954752-104954752 on Build GRCh38
rs142339077 Chr.11: 104955003-104955003 on Build GRCh38
rs750340377 Chr.11: 104954716-104954716 on Build GRCh38
rs754196122 Chr.11: 104954957-104954957 on Build GRCh38
rs376802358 Chr.11: 104954839-104954839 on Build GRCh38
rs777279432 Chr.11: 104955013-104955016 on Build GRCh38
rs369838930 Chr.11: 104954825-104954825 on Build GRCh38
rs367631972 Chr.11: 104954713-104954713 on Build GRCh38
rs55725009 Chr.11: 104954921-104954921 on Build GRCh38
rs752867454 Chr.11: 104954945-104954945 on Build GRCh38
rs760791999 Chr.11: 104954770-104954770 on Build GRCh38
rs764499117 Chr.11: 104954955-104954955 on Build GRCh38
rs778274568 Chr.11: 104954753-104954753 on Build GRCh38
rs112539250 Chr.11: 104955048-104955048 on Build GRCh38
rs776274459 Chr.11: 104954710-104954710 on Build GRCh38
rs903144597 Chr.11: 104955100-104955100 on Build GRCh38
rs372056674 Chr.11: 104954782-104954782 on Build GRCh38
rs145822235 Chr.11: 104954911-104954911 on Build GRCh38
rs781670566 Chr.11: 104954738-104954738 on Build GRCh38
rs779092072 Chr.11: 104954778-104954778 on Build GRCh38
rs770707997 Chr.11: 104955014-104955014 on Build GRCh38
rs898401067 Chr.11: 104955129-104955129 on Build GRCh38
rs1052419785 Chr.11: 104954926-104954926 on Build GRCh38
rs536340503 Chr.11: 104954800-104954800 on Build GRCh38
rs2298768 Chr.11: 104954853-104954853 on Build GRCh38
rs555193787 Chr.11: 104954999-104955000 on Build GRCh38
rs757340578 Chr.11: 104954729-104954730 on Build GRCh38
rs372646731 Chr.11: 104954885-104954885 on Build GRCh38
rs775625488 Chr.11: 104955045-104955045 on Build GRCh38
rs368133405 Chr.11: 104954776-104954776 on Build GRCh38
rs754450853 Chr.11: 104954978-104954978 on Build GRCh38
rs200174319 Chr.11: 104955002-104955002 on Build GRCh38
rs561133700 Chr.11: 104954951-104954951 on Build GRCh38
rs141431200 Chr.11: 104954835-104954835 on Build GRCh38
rs891066126 Chr.11: 104954784-104954784 on Build GRCh38
rs55767261 Chr.11: 104954931-104954931 on Build GRCh38
rs758585868 Chr.11: 104954728-104954728 on Build GRCh38
rs766500480 Chr.11: 104954735-104954735 on Build GRCh38
rs11539160 Chr.11: 104954965-104954965 on Build GRCh38
rs767589687 Chr.11: 104954934-104954934 on Build GRCh38
rs368899059 Chr.11: 104954871-104954871 on Build GRCh38
rs1039636249 Chr.11: 104955070-104955070 on Build GRCh38
rs774183391 Chr.11: 104955026-104955026 on Build GRCh38
rs369105517 Chr.11: 104954894-104954894 on Build GRCh38
rs772291949 Chr.11: 104954881-104954881 on Build GRCh38
rs56226603 Chr.11: 104954870-104954870 on Build GRCh38
rs1030418546 Chr.11: 104954846-104954846 on Build GRCh38
rs150997782 Chr.11: 104954812-104954812 on Build GRCh38
rs1018582802 Chr.11: 104954898-104954898 on Build GRCh38
rs757745508 Chr.11: 104954876-104954876 on Build GRCh38
rs777635897 Chr.11: 104954915-104954915 on Build GRCh38
rs929648640 Chr.11: 104954969-104954969 on Build GRCh38
rs997533999 Chr.11: 104955108-104955108 on Build GRCh38
rs764793096 Chr.11: 104954982-104954982 on Build GRCh38
rs371778039 Chr.11: 104954737-104954737 on Build GRCh38
rs766625268 Chr.11: 104954827-104954827 on Build GRCh38
rs551111748 Chr.11: 104954685-104954685 on Build GRCh38
rs767610532 Chr.11: 104954860-104954860 on Build GRCh38
rs765247554 Chr.11: 104954714-104954714 on Build GRCh38
rs778041398 Chr.11: 104954873-104954873 on Build GRCh38
rs56116531 Chr.11: 104955001-104955001 on Build GRCh38
rs192509490 Chr.11: 104954859-104954859 on Build GRCh38
rs1041736922 Chr.11: 104955055-104955055 on Build GRCh38
rs746168706 Chr.11: 104954829-104954830 on Build GRCh38
rs759553577 Chr.11: 104954933-104954933 on Build GRCh38
rs754353364 Chr.11: 104954874-104954874 on Build GRCh38
rs746480853 Chr.11: 104954884-104954884 on Build GRCh38
rs907327664 Chr.11: 104954686-104954686 on Build GRCh38
rs776317087 Chr.11: 104954826-104954826 on Build GRCh38
rs747870694 Chr.11: 104954992-104954992 on Build GRCh38
rs572881726 Chr.11: 104954813-104954813 on Build GRCh38
rs376811221 Chr.11: 104954768-104954768 on Build GRCh38
rs745747780 Chr.11: 104955032-104955032 on Build GRCh38
rs759840645 Chr.11: 104954845-104954845 on Build GRCh38
rs763961152 Chr.11: 104954698-104954698 on Build GRCh38
rs1045092904 Chr.11: 104954715-104954715 on Build GRCh38
rs772058463 Chr.11: 104955037-104955037 on Build GRCh38
rs750903321 Chr.11: 104954972-104954972 on Build GRCh38
rs747774766 Chr.11: 104954905-104954905 on Build GRCh38
rs146623912 Chr.11: 104954755-104954755 on Build GRCh38
rs375442545 Chr.11: 104954822-104954822 on Build GRCh38
rs775968861 Chr.11: 104954982-104954983 on Build GRCh38
rs769724709 Chr.11: 104954909-104954909 on Build GRCh38
rs1801319 Chr.11: 104954922-104954922 on Build GRCh38
rs567654660 Chr.11: 104954742-104954742 on Build GRCh38
rs558999956 Chr.11: 104955003-104955004 on Build GRCh38

View Full Product Details