Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104997261-104997766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP5
Entrez Gene ID
838
Gene Name
caspase 5
Gene Aliases
ICE(rel)III, ICEREL-III, ICH-3
UniGene
Hs.213327
Gene Chromosome Location
Chr.11: 104994240-105023902 on Build GRCh38
Transcripts
NM_001136109.1 NM_001136110.1 NM_001136112.1 NM_004347.3 NR_024239.1 NR_036562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1732768 c.1152A>T p.R384S Chr.11: 104997476-104997476 on Build GRCh38
1199644 c.1240C>T p.R414W Chr.11: 104997388-104997388 on Build GRCh38
1199645 c.1153C>T p.R385W Chr.11: 104997388-104997388 on Build GRCh38
2109098 c.1109G>C p.C370S Chr.11: 104997432-104997432 on Build GRCh38
5035824 c.1049-8C>T p.? Chr.11: 104997500-104997500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009361985 Chr.11: 104997342-104997342 on Build GRCh38
rs45439695 Chr.11: 104997507-104997507 on Build GRCh38
rs765997455 Chr.11: 104997446-104997446 on Build GRCh38
rs901372625 Chr.11: 104997426-104997426 on Build GRCh38
rs764297671 Chr.11: 104997476-104997476 on Build GRCh38
rs945012253 Chr.11: 104997572-104997572 on Build GRCh38
rs45620437 Chr.11: 104997491-104997491 on Build GRCh38
rs757545547 Chr.11: 104997394-104997394 on Build GRCh38
rs780271252 Chr.11: 104997495-104997495 on Build GRCh38
rs202025207 Chr.11: 104997485-104997485 on Build GRCh38
rs12222985 Chr.11: 104997621-104997621 on Build GRCh38
rs751328393 Chr.11: 104997355-104997355 on Build GRCh38
rs200585583 Chr.11: 104997484-104997484 on Build GRCh38
rs753698357 Chr.11: 104997593-104997593 on Build GRCh38
rs764071176 Chr.11: 104997513-104997513 on Build GRCh38
rs753830245 Chr.11: 104997479-104997479 on Build GRCh38
rs757099592 Chr.11: 104997636-104997636 on Build GRCh38
rs759361634 Chr.11: 104997449-104997449 on Build GRCh38
rs1044222152 Chr.11: 104997625-104997625 on Build GRCh38
rs550969473 Chr.11: 104997681-104997681 on Build GRCh38
rs143946090 Chr.11: 104997467-104997467 on Build GRCh38
rs747418836 Chr.11: 104997411-104997411 on Build GRCh38
rs767502229 Chr.11: 104997387-104997387 on Build GRCh38
rs200749075 Chr.11: 104997378-104997378 on Build GRCh38
rs375005378 Chr.11: 104997526-104997526 on Build GRCh38
rs769404945 Chr.11: 104997431-104997431 on Build GRCh38
rs370403880 Chr.11: 104997361-104997361 on Build GRCh38
rs371986422 Chr.11: 104997519-104997519 on Build GRCh38
rs565128873 Chr.11: 104997520-104997520 on Build GRCh38
rs747618230 Chr.11: 104997502-104997502 on Build GRCh38
rs889944029 Chr.11: 104997630-104997630 on Build GRCh38
rs766148367 Chr.11: 104997516-104997516 on Build GRCh38
rs900900291 Chr.11: 104997610-104997610 on Build GRCh38
rs767467723 Chr.11: 104997373-104997373 on Build GRCh38
rs201509715 Chr.11: 104997493-104997493 on Build GRCh38
rs772831264 Chr.11: 104997510-104997510 on Build GRCh38
rs778617757 Chr.11: 104997660-104997660 on Build GRCh38
rs45458695 Chr.11: 104997445-104997445 on Build GRCh38
rs190703418 Chr.11: 104997472-104997472 on Build GRCh38
rs763960854 Chr.11: 104997471-104997471 on Build GRCh38
rs372078561 Chr.11: 104997529-104997529 on Build GRCh38
rs1024359516 Chr.11: 104997691-104997691 on Build GRCh38
rs753779309 Chr.11: 104997544-104997544 on Build GRCh38
rs547070785 Chr.11: 104997447-104997447 on Build GRCh38
rs998764534 Chr.11: 104997626-104997626 on Build GRCh38
rs377357775 Chr.11: 104997344-104997344 on Build GRCh38
rs373942063 Chr.11: 104997482-104997482 on Build GRCh38
rs752540426 Chr.11: 104997377-104997377 on Build GRCh38
rs182955793 Chr.11: 104997527-104997527 on Build GRCh38
rs776071040 Chr.11: 104997408-104997408 on Build GRCh38
rs1014085032 Chr.11: 104997301-104997301 on Build GRCh38
rs554474023 Chr.11: 104997647-104997647 on Build GRCh38
rs867070721 Chr.11: 104997388-104997388 on Build GRCh38
rs200261759 Chr.11: 104997521-104997521 on Build GRCh38
rs577121203 Chr.11: 104997731-104997731 on Build GRCh38
rs1042175073 Chr.11: 104997605-104997605 on Build GRCh38
rs978436172 Chr.11: 104997338-104997338 on Build GRCh38
rs893565011 Chr.11: 104997317-104997317 on Build GRCh38
rs769012693 Chr.11: 104997608-104997608 on Build GRCh38
rs752497919 Chr.11: 104997375-104997375 on Build GRCh38
rs779458736 Chr.11: 104997397-104997397 on Build GRCh38
rs755908747 Chr.11: 104997466-104997466 on Build GRCh38
rs199566482 Chr.11: 104997522-104997522 on Build GRCh38
rs770390346 Chr.11: 104997515-104997515 on Build GRCh38
rs1049887585 Chr.11: 104997627-104997627 on Build GRCh38
rs1000142979 Chr.11: 104997655-104997655 on Build GRCh38
rs1014510010 Chr.11: 104997540-104997540 on Build GRCh38
rs748822565 Chr.11: 104997514-104997514 on Build GRCh38
rs753829255 Chr.11: 104997536-104997537 on Build GRCh38
rs969928170 Chr.11: 104997331-104997331 on Build GRCh38
rs752690490 Chr.11: 104997471-104997471 on Build GRCh38
rs746213106 Chr.11: 104997398-104997398 on Build GRCh38
rs542455828 Chr.11: 104997313-104997313 on Build GRCh38
rs994098873 Chr.11: 104997685-104997685 on Build GRCh38
rs772515825 Chr.11: 104997406-104997406 on Build GRCh38
rs772593433 Chr.11: 104997434-104997434 on Build GRCh38

View Full Product Details