Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105031079-105031584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP1
Entrez Gene ID
834
Gene Name
caspase 1
Gene Aliases
ICE, IL1BC, P45
UniGene
Hs.2490
Gene Chromosome Location
Chr.11: 105025508-105035591 on Build GRCh38
Transcripts
NM_001223.4 NM_001257118.2 NM_001257119.2 NM_033292.3 NM_033293.3 NM_033294.3 NM_033295.3 XM_017018393.1 XM_017018394.1 XM_017018395.1 XM_017018396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283928 c.453G>A p.E151E Chr.11: 105031165-105031165 on Build GRCh38
5817834 c.416A>G p.E139G Chr.11: 105031202-105031202 on Build GRCh38
6294244 c.446C>A p.S149* Chr.11: 105031172-105031172 on Build GRCh38
5528152 c.429G>A p.R143R Chr.11: 105031189-105031189 on Build GRCh38
7129727 c.372C>A p.P124P Chr.11: 105031246-105031246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367931430 Chr.11: 105031318-105031318 on Build GRCh38
rs948310409 Chr.11: 105031414-105031414 on Build GRCh38
rs902927434 Chr.11: 105031408-105031408 on Build GRCh38
rs189373708 Chr.11: 105031441-105031441 on Build GRCh38
rs1699076 Chr.11: 105031115-105031115 on Build GRCh38
rs375010411 Chr.11: 105031283-105031283 on Build GRCh38
rs1000787732 Chr.11: 105031461-105031461 on Build GRCh38
rs564568441 Chr.11: 105031232-105031232 on Build GRCh38
rs914212069 Chr.11: 105031403-105031403 on Build GRCh38
rs773356480 Chr.11: 105031241-105031241 on Build GRCh38
rs766086112 Chr.11: 105031291-105031291 on Build GRCh38
rs781140786 Chr.11: 105031316-105031316 on Build GRCh38
rs750278836 Chr.11: 105031179-105031179 on Build GRCh38
rs1006830180 Chr.11: 105031556-105031556 on Build GRCh38
rs3203616 Chr.11: 105031208-105031208 on Build GRCh38
rs549437453 Chr.11: 105031380-105031380 on Build GRCh38
rs770191842 Chr.11: 105031141-105031141 on Build GRCh38
rs748227609 Chr.11: 105031317-105031317 on Build GRCh38
rs755154041 Chr.11: 105031220-105031220 on Build GRCh38
rs761608796 Chr.11: 105031113-105031113 on Build GRCh38
rs997621124 Chr.11: 105031175-105031175 on Build GRCh38
rs755897950 Chr.11: 105031263-105031263 on Build GRCh38
rs529194780 Chr.11: 105031404-105031404 on Build GRCh38
rs764985999 Chr.11: 105031279-105031279 on Build GRCh38
rs753570094 Chr.11: 105031172-105031172 on Build GRCh38
rs1010519081 Chr.11: 105031510-105031510 on Build GRCh38
rs773173695 Chr.11: 105031291-105031292 on Build GRCh38
rs774987068 Chr.11: 105031253-105031253 on Build GRCh38
rs746909649 Chr.11: 105031206-105031206 on Build GRCh38
rs762850041 Chr.11: 105031300-105031300 on Build GRCh38
rs766783673 Chr.11: 105031442-105031442 on Build GRCh38
rs1016243644 Chr.11: 105031231-105031231 on Build GRCh38
rs763505053 Chr.11: 105031171-105031171 on Build GRCh38
rs749149374 Chr.11: 105031405-105031405 on Build GRCh38
rs551204273 Chr.11: 105031511-105031511 on Build GRCh38
rs562853587 Chr.11: 105031446-105031446 on Build GRCh38
rs865793662 Chr.11: 105031260-105031260 on Build GRCh38
rs200659098 Chr.11: 105031153-105031153 on Build GRCh38
rs973834616 Chr.11: 105031348-105031348 on Build GRCh38
rs1026435281 Chr.11: 105031501-105031501 on Build GRCh38
rs199932541 Chr.11: 105031248-105031248 on Build GRCh38
rs758330498 Chr.11: 105031290-105031290 on Build GRCh38
rs763263420 Chr.11: 105031151-105031151 on Build GRCh38
rs774705591 Chr.11: 105031463-105031463 on Build GRCh38
rs955311366 Chr.11: 105031287-105031287 on Build GRCh38
rs201549047 Chr.11: 105031149-105031149 on Build GRCh38
rs892878435 Chr.11: 105031138-105031138 on Build GRCh38
rs776030696 Chr.11: 105031272-105031272 on Build GRCh38
rs186737339 Chr.11: 105031421-105031421 on Build GRCh38
rs3203615 Chr.11: 105031233-105031233 on Build GRCh38
rs981267806 Chr.11: 105031340-105031340 on Build GRCh38
rs991169367 Chr.11: 105031398-105031398 on Build GRCh38
rs955497970 Chr.11: 105031541-105031541 on Build GRCh38
rs760286520 Chr.11: 105031167-105031167 on Build GRCh38
rs1030579352 Chr.11: 105031177-105031177 on Build GRCh38
rs970770370 Chr.11: 105031512-105031512 on Build GRCh38
rs769532325 Chr.11: 105031197-105031199 on Build GRCh38
rs985154923 Chr.11: 105031295-105031295 on Build GRCh38
rs571295206 Chr.11: 105031549-105031549 on Build GRCh38
rs763711195 Chr.11: 105031270-105031270 on Build GRCh38
rs751558009 Chr.11: 105031310-105031310 on Build GRCh38
rs770034012 Chr.11: 105031235-105031235 on Build GRCh38
rs899190560 Chr.11: 105031467-105031467 on Build GRCh38
rs45500199 Chr.11: 105031324-105031324 on Build GRCh38
rs981274352 Chr.11: 105031533-105031533 on Build GRCh38
rs199570136 Chr.11: 105031251-105031251 on Build GRCh38
rs761368441 Chr.11: 105031278-105031278 on Build GRCh38
rs1051842443 Chr.11: 105031438-105031438 on Build GRCh38
rs1045029850 Chr.11: 105031110-105031110 on Build GRCh38
rs1031016918 Chr.11: 105031157-105031157 on Build GRCh38
rs1029485082 Chr.11: 105031537-105031537 on Build GRCh38
rs369343501 Chr.11: 105031130-105031130 on Build GRCh38
rs529939902 Chr.11: 105031378-105031378 on Build GRCh38
rs370258690 Chr.11: 105031120-105031120 on Build GRCh38
rs774658840 Chr.11: 105031160-105031160 on Build GRCh38
rs150956450 Chr.11: 105031498-105031498 on Build GRCh38
rs367873337 Chr.11: 105031174-105031174 on Build GRCh38
rs3203614 Chr.11: 105031236-105031236 on Build GRCh38
rs754891981 Chr.11: 105031312-105031312 on Build GRCh38
rs541150316 Chr.11: 105031196-105031196 on Build GRCh38
rs781107669 Chr.11: 105031227-105031227 on Build GRCh38
rs1035497038 Chr.11: 105031543-105031543 on Build GRCh38
rs143095678 Chr.11: 105031247-105031247 on Build GRCh38
rs746610950 Chr.11: 105031185-105031185 on Build GRCh38
rs149065878 Chr.11: 105031189-105031189 on Build GRCh38

View Full Product Details