Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105035015-105035513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP1
Entrez Gene ID
834
Gene Name
caspase 1
Gene Aliases
ICE, IL1BC, P45
UniGene
Hs.2490
Gene Chromosome Location
Chr.11: 105025508-105035591 on Build GRCh38
Transcripts
NM_001223.4 NM_001257118.2 NM_001257119.2 NM_033292.3 NM_033293.3 NM_033294.3 NM_033295.3 XM_017018393.1 XM_017018394.1 XM_017018395.1 XM_017018396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620544 c.5C>T p.A2V Chr.11: 105035109-105035109 on Build GRCh38
7239892 c.6C>A p.A2A Chr.11: 105035108-105035108 on Build GRCh38
922453 c.4delG p.A2fs*5 Chr.11: 105035110-105035110 on Build GRCh38
6580903 c.1_7+5delATGGCCGGTGAG p.? Chr.11: 105035102-105035125 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893738549 Chr.11: 105035147-105035147 on Build GRCh38
rs374423061 Chr.11: 105035176-105035176 on Build GRCh38
rs778925999 Chr.11: 105035089-105035089 on Build GRCh38
rs767791240 Chr.11: 105035077-105035078 on Build GRCh38
rs998260496 Chr.11: 105035474-105035474 on Build GRCh38
rs976842960 Chr.11: 105035266-105035266 on Build GRCh38
rs766901394 Chr.11: 105035126-105035126 on Build GRCh38
rs560806044 Chr.11: 105035455-105035455 on Build GRCh38
rs941959206 Chr.11: 105035434-105035434 on Build GRCh38
rs775848383 Chr.11: 105035097-105035097 on Build GRCh38
rs950061207 Chr.11: 105035087-105035087 on Build GRCh38
rs765741146 Chr.11: 105035109-105035109 on Build GRCh38
rs531602109 Chr.11: 105035180-105035180 on Build GRCh38
rs945207719 Chr.11: 105035379-105035379 on Build GRCh38
rs749042137 Chr.11: 105035076-105035076 on Build GRCh38
rs3809024 Chr.11: 105035212-105035212 on Build GRCh38
rs570568715 Chr.11: 105035241-105035241 on Build GRCh38
rs373997855 Chr.11: 105035158-105035158 on Build GRCh38
rs971837565 Chr.11: 105035287-105035287 on Build GRCh38
rs1028500924 Chr.11: 105035207-105035207 on Build GRCh38
rs1002589916 Chr.11: 105035192-105035192 on Build GRCh38
rs969363290 Chr.11: 105035232-105035232 on Build GRCh38
rs553961449 Chr.11: 105035224-105035224 on Build GRCh38
rs200219743 Chr.11: 105035131-105035131 on Build GRCh38
rs777404332 Chr.11: 105035059-105035059 on Build GRCh38
rs375305868 Chr.11: 105035345-105035345 on Build GRCh38
rs1021912563 Chr.11: 105035226-105035226 on Build GRCh38
rs541884770 Chr.11: 105035349-105035349 on Build GRCh38
rs1049384046 Chr.11: 105035476-105035476 on Build GRCh38
rs745959532 Chr.11: 105035092-105035092 on Build GRCh38
rs757290519 Chr.11: 105035077-105035077 on Build GRCh38
rs921311602 Chr.11: 105035268-105035268 on Build GRCh38
rs927388395 Chr.11: 105035296-105035296 on Build GRCh38
rs752439028 Chr.11: 105035129-105035129 on Build GRCh38
rs781376071 Chr.11: 105035462-105035462 on Build GRCh38
rs889549168 Chr.11: 105035488-105035488 on Build GRCh38
rs917282465 Chr.11: 105035086-105035086 on Build GRCh38
rs1013036435 Chr.11: 105035213-105035213 on Build GRCh38
rs747169291 Chr.11: 105035101-105035101 on Build GRCh38
rs556296093 Chr.11: 105035324-105035324 on Build GRCh38
rs146669228 Chr.11: 105035203-105035203 on Build GRCh38
rs116982264 Chr.11: 105035364-105035364 on Build GRCh38
rs376222697 Chr.11: 105035045-105035045 on Build GRCh38
rs977473022 Chr.11: 105035242-105035242 on Build GRCh38
rs973003104 Chr.11: 105035257-105035257 on Build GRCh38
rs772034057 Chr.11: 105035096-105035096 on Build GRCh38
rs180804835 Chr.11: 105035163-105035163 on Build GRCh38
rs924625305 Chr.11: 105035244-105035244 on Build GRCh38
rs923114617 Chr.11: 105035083-105035083 on Build GRCh38
rs190709302 Chr.11: 105035247-105035247 on Build GRCh38
rs150005656 Chr.11: 105035107-105035107 on Build GRCh38
rs45541843 Chr.11: 105035181-105035181 on Build GRCh38
rs944086831 Chr.11: 105035063-105035066 on Build GRCh38
rs576166510 Chr.11: 105035330-105035330 on Build GRCh38
rs755743530 Chr.11: 105035148-105035148 on Build GRCh38
rs980092400 Chr.11: 105035288-105035288 on Build GRCh38
rs773692055 Chr.11: 105035112-105035112 on Build GRCh38
rs1006149852 Chr.11: 105035221-105035221 on Build GRCh38
rs540553575 Chr.11: 105035432-105035432 on Build GRCh38
rs199725474 Chr.11: 105035075-105035075 on Build GRCh38
rs370647445 Chr.11: 105035119-105035119 on Build GRCh38
rs368205186 Chr.11: 105035162-105035162 on Build GRCh38
rs917502646 Chr.11: 105035274-105035274 on Build GRCh38
rs753028520 Chr.11: 105035127-105035128 on Build GRCh38
rs777006645 Chr.11: 105035108-105035108 on Build GRCh38
rs953779444 Chr.11: 105035214-105035214 on Build GRCh38
rs575102051 Chr.11: 105035378-105035378 on Build GRCh38

View Full Product Details