Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111298181-111298685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
UniGene
Hs.729225
Gene Chromosome Location
Chr.11: 111293389-111305048 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302646.1 NM_001302647.1 NM_001302648.1 NM_207429.3 NM_001271458.1 XM_017017196.1

Gene Symbol
COLCA2
Entrez Gene ID
120376
Gene Name
colorectal cancer associated 2
Gene Aliases
C11orf93, CASC13, LOH11CR1G
UniGene
Hs.31409
Gene Chromosome Location
Chr.11: 111298517-111308735 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302646.1 NM_001302647.1 NM_001302648.1 NM_207429.3 NM_001271458.1 XM_017017196.1

SNP Detail

SNP ID Chromosome Location
rs1039548823 Chr.11: 111298615-111298615 on Build GRCh38
rs570067149 Chr.11: 111298483-111298483 on Build GRCh38
rs1006831497 Chr.11: 111298334-111298334 on Build GRCh38
rs1015430336 Chr.11: 111298572-111298572 on Build GRCh38
rs564071302 Chr.11: 111298595-111298595 on Build GRCh38
rs938471068 Chr.11: 111298380-111298380 on Build GRCh38
rs10244 Chr.11: 111298385-111298385 on Build GRCh38
rs370780567 Chr.11: 111298624-111298624 on Build GRCh38
rs954820896 Chr.11: 111298218-111298218 on Build GRCh38
rs946834430 Chr.11: 111298541-111298541 on Build GRCh38
rs376671617 Chr.11: 111298494-111298494 on Build GRCh38
rs1008953572 Chr.11: 111298545-111298545 on Build GRCh38
rs904815264 Chr.11: 111298487-111298487 on Build GRCh38
rs972643083 Chr.11: 111298282-111298282 on Build GRCh38
rs1042921626 Chr.11: 111298319-111298319 on Build GRCh38
rs910064938 Chr.11: 111298392-111298392 on Build GRCh38
rs530117684 Chr.11: 111298395-111298395 on Build GRCh38
rs927019627 Chr.11: 111298372-111298372 on Build GRCh38
rs192920581 Chr.11: 111298455-111298455 on Build GRCh38
rs913860895 Chr.11: 111298496-111298497 on Build GRCh38
rs889186979 Chr.11: 111298546-111298546 on Build GRCh38
rs891936825 Chr.11: 111298397-111298397 on Build GRCh38
rs969502433 Chr.11: 111298421-111298422 on Build GRCh38
rs1052451779 Chr.11: 111298481-111298481 on Build GRCh38
rs776236691 Chr.11: 111298222-111298222 on Build GRCh38
rs997894903 Chr.11: 111298353-111298353 on Build GRCh38
rs886744214 Chr.11: 111298658-111298658 on Build GRCh38
rs1018228553 Chr.11: 111298342-111298342 on Build GRCh38
rs1029023616 Chr.11: 111298206-111298206 on Build GRCh38
rs919844731 Chr.11: 111298292-111298292 on Build GRCh38
rs942540207 Chr.11: 111298577-111298577 on Build GRCh38
rs763485596 Chr.11: 111298497-111298497 on Build GRCh38
rs752858632 Chr.11: 111298375-111298376 on Build GRCh38
rs550581412 Chr.11: 111298477-111298477 on Build GRCh38
rs1010821827 Chr.11: 111298426-111298426 on Build GRCh38
rs904327860 Chr.11: 111298320-111298320 on Build GRCh38
rs1028973022 Chr.11: 111298498-111298498 on Build GRCh38
rs951164855 Chr.11: 111298382-111298382 on Build GRCh38
rs961514839 Chr.11: 111298268-111298268 on Build GRCh38
rs75721472 Chr.11: 111298293-111298293 on Build GRCh38
rs537413560 Chr.11: 111298500-111298500 on Build GRCh38
rs1011371783 Chr.11: 111298315-111298315 on Build GRCh38
rs987019471 Chr.11: 111298245-111298245 on Build GRCh38
rs188072336 Chr.11: 111298347-111298347 on Build GRCh38
rs912373183 Chr.11: 111298261-111298261 on Build GRCh38
rs548719297 Chr.11: 111298374-111298374 on Build GRCh38

View Full Product Details