Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111299130-111299630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
UniGene
Hs.729225
Gene Chromosome Location
Chr.11: 111293389-111305048 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302646.1 NM_001302647.1 NM_001302648.1 NM_207429.3 NM_001136105.2 NM_001271457.1 NM_001271458.1 XM_017017196.1 XM_017017199.1

Gene Symbol
COLCA2
Entrez Gene ID
120376
Gene Name
colorectal cancer associated 2
Gene Aliases
C11orf93, CASC13, LOH11CR1G
UniGene
Hs.31409
Gene Chromosome Location
Chr.11: 111298517-111308735 on Build GRCh38
Transcripts
NM_001302644.1 NM_001302645.1 NM_001302646.1 NM_001302647.1 NM_001302648.1 NM_207429.3 NM_001136105.2 NM_001271457.1 NM_001271458.1 XM_017017196.1 XM_017017199.1

SNP Detail

SNP ID Chromosome Location
rs896265585 Chr.11: 111299358-111299358 on Build GRCh38
rs895604780 Chr.11: 111299499-111299499 on Build GRCh38
rs903992512 Chr.11: 111299370-111299370 on Build GRCh38
rs908402822 Chr.11: 111299563-111299563 on Build GRCh38
rs548508768 Chr.11: 111299248-111299248 on Build GRCh38
rs1049118874 Chr.11: 111299462-111299462 on Build GRCh38
rs1056055877 Chr.11: 111299286-111299286 on Build GRCh38
rs968290810 Chr.11: 111299175-111299175 on Build GRCh38
rs953545491 Chr.11: 111299189-111299189 on Build GRCh38
rs986297415 Chr.11: 111299190-111299190 on Build GRCh38
rs949915760 Chr.11: 111299554-111299554 on Build GRCh38
rs566952852 Chr.11: 111299222-111299222 on Build GRCh38
rs926673662 Chr.11: 111299464-111299464 on Build GRCh38
rs912025303 Chr.11: 111299405-111299405 on Build GRCh38
rs895666734 Chr.11: 111299290-111299290 on Build GRCh38
rs546981072 Chr.11: 111299489-111299489 on Build GRCh38
rs1020209060 Chr.11: 111299348-111299348 on Build GRCh38
rs944977099 Chr.11: 111299212-111299212 on Build GRCh38
rs530946959 Chr.11: 111299347-111299347 on Build GRCh38
rs972692592 Chr.11: 111299441-111299441 on Build GRCh38
rs953162246 Chr.11: 111299385-111299385 on Build GRCh38
rs976090169 Chr.11: 111299231-111299231 on Build GRCh38
rs1042023054 Chr.11: 111299580-111299580 on Build GRCh38
rs903143448 Chr.11: 111299612-111299612 on Build GRCh38
rs1036637398 Chr.11: 111299215-111299215 on Build GRCh38
rs937709826 Chr.11: 111299332-111299332 on Build GRCh38
rs552587809 Chr.11: 111299419-111299419 on Build GRCh38
rs908337915 Chr.11: 111299276-111299276 on Build GRCh38
rs570887389 Chr.11: 111299472-111299472 on Build GRCh38
rs978446202 Chr.11: 111299158-111299158 on Build GRCh38
rs919522275 Chr.11: 111299449-111299449 on Build GRCh38
rs930621464 Chr.11: 111299461-111299461 on Build GRCh38
rs1032716417 Chr.11: 111299365-111299365 on Build GRCh38
rs925706554 Chr.11: 111299161-111299161 on Build GRCh38
rs986297905 Chr.11: 111299394-111299394 on Build GRCh38
rs11213823 Chr.11: 111299265-111299265 on Build GRCh38
rs983527412 Chr.11: 111299545-111299545 on Build GRCh38
rs763908474 Chr.11: 111299462-111299462 on Build GRCh38
rs962542702 Chr.11: 111299564-111299564 on Build GRCh38
rs1001354288 Chr.11: 111299269-111299269 on Build GRCh38
rs1001133494 Chr.11: 111299373-111299373 on Build GRCh38
rs1014558788 Chr.11: 111299317-111299317 on Build GRCh38
rs753502481 Chr.11: 111299155-111299155 on Build GRCh38
rs999856620 Chr.11: 111299360-111299360 on Build GRCh38
rs941296289 Chr.11: 111299277-111299277 on Build GRCh38
rs912133558 Chr.11: 111299211-111299211 on Build GRCh38
rs938052040 Chr.11: 111299466-111299466 on Build GRCh38
rs919225472 Chr.11: 111299237-111299237 on Build GRCh38
rs373839670 Chr.11: 111299241-111299241 on Build GRCh38
rs966976625 Chr.11: 111299349-111299349 on Build GRCh38

View Full Product Details