Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125903076-125903616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX25
Entrez Gene ID
29118
Gene Name
DEAD-box helicase 25
Gene Aliases
GRTH
UniGene
Hs.420263
Gene Chromosome Location
Chr.11: 125903275-125923110 on Build GRCh38
Transcripts
XM_017017626.1 NM_001271985.1 NM_031307.3 XM_005271687.3 XM_005271688.3

Gene Symbol
PUS3
Entrez Gene ID
83480
Gene Name
pseudouridylate synthase 3
Gene Aliases
2610020J05Rik, FKSG32
UniGene
Hs.660922
Gene Chromosome Location
Chr.11: 125893485-125903774 on Build GRCh38
Transcripts
XM_017017626.1 NM_001271985.1 NM_031307.3 XM_005271687.3 XM_005271688.3

SNP Detail

SNP ID Chromosome Location
rs571079 Chr.11: 125903377-125903377 on Build GRCh38
rs922479270 Chr.11: 125903360-125903360 on Build GRCh38
rs775105908 Chr.11: 125903135-125903135 on Build GRCh38
rs909873916 Chr.11: 125903119-125903119 on Build GRCh38
rs964338085 Chr.11: 125903430-125903430 on Build GRCh38
rs547232 Chr.11: 125903104-125903104 on Build GRCh38
rs937978976 Chr.11: 125903295-125903295 on Build GRCh38
rs369095457 Chr.11: 125903346-125903346 on Build GRCh38
rs564173799 Chr.11: 125903229-125903229 on Build GRCh38
rs545722551 Chr.11: 125903356-125903356 on Build GRCh38
rs569262300 Chr.11: 125903561-125903561 on Build GRCh38
rs975424980 Chr.11: 125903348-125903348 on Build GRCh38
rs975994181 Chr.11: 125903192-125903192 on Build GRCh38
rs111725242 Chr.11: 125903164-125903164 on Build GRCh38
rs922829441 Chr.11: 125903232-125903232 on Build GRCh38
rs764158546 Chr.11: 125903190-125903190 on Build GRCh38
rs1025805753 Chr.11: 125903303-125903303 on Build GRCh38
rs113827647 Chr.11: 125903552-125903569 on Build GRCh38
rs1012561738 Chr.11: 125903271-125903271 on Build GRCh38
rs975307362 Chr.11: 125903187-125903187 on Build GRCh38
rs948354916 Chr.11: 125903483-125903483 on Build GRCh38
rs140951817 Chr.11: 125903335-125903335 on Build GRCh38
rs192555083 Chr.11: 125903485-125903485 on Build GRCh38
rs578151432 Chr.11: 125903347-125903347 on Build GRCh38
rs1054194909 Chr.11: 125903470-125903470 on Build GRCh38
rs537073873 Chr.11: 125903501-125903504 on Build GRCh38
rs776657309 Chr.11: 125903551-125903552 on Build GRCh38
rs915673655 Chr.11: 125903473-125903473 on Build GRCh38
rs935762495 Chr.11: 125903253-125903253 on Build GRCh38
rs1005846330 Chr.11: 125903359-125903359 on Build GRCh38
rs984649140 Chr.11: 125903113-125903113 on Build GRCh38
rs924463153 Chr.11: 125903446-125903446 on Build GRCh38
rs761642281 Chr.11: 125903256-125903256 on Build GRCh38
rs1047791338 Chr.11: 125903527-125903527 on Build GRCh38
rs977783378 Chr.11: 125903433-125903433 on Build GRCh38
rs1030308674 Chr.11: 125903481-125903481 on Build GRCh38
rs946711739 Chr.11: 125903265-125903265 on Build GRCh38
rs79013855 Chr.11: 125903523-125903523 on Build GRCh38
rs563641629 Chr.11: 125903307-125903307 on Build GRCh38
rs550332389 Chr.11: 125903240-125903240 on Build GRCh38
rs1005621072 Chr.11: 125903342-125903342 on Build GRCh38
rs12290595 Chr.11: 125903099-125903099 on Build GRCh38
rs186664493 Chr.11: 125903453-125903453 on Build GRCh38
rs571779383 Chr.11: 125903482-125903482 on Build GRCh38
rs988264082 Chr.11: 125903243-125903243 on Build GRCh38
rs533443763 Chr.11: 125903236-125903236 on Build GRCh38
rs755623964 Chr.11: 125903519-125903519 on Build GRCh38
rs781094080 Chr.11: 125903294-125903294 on Build GRCh38
rs534570905 Chr.11: 125903167-125903167 on Build GRCh38
rs964223989 Chr.11: 125903184-125903184 on Build GRCh38
rs549113400 Chr.11: 125903191-125903191 on Build GRCh38
rs896345371 Chr.11: 125903312-125903312 on Build GRCh38
rs984393199 Chr.11: 125903157-125903157 on Build GRCh38
rs371973142 Chr.11: 125903555-125903572 on Build GRCh38
rs906914910 Chr.11: 125903512-125903512 on Build GRCh38
rs906800372 Chr.11: 125903288-125903288 on Build GRCh38
rs1045259866 Chr.11: 125903272-125903272 on Build GRCh38
rs933785518 Chr.11: 125903211-125903211 on Build GRCh38
rs552936471 Chr.11: 125903319-125903319 on Build GRCh38
rs530591163 Chr.11: 125903188-125903188 on Build GRCh38
rs913972737 Chr.11: 125903260-125903260 on Build GRCh38
rs894133640 Chr.11: 125903261-125903261 on Build GRCh38
rs541211370 Chr.11: 125903179-125903179 on Build GRCh38
rs549653832 Chr.11: 125903339-125903339 on Build GRCh38

View Full Product Details