Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1459016-1459553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3C
Entrez Gene ID
219293
Gene Name
ATPase family, AAA domain containing 3C
Gene Aliases
-
UniGene
Hs.724767
Gene Chromosome Location
Chr.1: 1449689-1470158 on Build GRCh38
Transcripts
NM_001039211.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4418702 c.742-2A>G p.? Chr.1: 1459159-1459159 on Build GRCh38
3472810 c.783C>T p.F261F Chr.1: 1459202-1459202 on Build GRCh38
1333440 c.794C>T p.T265M Chr.1: 1459213-1459213 on Build GRCh38
2230043 c.749T>C p.I250T Chr.1: 1459168-1459168 on Build GRCh38
2230044 c.777C>T p.N259N Chr.1: 1459196-1459196 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1039118383 Chr.1: 1459322-1459322 on Build GRCh38
rs779090181 Chr.1: 1459214-1459214 on Build GRCh38
rs981144520 Chr.1: 1459401-1459401 on Build GRCh38
rs1011485595 Chr.1: 1459533-1459534 on Build GRCh38
rs566195692 Chr.1: 1459400-1459400 on Build GRCh38
rs1751468 Chr.1: 1459045-1459045 on Build GRCh38
rs780699793 Chr.1: 1459259-1459259 on Build GRCh38
rs777535435 Chr.1: 1459255-1459255 on Build GRCh38
rs533302410 Chr.1: 1459313-1459313 on Build GRCh38
rs775924265 Chr.1: 1459262-1459262 on Build GRCh38
rs999899776 Chr.1: 1459493-1459493 on Build GRCh38
rs190264554 Chr.1: 1459247-1459247 on Build GRCh38
rs561467649 Chr.1: 1459218-1459218 on Build GRCh38
rs763502640 Chr.1: 1459269-1459269 on Build GRCh38
rs950365254 Chr.1: 1459426-1459426 on Build GRCh38
rs34719940 Chr.1: 1459273-1459273 on Build GRCh38
rs1054786931 Chr.1: 1459334-1459334 on Build GRCh38
rs560911953 Chr.1: 1459127-1459127 on Build GRCh38
rs555941983 Chr.1: 1459468-1459468 on Build GRCh38
rs765685109 Chr.1: 1459115-1459115 on Build GRCh38
rs535981931 Chr.1: 1459159-1459159 on Build GRCh38
rs755256408 Chr.1: 1459188-1459188 on Build GRCh38
rs774824584 Chr.1: 1459274-1459274 on Build GRCh38
rs1022750549 Chr.1: 1459392-1459392 on Build GRCh38
rs915597377 Chr.1: 1459060-1459060 on Build GRCh38
rs750531612 Chr.1: 1459280-1459280 on Build GRCh38
rs1048210317 Chr.1: 1459510-1459510 on Build GRCh38
rs897074704 Chr.1: 1459482-1459482 on Build GRCh38
rs770277138 Chr.1: 1459384-1459384 on Build GRCh38
rs900625479 Chr.1: 1459341-1459341 on Build GRCh38
rs1055412487 Chr.1: 1459164-1459164 on Build GRCh38
rs767744235 Chr.1: 1459234-1459234 on Build GRCh38
rs558457491 Chr.1: 1459197-1459197 on Build GRCh38
rs555910000 Chr.1: 1459161-1459161 on Build GRCh38
rs778659209 Chr.1: 1459246-1459246 on Build GRCh38
rs543633168 Chr.1: 1459202-1459202 on Build GRCh38
rs199523351 Chr.1: 1459214-1459214 on Build GRCh38
rs535041748 Chr.1: 1459196-1459196 on Build GRCh38
rs965774733 Chr.1: 1459414-1459414 on Build GRCh38
rs78193071 Chr.1: 1459391-1459391 on Build GRCh38
rs901146641 Chr.1: 1459522-1459522 on Build GRCh38
rs1013485406 Chr.1: 1459352-1459352 on Build GRCh38
rs752961653 Chr.1: 1459125-1459125 on Build GRCh38
rs762155623 Chr.1: 1459275-1459275 on Build GRCh38
rs1018787747 Chr.1: 1459520-1459520 on Build GRCh38
rs758665962 Chr.1: 1459129-1459129 on Build GRCh38
rs372776056 Chr.1: 1459474-1459474 on Build GRCh38
rs1024795738 Chr.1: 1459184-1459184 on Build GRCh38
rs569794156 Chr.1: 1459363-1459363 on Build GRCh38
rs1047927969 Chr.1: 1459157-1459158 on Build GRCh38
rs570738138 Chr.1: 1459056-1459056 on Build GRCh38
rs1020500909 Chr.1: 1459433-1459433 on Build GRCh38
rs1053045145 Chr.1: 1459516-1459516 on Build GRCh38
rs866226462 Chr.1: 1459351-1459351 on Build GRCh38
rs888298981 Chr.1: 1459517-1459517 on Build GRCh38
rs758434462 Chr.1: 1459261-1459262 on Build GRCh38
rs766720896 Chr.1: 1459175-1459176 on Build GRCh38
rs754183955 Chr.1: 1459186-1459187 on Build GRCh38
rs767801504 Chr.1: 1459279-1459279 on Build GRCh38
rs200778422 Chr.1: 1459168-1459168 on Build GRCh38
rs760116888 Chr.1: 1459209-1459209 on Build GRCh38
rs746715371 Chr.1: 1459257-1459257 on Build GRCh38
rs928328338 Chr.1: 1459410-1459410 on Build GRCh38
rs1045970686 Chr.1: 1459104-1459104 on Build GRCh38
rs894828516 Chr.1: 1459342-1459342 on Build GRCh38
rs948361479 Chr.1: 1459101-1459101 on Build GRCh38
rs1031525905 Chr.1: 1459243-1459243 on Build GRCh38
rs752784777 Chr.1: 1459240-1459241 on Build GRCh38
rs751690138 Chr.1: 1459148-1459148 on Build GRCh38
rs757545026 Chr.1: 1459359-1459359 on Build GRCh38
rs758198400 Chr.1: 1459253-1459253 on Build GRCh38
rs557372840 Chr.1: 1459479-1459479 on Build GRCh38
rs527258865 Chr.1: 1459244-1459244 on Build GRCh38
rs181723185 Chr.1: 1459434-1459434 on Build GRCh38
rs926143903 Chr.1: 1459463-1459463 on Build GRCh38
rs929913200 Chr.1: 1459483-1459483 on Build GRCh38
rs557580508 Chr.1: 1459210-1459210 on Build GRCh38
rs372248146 Chr.1: 1459261-1459261 on Build GRCh38
rs757478058 Chr.1: 1459151-1459151 on Build GRCh38
rs140945924 Chr.1: 1459370-1459370 on Build GRCh38
rs896958844 Chr.1: 1459360-1459360 on Build GRCh38
rs574149418 Chr.1: 1459518-1459518 on Build GRCh38
rs372265864 Chr.1: 1459422-1459422 on Build GRCh38
rs762104239 Chr.1: 1459231-1459231 on Build GRCh38
rs772784905 Chr.1: 1459189-1459189 on Build GRCh38
rs978896094 Chr.1: 1459459-1459459 on Build GRCh38
rs574161074 Chr.1: 1459213-1459213 on Build GRCh38
rs770541714 Chr.1: 1459258-1459258 on Build GRCh38
rs945109593 Chr.1: 1459055-1459055 on Build GRCh38
rs549973599 Chr.1: 1459111-1459111 on Build GRCh38
rs752427001 Chr.1: 1459283-1459283 on Build GRCh38
rs976943161 Chr.1: 1459423-1459423 on Build GRCh38
rs76548509 Chr.1: 1459284-1459284 on Build GRCh38
rs571844284 Chr.1: 1459476-1459476 on Build GRCh38
rs570046441 Chr.1: 1459147-1459147 on Build GRCh38
rs902528174 Chr.1: 1459484-1459484 on Build GRCh38
rs75201061 Chr.1: 1459357-1459357 on Build GRCh38
rs751887903 Chr.1: 1459220-1459220 on Build GRCh38
rs747849727 Chr.1: 1459173-1459173 on Build GRCh38
rs371333978 Chr.1: 1459050-1459050 on Build GRCh38
rs778801039 Chr.1: 1459172-1459172 on Build GRCh38
rs539239230 Chr.1: 1459106-1459106 on Build GRCh38
rs537790499 Chr.1: 1459477-1459477 on Build GRCh38
rs925058287 Chr.1: 1459298-1459298 on Build GRCh38
rs777150602 Chr.1: 1459040-1459040 on Build GRCh38
rs944326327 Chr.1: 1459039-1459039 on Build GRCh38
rs578222969 Chr.1: 1459199-1459199 on Build GRCh38

View Full Product Details