Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134364237-134364744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L2
Entrez Gene ID
89944
Gene Name
galactosidase beta 1 like 2
Gene Aliases
MST114, MSTP114
UniGene
Hs.436178
Gene Chromosome Location
Chr.11: 134331874-134376324 on Build GRCh38
Transcripts
NM_138342.3 XM_006718937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244707 c.763C>A p.H255N Chr.11: 134364357-134364357 on Build GRCh38
2090437 c.774G>C p.Q258H Chr.11: 134364368-134364368 on Build GRCh38
5665628 c.804+1G>T p.? Chr.11: 134364399-134364399 on Build GRCh38
925229 c.766G>A p.E256K Chr.11: 134364360-134364360 on Build GRCh38
6671370 c.765C>T p.H255H Chr.11: 134364359-134364359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544158686 Chr.11: 134364616-134364616 on Build GRCh38
rs60460137 Chr.11: 134364292-134364292 on Build GRCh38
rs767426605 Chr.11: 134364363-134364363 on Build GRCh38
rs979847279 Chr.11: 134364465-134364465 on Build GRCh38
rs190239347 Chr.11: 134364591-134364591 on Build GRCh38
rs903951595 Chr.11: 134364366-134364366 on Build GRCh38
rs913183032 Chr.11: 134364488-134364488 on Build GRCh38
rs936526423 Chr.11: 134364426-134364426 on Build GRCh38
rs754187787 Chr.11: 134364287-134364287 on Build GRCh38
rs7103363 Chr.11: 134364514-134364514 on Build GRCh38
rs1024950257 Chr.11: 134364554-134364554 on Build GRCh38
rs997561515 Chr.11: 134364562-134364562 on Build GRCh38
rs757932900 Chr.11: 134364404-134364404 on Build GRCh38
rs112886248 Chr.11: 134364393-134364393 on Build GRCh38
rs374456329 Chr.11: 134364440-134364440 on Build GRCh38
rs994336226 Chr.11: 134364307-134364307 on Build GRCh38
rs1008077433 Chr.11: 134364529-134364529 on Build GRCh38
rs889589071 Chr.11: 134364524-134364524 on Build GRCh38
rs1042932368 Chr.11: 134364690-134364690 on Build GRCh38
rs950946435 Chr.11: 134364573-134364573 on Build GRCh38
rs532988700 Chr.11: 134364648-134364648 on Build GRCh38
rs551570910 Chr.11: 134364425-134364425 on Build GRCh38
rs200983096 Chr.11: 134364368-134364368 on Build GRCh38
rs891694128 Chr.11: 134364337-134364337 on Build GRCh38
rs764534807 Chr.11: 134364267-134364267 on Build GRCh38
rs772336998 Chr.11: 134364434-134364434 on Build GRCh38
rs966050225 Chr.11: 134364377-134364377 on Build GRCh38
rs150932563 Chr.11: 134364470-134364470 on Build GRCh38
rs755760073 Chr.11: 134364392-134364392 on Build GRCh38
rs768636635 Chr.11: 134364336-134364336 on Build GRCh38
rs769649838 Chr.11: 134364428-134364428 on Build GRCh38
rs912867554 Chr.11: 134364668-134364668 on Build GRCh38
rs911040370 Chr.11: 134364711-134364711 on Build GRCh38
rs750784757 Chr.11: 134364447-134364447 on Build GRCh38
rs760269880 Chr.11: 134364528-134364528 on Build GRCh38
rs114723326 Chr.11: 134364375-134364375 on Build GRCh38
rs200333553 Chr.11: 134364646-134364646 on Build GRCh38
rs368269712 Chr.11: 134364600-134364600 on Build GRCh38
rs942603354 Chr.11: 134364471-134364471 on Build GRCh38
rs764607442 Chr.11: 134364441-134364441 on Build GRCh38
rs959580023 Chr.11: 134364528-134364528 on Build GRCh38
rs777082395 Chr.11: 134364444-134364444 on Build GRCh38
rs139338435 Chr.11: 134364359-134364359 on Build GRCh38
rs1005265090 Chr.11: 134364641-134364641 on Build GRCh38
rs771323713 Chr.11: 134364357-134364357 on Build GRCh38
rs550605133 Chr.11: 134364299-134364300 on Build GRCh38
rs747938335 Chr.11: 134364348-134364348 on Build GRCh38
rs960048832 Chr.11: 134364631-134364631 on Build GRCh38
rs778505587 Chr.11: 134364305-134364305 on Build GRCh38
rs759815828 Chr.11: 134364300-134364301 on Build GRCh38
rs746137587 Chr.11: 134364431-134364431 on Build GRCh38
rs780056198 Chr.11: 134364335-134364335 on Build GRCh38
rs370456525 Chr.11: 134364438-134364438 on Build GRCh38
rs776932132 Chr.11: 134364695-134364695 on Build GRCh38
rs550307967 Chr.11: 134364421-134364421 on Build GRCh38
rs1041269560 Chr.11: 134364701-134364701 on Build GRCh38
rs563132013 Chr.11: 134364597-134364597 on Build GRCh38
rs140590685 Chr.11: 134364489-134364489 on Build GRCh38
rs757799529 Chr.11: 134364334-134364334 on Build GRCh38
rs57782915 Chr.11: 134364483-134364483 on Build GRCh38
rs942578284 Chr.11: 134364696-134364696 on Build GRCh38
rs762168887 Chr.11: 134364446-134364446 on Build GRCh38
rs386759206 Chr.11: 134364512-134364514 on Build GRCh38
rs553352778 Chr.11: 134364709-134364709 on Build GRCh38
rs1026317487 Chr.11: 134364565-134364565 on Build GRCh38
rs560331547 Chr.11: 134364713-134364713 on Build GRCh38
rs374794020 Chr.11: 134364406-134364406 on Build GRCh38
rs199672296 Chr.11: 134364523-134364523 on Build GRCh38
rs754241077 Chr.11: 134364383-134364383 on Build GRCh38
rs759671556 Chr.11: 134364360-134364360 on Build GRCh38
rs551320527 Chr.11: 134364596-134364596 on Build GRCh38
rs775633466 Chr.11: 134364367-134364367 on Build GRCh38
rs79881383 Chr.11: 134364482-134364482 on Build GRCh38
rs200378528 Chr.11: 134364422-134364422 on Build GRCh38
rs543221153 Chr.11: 134364423-134364423 on Build GRCh38
rs4262740 Chr.11: 134364512-134364512 on Build GRCh38
rs989279168 Chr.11: 134364566-134364566 on Build GRCh38
rs1016503737 Chr.11: 134364390-134364390 on Build GRCh38
rs749987929 Chr.11: 134364399-134364399 on Build GRCh38
rs1006444063 Chr.11: 134364581-134364581 on Build GRCh38
rs201749594 Chr.11: 134364423-134364423 on Build GRCh38
rs201765506 Chr.11: 134364429-134364429 on Build GRCh38
rs551487709 Chr.11: 134364702-134364702 on Build GRCh38
rs1025684807 Chr.11: 134364657-134364657 on Build GRCh38
rs771303650 Chr.11: 134364678-134364678 on Build GRCh38
rs1007610787 Chr.11: 134364374-134364374 on Build GRCh38
rs888156684 Chr.11: 134364624-134364624 on Build GRCh38
rs745414183 Chr.11: 134364332-134364332 on Build GRCh38
rs557863989 Chr.11: 134364557-134364557 on Build GRCh38
rs1019702042 Chr.11: 134364295-134364295 on Build GRCh38
rs7103469 Chr.11: 134364558-134364558 on Build GRCh38
rs549252055 Chr.11: 134364473-134364473 on Build GRCh38
rs577924366 Chr.11: 134364719-134364719 on Build GRCh38
rs1006384078 Chr.11: 134364522-134364523 on Build GRCh38
rs776629486 Chr.11: 134364344-134364344 on Build GRCh38
rs767672333 Chr.11: 134364300-134364303 on Build GRCh38
rs947321450 Chr.11: 134364688-134364688 on Build GRCh38

View Full Product Details