Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134368554-134369047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L2
Entrez Gene ID
89944
Gene Name
galactosidase beta 1 like 2
Gene Aliases
MST114, MSTP114
UniGene
Hs.436178
Gene Chromosome Location
Chr.11: 134331874-134376324 on Build GRCh38
Transcripts
NM_138342.3 XM_006718937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
328729 c.966C>G p.N322K Chr.11: 134368720-134368720 on Build GRCh38
2090441 c.903C>T p.T301T Chr.11: 134368657-134368657 on Build GRCh38
925231 c.955G>A p.G319R Chr.11: 134368709-134368709 on Build GRCh38
7154076 c.939C>A p.N313K Chr.11: 134368693-134368693 on Build GRCh38
4019559 c.904G>A p.V302M Chr.11: 134368658-134368658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887683158 Chr.11: 134368621-134368621 on Build GRCh38
rs78013315 Chr.11: 134368856-134368856 on Build GRCh38
rs73589616 Chr.11: 134368971-134368971 on Build GRCh38
rs950786909 Chr.11: 134368929-134368929 on Build GRCh38
rs957621117 Chr.11: 134368590-134368590 on Build GRCh38
rs147889120 Chr.11: 134368642-134368642 on Build GRCh38
rs35217320 Chr.11: 134368705-134368705 on Build GRCh38
rs199527684 Chr.11: 134368669-134368669 on Build GRCh38
rs767249800 Chr.11: 134368641-134368641 on Build GRCh38
rs781367414 Chr.11: 134368698-134368698 on Build GRCh38
rs781314364 Chr.11: 134368613-134368613 on Build GRCh38
rs201643519 Chr.11: 134368686-134368686 on Build GRCh38
rs1055486908 Chr.11: 134368717-134368717 on Build GRCh38
rs747918955 Chr.11: 134368825-134368829 on Build GRCh38
rs567269407 Chr.11: 134368972-134368972 on Build GRCh38
rs369853573 Chr.11: 134368678-134368678 on Build GRCh38
rs374163223 Chr.11: 134368639-134368639 on Build GRCh38
rs78260611 Chr.11: 134368624-134368624 on Build GRCh38
rs551369142 Chr.11: 134368624-134368625 on Build GRCh38
rs772768983 Chr.11: 134368936-134368936 on Build GRCh38
rs762136402 Chr.11: 134368920-134368920 on Build GRCh38
rs1017853333 Chr.11: 134369026-134369026 on Build GRCh38
rs968336251 Chr.11: 134369018-134369018 on Build GRCh38
rs892534936 Chr.11: 134368990-134368990 on Build GRCh38
rs1048865638 Chr.11: 134368691-134368691 on Build GRCh38
rs780858394 Chr.11: 134368801-134368802 on Build GRCh38
rs750265921 Chr.11: 134368818-134368818 on Build GRCh38
rs754690463 Chr.11: 134368695-134368695 on Build GRCh38
rs372771379 Chr.11: 134368731-134368731 on Build GRCh38
rs760215327 Chr.11: 134368657-134368657 on Build GRCh38
rs140813422 Chr.11: 134368708-134368708 on Build GRCh38
rs968702182 Chr.11: 134368695-134368695 on Build GRCh38
rs532540966 Chr.11: 134368919-134368919 on Build GRCh38
rs966275665 Chr.11: 134368880-134368880 on Build GRCh38
rs563854435 Chr.11: 134368766-134368766 on Build GRCh38
rs148929536 Chr.11: 134368676-134368676 on Build GRCh38
rs748245997 Chr.11: 134368700-134368700 on Build GRCh38
rs562525799 Chr.11: 134368789-134368789 on Build GRCh38
rs770249861 Chr.11: 134368614-134368614 on Build GRCh38
rs948612108 Chr.11: 134368863-134368863 on Build GRCh38
rs747176431 Chr.11: 134368601-134368601 on Build GRCh38
rs560069347 Chr.11: 134368884-134368884 on Build GRCh38
rs773716055 Chr.11: 134368620-134368620 on Build GRCh38
rs751604823 Chr.11: 134368985-134368985 on Build GRCh38
rs539605151 Chr.11: 134368658-134368658 on Build GRCh38
rs774898396 Chr.11: 134368747-134368747 on Build GRCh38
rs1009610269 Chr.11: 134368991-134368991 on Build GRCh38
rs544226848 Chr.11: 134368726-134368726 on Build GRCh38
rs909136531 Chr.11: 134368857-134368857 on Build GRCh38
rs527376380 Chr.11: 134368636-134368636 on Build GRCh38
rs372904369 Chr.11: 134368637-134368637 on Build GRCh38
rs557032028 Chr.11: 134368587-134368587 on Build GRCh38
rs1022677627 Chr.11: 134369008-134369008 on Build GRCh38
rs751100845 Chr.11: 134368679-134368679 on Build GRCh38
rs765052401 Chr.11: 134368817-134368817 on Build GRCh38
rs375488325 Chr.11: 134368638-134368638 on Build GRCh38
rs369111573 Chr.11: 134368625-134368626 on Build GRCh38
rs753742914 Chr.11: 134368667-134368667 on Build GRCh38
rs1044223920 Chr.11: 134368961-134368961 on Build GRCh38
rs767754194 Chr.11: 134368827-134368827 on Build GRCh38
rs957799897 Chr.11: 134368859-134368859 on Build GRCh38
rs771875565 Chr.11: 134368787-134368787 on Build GRCh38
rs143581329 Chr.11: 134368679-134368679 on Build GRCh38
rs149574035 Chr.11: 134368820-134368820 on Build GRCh38
rs527517764 Chr.11: 134368903-134368903 on Build GRCh38
rs746215244 Chr.11: 134368785-134368785 on Build GRCh38
rs138574847 Chr.11: 134368730-134368730 on Build GRCh38
rs760266441 Chr.11: 134368803-134368803 on Build GRCh38
rs377274952 Chr.11: 134368720-134368720 on Build GRCh38
rs909040559 Chr.11: 134368939-134368939 on Build GRCh38
rs765136821 Chr.11: 134368670-134368670 on Build GRCh38
rs929022136 Chr.11: 134368844-134368844 on Build GRCh38
rs1035000024 Chr.11: 134368675-134368675 on Build GRCh38
rs768850751 Chr.11: 134368612-134368612 on Build GRCh38
rs138130138 Chr.11: 134368709-134368709 on Build GRCh38
rs761876285 Chr.11: 134368814-134368814 on Build GRCh38
rs1007785054 Chr.11: 134369024-134369024 on Build GRCh38
rs1022579708 Chr.11: 134368680-134368680 on Build GRCh38
rs1018235711 Chr.11: 134368879-134368879 on Build GRCh38
rs949293322 Chr.11: 134368977-134368977 on Build GRCh38
rs377193057 Chr.11: 134368804-134368804 on Build GRCh38
rs752987312 Chr.11: 134368872-134368872 on Build GRCh38
rs919278293 Chr.11: 134368916-134368916 on Build GRCh38
rs775989937 Chr.11: 134368806-134368806 on Build GRCh38

View Full Product Details