Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7483244-7483741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163
Entrez Gene ID
9332
Gene Name
CD163 molecule
Gene Aliases
M130, MM130, SCARI1
UniGene
Hs.504641
Gene Chromosome Location
Chr.12: 7470816-7503818 on Build GRCh38
Transcripts
NM_004244.5 NM_203416.3 XM_005253528.4 XM_005253529.4 XM_017020231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2152637 c.3133C>T p.R1045C Chr.12: 7483421-7483421 on Build GRCh38
2152636 c.3034C>T p.R1012C Chr.12: 7483421-7483421 on Build GRCh38
4540191 c.2801G>A p.G934E Chr.12: 7483654-7483654 on Build GRCh38
3872519 c.2941G>A p.E981K Chr.12: 7483514-7483514 on Build GRCh38
5841903 c.3076G>A p.V1026M Chr.12: 7483379-7483379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368492058 Chr.12: 7483377-7483377 on Build GRCh38
rs755454282 Chr.12: 7483391-7483391 on Build GRCh38
rs771733043 Chr.12: 7483474-7483474 on Build GRCh38
rs755265817 Chr.12: 7483408-7483408 on Build GRCh38
rs760675429 Chr.12: 7483691-7483691 on Build GRCh38
rs1045655047 Chr.12: 7483488-7483488 on Build GRCh38
rs554863113 Chr.12: 7483358-7483358 on Build GRCh38
rs775082662 Chr.12: 7483485-7483485 on Build GRCh38
rs757152352 Chr.12: 7483617-7483617 on Build GRCh38
rs371406699 Chr.12: 7483572-7483572 on Build GRCh38
rs753885158 Chr.12: 7483613-7483613 on Build GRCh38
rs764142833 Chr.12: 7483574-7483574 on Build GRCh38
rs747646809 Chr.12: 7483695-7483695 on Build GRCh38
rs760010924 Chr.12: 7483367-7483367 on Build GRCh38
rs777891066 Chr.12: 7483425-7483425 on Build GRCh38
rs770685020 Chr.12: 7483325-7483325 on Build GRCh38
rs776200093 Chr.12: 7483499-7483499 on Build GRCh38
rs199560015 Chr.12: 7483641-7483641 on Build GRCh38
rs771413158 Chr.12: 7483518-7483518 on Build GRCh38
rs200469785 Chr.12: 7483442-7483442 on Build GRCh38
rs748577162 Chr.12: 7483439-7483439 on Build GRCh38
rs746721451 Chr.12: 7483635-7483635 on Build GRCh38
rs771374208 Chr.12: 7483709-7483709 on Build GRCh38
rs774603375 Chr.12: 7483527-7483527 on Build GRCh38
rs141756075 Chr.12: 7483564-7483564 on Build GRCh38
rs753020548 Chr.12: 7483563-7483563 on Build GRCh38
rs199986440 Chr.12: 7483475-7483475 on Build GRCh38
rs767965678 Chr.12: 7483548-7483548 on Build GRCh38
rs774964934 Chr.12: 7483716-7483716 on Build GRCh38
rs759943157 Chr.12: 7483531-7483531 on Build GRCh38
rs374982629 Chr.12: 7483420-7483420 on Build GRCh38
rs779801921 Chr.12: 7483631-7483631 on Build GRCh38
rs145413017 Chr.12: 7483385-7483385 on Build GRCh38
rs749221257 Chr.12: 7483324-7483324 on Build GRCh38
rs964444440 Chr.12: 7483634-7483634 on Build GRCh38
rs192304862 Chr.12: 7483315-7483315 on Build GRCh38
rs774178726 Chr.12: 7483342-7483342 on Build GRCh38
rs768204023 Chr.12: 7483486-7483486 on Build GRCh38
rs112063621 Chr.12: 7483366-7483366 on Build GRCh38
rs993701744 Chr.12: 7483428-7483428 on Build GRCh38
rs866293964 Chr.12: 7483456-7483456 on Build GRCh38
rs776402823 Chr.12: 7483494-7483494 on Build GRCh38
rs751885503 Chr.12: 7483404-7483404 on Build GRCh38
rs945949258 Chr.12: 7483464-7483464 on Build GRCh38
rs371546077 Chr.12: 7483386-7483386 on Build GRCh38
rs750247723 Chr.12: 7483625-7483625 on Build GRCh38
rs763870982 Chr.12: 7483571-7483571 on Build GRCh38
rs775146850 Chr.12: 7483300-7483300 on Build GRCh38
rs867643659 Chr.12: 7483472-7483472 on Build GRCh38
rs778120642 Chr.12: 7483293-7483293 on Build GRCh38
rs766720451 Chr.12: 7483399-7483399 on Build GRCh38
rs780582176 Chr.12: 7483669-7483669 on Build GRCh38
rs144924673 Chr.12: 7483455-7483455 on Build GRCh38
rs763374425 Chr.12: 7483511-7483511 on Build GRCh38
rs766442796 Chr.12: 7483319-7483319 on Build GRCh38
rs761565641 Chr.12: 7483626-7483626 on Build GRCh38
rs78817356 Chr.12: 7483431-7483431 on Build GRCh38
rs372790972 Chr.12: 7483557-7483557 on Build GRCh38
rs752858863 Chr.12: 7483421-7483421 on Build GRCh38
rs867419942 Chr.12: 7483444-7483444 on Build GRCh38
rs986972938 Chr.12: 7483303-7483303 on Build GRCh38
rs190006448 Chr.12: 7483285-7483285 on Build GRCh38
rs756305706 Chr.12: 7483422-7483422 on Build GRCh38
rs975931931 Chr.12: 7483352-7483352 on Build GRCh38
rs368897599 Chr.12: 7483566-7483566 on Build GRCh38
rs745473362 Chr.12: 7483618-7483618 on Build GRCh38
rs773631588 Chr.12: 7483389-7483389 on Build GRCh38
rs147938240 Chr.12: 7483675-7483675 on Build GRCh38
rs79260013 Chr.12: 7483345-7483345 on Build GRCh38

View Full Product Details