Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9732850-9733390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLECL1
Entrez Gene ID
160365
Gene Name
C-type lectin like 1
Gene Aliases
DCAL-1, DCAL1
UniGene
Hs.560087
Gene Chromosome Location
Chr.12: 9715860-9733435 on Build GRCh38
Transcripts
NM_001253750.1 NM_001267701.1 NM_172004.3 XM_011520574.2 XM_017018885.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466758 c.129G>A p.R43R Chr.12: 9733136-9733136 on Build GRCh38
3466757 c.213C>T p.F71F Chr.12: 9733052-9733052 on Build GRCh38
3688508 c.283C>T p.P95S Chr.12: 9732982-9732982 on Build GRCh38
3664880 c.316+5G>T p.? Chr.12: 9732944-9732944 on Build GRCh38
3753550 c.48T>A p.A16A Chr.12: 9733217-9733217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61913428 Chr.12: 9733217-9733217 on Build GRCh38
rs777393740 Chr.12: 9733000-9733000 on Build GRCh38
rs761669698 Chr.12: 9733073-9733073 on Build GRCh38
rs1023499997 Chr.12: 9732969-9732969 on Build GRCh38
rs779195515 Chr.12: 9733037-9733037 on Build GRCh38
rs779423983 Chr.12: 9733316-9733320 on Build GRCh38
rs754111425 Chr.12: 9733029-9733029 on Build GRCh38
rs79809727 Chr.12: 9732982-9732982 on Build GRCh38
rs896597470 Chr.12: 9733163-9733163 on Build GRCh38
rs776643093 Chr.12: 9733242-9733242 on Build GRCh38
rs764597759 Chr.12: 9733261-9733261 on Build GRCh38
rs748653902 Chr.12: 9733039-9733039 on Build GRCh38
rs779506004 Chr.12: 9732968-9732968 on Build GRCh38
rs752781934 Chr.12: 9732993-9732994 on Build GRCh38
rs962384674 Chr.12: 9733192-9733192 on Build GRCh38
rs34201389 Chr.12: 9733244-9733244 on Build GRCh38
rs1016617605 Chr.12: 9732933-9732933 on Build GRCh38
rs12304510 Chr.12: 9732930-9732930 on Build GRCh38
rs142098692 Chr.12: 9733079-9733079 on Build GRCh38
rs755854874 Chr.12: 9733155-9733155 on Build GRCh38
rs776471903 Chr.12: 9732981-9732981 on Build GRCh38
rs759580248 Chr.12: 9733307-9733307 on Build GRCh38
rs138470841 Chr.12: 9733250-9733250 on Build GRCh38
rs547074560 Chr.12: 9733366-9733366 on Build GRCh38
rs762736859 Chr.12: 9732912-9732912 on Build GRCh38
rs144269074 Chr.12: 9733086-9733086 on Build GRCh38
rs148811159 Chr.12: 9733234-9733234 on Build GRCh38
rs1056207902 Chr.12: 9733153-9733153 on Build GRCh38
rs760774428 Chr.12: 9733101-9733101 on Build GRCh38
rs776756337 Chr.12: 9733304-9733304 on Build GRCh38
rs777603186 Chr.12: 9733296-9733296 on Build GRCh38
rs757815843 Chr.12: 9733078-9733078 on Build GRCh38
rs750627618 Chr.12: 9732931-9732931 on Build GRCh38
rs564859966 Chr.12: 9733128-9733128 on Build GRCh38
rs751550011 Chr.12: 9732922-9732922 on Build GRCh38
rs60145243 Chr.12: 9733344-9733344 on Build GRCh38
rs779484191 Chr.12: 9733170-9733170 on Build GRCh38
rs375872847 Chr.12: 9733278-9733278 on Build GRCh38
rs755624366 Chr.12: 9733160-9733160 on Build GRCh38
rs752096263 Chr.12: 9733264-9733264 on Build GRCh38
rs767578087 Chr.12: 9733010-9733010 on Build GRCh38
rs10492165 Chr.12: 9733365-9733365 on Build GRCh38
rs771833157 Chr.12: 9733290-9733290 on Build GRCh38
rs753065500 Chr.12: 9733246-9733246 on Build GRCh38
rs113222621 Chr.12: 9733114-9733115 on Build GRCh38
rs754069990 Chr.12: 9733106-9733106 on Build GRCh38
rs369340014 Chr.12: 9733143-9733143 on Build GRCh38
rs750439588 Chr.12: 9733345-9733345 on Build GRCh38
rs768195683 Chr.12: 9733180-9733180 on Build GRCh38
rs751488466 Chr.12: 9733001-9733001 on Build GRCh38
rs770544921 Chr.12: 9732971-9732971 on Build GRCh38
rs71929655 Chr.12: 9733112-9733113 on Build GRCh38
rs773102642 Chr.12: 9733228-9733228 on Build GRCh38
rs71045297 Chr.12: 9733111-9733112 on Build GRCh38
rs113575400 Chr.12: 9733111-9733111 on Build GRCh38
rs764110641 Chr.12: 9732920-9732920 on Build GRCh38
rs201053431 Chr.12: 9733063-9733063 on Build GRCh38
rs774280673 Chr.12: 9732897-9732897 on Build GRCh38
rs747838636 Chr.12: 9733283-9733283 on Build GRCh38
rs373212551 Chr.12: 9733046-9733046 on Build GRCh38
rs760579440 Chr.12: 9733229-9733229 on Build GRCh38
rs1035203934 Chr.12: 9732886-9732886 on Build GRCh38
rs374092196 Chr.12: 9733120-9733120 on Build GRCh38
rs1043890968 Chr.12: 9733093-9733093 on Build GRCh38
rs762833109 Chr.12: 9732991-9732991 on Build GRCh38
rs757292060 Chr.12: 9732924-9732924 on Build GRCh38
rs774073390 Chr.12: 9733199-9733199 on Build GRCh38
rs766409260 Chr.12: 9733105-9733105 on Build GRCh38
rs779546947 Chr.12: 9733271-9733271 on Build GRCh38
rs758921296 Chr.12: 9733123-9733123 on Build GRCh38
rs750504659 Chr.12: 9733012-9733012 on Build GRCh38
rs772775518 Chr.12: 9733042-9733042 on Build GRCh38
rs765114525 Chr.12: 9732901-9732901 on Build GRCh38
rs200639830 Chr.12: 9733117-9733117 on Build GRCh38
rs755339740 Chr.12: 9733030-9733030 on Build GRCh38
rs747782269 Chr.12: 9733110-9733111 on Build GRCh38
rs753118623 Chr.12: 9733122-9733122 on Build GRCh38
rs755254210 Chr.12: 9732990-9732990 on Build GRCh38
rs372501647 Chr.12: 9733172-9733172 on Build GRCh38
rs142778932 Chr.12: 9733361-9733361 on Build GRCh38
rs530695832 Chr.12: 9733173-9733173 on Build GRCh38
rs145268017 Chr.12: 9733053-9733053 on Build GRCh38
rs780785881 Chr.12: 9733050-9733050 on Build GRCh38
rs779620162 Chr.12: 9733062-9733062 on Build GRCh38
rs139676111 Chr.12: 9733008-9733008 on Build GRCh38
rs985759925 Chr.12: 9733355-9733355 on Build GRCh38
rs759408171 Chr.12: 9732893-9732893 on Build GRCh38
rs180674572 Chr.12: 9733198-9733198 on Build GRCh38
rs1045069955 Chr.12: 9733109-9733110 on Build GRCh38
rs756270202 Chr.12: 9733016-9733016 on Build GRCh38
rs780216106 Chr.12: 9732956-9732956 on Build GRCh38
rs986886282 Chr.12: 9733044-9733044 on Build GRCh38
rs376988344 Chr.12: 9733301-9733301 on Build GRCh38
rs774196156 Chr.12: 9733070-9733070 on Build GRCh38
rs147608426 Chr.12: 9733057-9733057 on Build GRCh38
rs776340468 Chr.12: 9732976-9732976 on Build GRCh38
rs151210429 Chr.12: 9732995-9732995 on Build GRCh38
rs202221764 Chr.12: 9732960-9732960 on Build GRCh38
rs749676733 Chr.12: 9732958-9732958 on Build GRCh38
rs748119457 Chr.12: 9733277-9733277 on Build GRCh38
rs750671734 Chr.12: 9732970-9732970 on Build GRCh38
rs756393100 Chr.12: 9732952-9732952 on Build GRCh38
rs375808663 Chr.12: 9733220-9733220 on Build GRCh38
rs780345412 Chr.12: 9733026-9733026 on Build GRCh38
rs373298472 Chr.12: 9733269-9733269 on Build GRCh38
rs115549012 Chr.12: 9732907-9732907 on Build GRCh38
rs914559593 Chr.12: 9733320-9733320 on Build GRCh38
rs749063416 Chr.12: 9732874-9732874 on Build GRCh38
rs370148784 Chr.12: 9733243-9733243 on Build GRCh38
rs746360704 Chr.12: 9732881-9732881 on Build GRCh38
rs200640673 Chr.12: 9732980-9732980 on Build GRCh38
rs398070028 Chr.12: 9733116-9733117 on Build GRCh38

View Full Product Details