Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12094293-12094827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L14
Entrez Gene ID
79370
Gene Name
BCL2 like 14
Gene Aliases
BCLG
UniGene
Hs.210343
Gene Chromosome Location
Chr.12: 12049844-12099695 on Build GRCh38
Transcripts
NM_030766.1 NM_138722.1 NM_138723.1 XM_006719151.3 XM_011520848.1 XM_011520849.1 XM_017019963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4039525 c.790G>A p.E264K Chr.12: 12094775-12094775 on Build GRCh38
4039526 c.821T>C p.L274P Chr.12: 12094806-12094806 on Build GRCh38
1359595 c.765G>A p.M255I Chr.12: 12094750-12094750 on Build GRCh38
7247929 c.697T>C p.L233L Chr.12: 12094682-12094682 on Build GRCh38
546938 c.730C>T p.P244S Chr.12: 12094562-12094562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761569693 Chr.12: 12094666-12094666 on Build GRCh38
rs762964230 Chr.12: 12094388-12094388 on Build GRCh38
rs1641728 Chr.12: 12094426-12094426 on Build GRCh38
rs753588720 Chr.12: 12094702-12094702 on Build GRCh38
rs61738409 Chr.12: 12094760-12094760 on Build GRCh38
rs963796823 Chr.12: 12094347-12094347 on Build GRCh38
rs4763779 Chr.12: 12094588-12094588 on Build GRCh38
rs895188353 Chr.12: 12094657-12094657 on Build GRCh38
rs772203208 Chr.12: 12094494-12094494 on Build GRCh38
rs775650764 Chr.12: 12094665-12094665 on Build GRCh38
rs752680306 Chr.12: 12094618-12094618 on Build GRCh38
rs535131543 Chr.12: 12094517-12094517 on Build GRCh38
rs973522821 Chr.12: 12094377-12094377 on Build GRCh38
rs758270759 Chr.12: 12094619-12094619 on Build GRCh38
rs781524706 Chr.12: 12094645-12094645 on Build GRCh38
rs760547461 Chr.12: 12094572-12094572 on Build GRCh38
rs551408987 Chr.12: 12094389-12094389 on Build GRCh38
rs753403340 Chr.12: 12094582-12094582 on Build GRCh38
rs142455025 Chr.12: 12094425-12094425 on Build GRCh38
rs529228448 Chr.12: 12094423-12094423 on Build GRCh38
rs1051750089 Chr.12: 12094394-12094394 on Build GRCh38
rs931998987 Chr.12: 12094384-12094384 on Build GRCh38
rs754906258 Chr.12: 12094474-12094474 on Build GRCh38
rs977560761 Chr.12: 12094800-12094800 on Build GRCh38
rs373995100 Chr.12: 12094641-12094641 on Build GRCh38
rs755854881 Chr.12: 12094564-12094564 on Build GRCh38
rs963494254 Chr.12: 12094724-12094724 on Build GRCh38
rs538608614 Chr.12: 12094568-12094569 on Build GRCh38
rs370468556 Chr.12: 12094732-12094732 on Build GRCh38
rs760305149 Chr.12: 12094697-12094697 on Build GRCh38
rs575884005 Chr.12: 12094669-12094669 on Build GRCh38
rs267603353 Chr.12: 12094745-12094745 on Build GRCh38
rs369434316 Chr.12: 12094545-12094545 on Build GRCh38
rs758360894 Chr.12: 12094738-12094738 on Build GRCh38
rs922004166 Chr.12: 12094381-12094381 on Build GRCh38
rs777702021 Chr.12: 12094361-12094361 on Build GRCh38
rs775406284 Chr.12: 12094535-12094535 on Build GRCh38
rs911817985 Chr.12: 12094410-12094410 on Build GRCh38
rs781127816 Chr.12: 12094782-12094782 on Build GRCh38
rs386760448 Chr.12: 12094659-12094660 on Build GRCh38
rs1029106525 Chr.12: 12094784-12094784 on Build GRCh38
rs1012362856 Chr.12: 12094362-12094362 on Build GRCh38
rs539198572 Chr.12: 12094659-12094659 on Build GRCh38
rs1015589120 Chr.12: 12094752-12094752 on Build GRCh38
rs74064622 Chr.12: 12094480-12094480 on Build GRCh38
rs771089400 Chr.12: 12094527-12094527 on Build GRCh38
rs569913421 Chr.12: 12094417-12094417 on Build GRCh38
rs188940783 Chr.12: 12094615-12094615 on Build GRCh38
rs143976011 Chr.12: 12094556-12094556 on Build GRCh38
rs765787822 Chr.12: 12094699-12094699 on Build GRCh38
rs987114746 Chr.12: 12094773-12094773 on Build GRCh38
rs765496952 Chr.12: 12094541-12094541 on Build GRCh38
rs893603651 Chr.12: 12094450-12094450 on Build GRCh38
rs11054683 Chr.12: 12094682-12094682 on Build GRCh38
rs973057363 Chr.12: 12094486-12094486 on Build GRCh38
rs778563211 Chr.12: 12094610-12094610 on Build GRCh38
rs759039939 Chr.12: 12094707-12094707 on Build GRCh38
rs777850411 Chr.12: 12094630-12094630 on Build GRCh38
rs769113790 Chr.12: 12094804-12094804 on Build GRCh38
rs749065278 Chr.12: 12094464-12094464 on Build GRCh38
rs769850390 Chr.12: 12094802-12094802 on Build GRCh38
rs773204969 Chr.12: 12094497-12094497 on Build GRCh38
rs765089911 Chr.12: 12094728-12094728 on Build GRCh38
rs867780243 Chr.12: 12094567-12094568 on Build GRCh38
rs747048001 Chr.12: 12094640-12094640 on Build GRCh38
rs1015066792 Chr.12: 12094709-12094709 on Build GRCh38
rs148640639 Chr.12: 12094767-12094767 on Build GRCh38
rs754856251 Chr.12: 12094590-12094590 on Build GRCh38
rs200229629 Chr.12: 12094526-12094526 on Build GRCh38
rs778175117 Chr.12: 12094484-12094485 on Build GRCh38
rs763934174 Chr.12: 12094655-12094655 on Build GRCh38
rs11054682 Chr.12: 12094660-12094660 on Build GRCh38
rs911609268 Chr.12: 12094570-12094570 on Build GRCh38
rs202074056 Chr.12: 12094635-12094635 on Build GRCh38
rs749632790 Chr.12: 12094803-12094803 on Build GRCh38
rs749896783 Chr.12: 12094562-12094562 on Build GRCh38
rs777725808 Chr.12: 12094758-12094758 on Build GRCh38
rs759983720 Chr.12: 12094539-12094539 on Build GRCh38
rs750044386 Chr.12: 12094775-12094775 on Build GRCh38
rs748017556 Chr.12: 12094481-12094481 on Build GRCh38
rs921624848 Chr.12: 12094520-12094520 on Build GRCh38
rs772882119 Chr.12: 12094696-12094696 on Build GRCh38
rs139549265 Chr.12: 12094786-12094786 on Build GRCh38

View Full Product Details