Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13089121-13089617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSG1
Entrez Gene ID
83445
Gene Name
germ cell associated 1
Gene Aliases
-
UniGene
Hs.240053
Gene Chromosome Location
Chr.12: 13083537-13103720 on Build GRCh38
Transcripts
NM_001080554.2 NM_001080555.2 NM_001206842.1 NM_001206843.1 NM_001206845.1 NM_031289.3 NM_153823.3 XM_005253493.2 XM_005253495.1 XM_011520858.1 XM_011520859.1 XM_011520860.1 XM_011520861.1 XM_011520862.1 XM_011520863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025282 c.423G>A p.A141A Chr.12: 13089218-13089218 on Build GRCh38
2228980 c.422C>T p.A141V Chr.12: 13089219-13089219 on Build GRCh38
6194516 c.381G>C p.Q127H Chr.12: 13089260-13089260 on Build GRCh38
5657463 c.421G>A p.A141T Chr.12: 13089220-13089220 on Build GRCh38
281574 c.409T>C p.S137P Chr.12: 13089232-13089232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889800434 Chr.12: 13089154-13089154 on Build GRCh38
rs145349009 Chr.12: 13089544-13089544 on Build GRCh38
rs1044830250 Chr.12: 13089486-13089486 on Build GRCh38
rs766728686 Chr.12: 13089273-13089273 on Build GRCh38
rs1003245498 Chr.12: 13089277-13089277 on Build GRCh38
rs6488584 Chr.12: 13089363-13089363 on Build GRCh38
rs371613978 Chr.12: 13089178-13089178 on Build GRCh38
rs953013652 Chr.12: 13089584-13089584 on Build GRCh38
rs150827186 Chr.12: 13089498-13089498 on Build GRCh38
rs558290346 Chr.12: 13089316-13089316 on Build GRCh38
rs1010773666 Chr.12: 13089472-13089472 on Build GRCh38
rs751179818 Chr.12: 13089323-13089323 on Build GRCh38
rs969031333 Chr.12: 13089237-13089237 on Build GRCh38
rs1035942426 Chr.12: 13089282-13089282 on Build GRCh38
rs1052599148 Chr.12: 13089354-13089354 on Build GRCh38
rs952880644 Chr.12: 13089328-13089328 on Build GRCh38
rs368946693 Chr.12: 13089218-13089218 on Build GRCh38
rs188527485 Chr.12: 13089331-13089331 on Build GRCh38
rs749398869 Chr.12: 13089397-13089397 on Build GRCh38
rs575649011 Chr.12: 13089219-13089219 on Build GRCh38
rs769294832 Chr.12: 13089317-13089317 on Build GRCh38
rs761230670 Chr.12: 13089245-13089245 on Build GRCh38
rs751183582 Chr.12: 13089340-13089340 on Build GRCh38
rs1010674960 Chr.12: 13089234-13089234 on Build GRCh38
rs765826295 Chr.12: 13089266-13089266 on Build GRCh38
rs745863860 Chr.12: 13089375-13089375 on Build GRCh38
rs773048049 Chr.12: 13089356-13089357 on Build GRCh38
rs763056375 Chr.12: 13089194-13089194 on Build GRCh38
rs146117765 Chr.12: 13089292-13089292 on Build GRCh38
rs542600236 Chr.12: 13089243-13089243 on Build GRCh38
rs548138170 Chr.12: 13089513-13089513 on Build GRCh38
rs747639565 Chr.12: 13089316-13089317 on Build GRCh38
rs1022294157 Chr.12: 13089166-13089166 on Build GRCh38
rs922471573 Chr.12: 13089435-13089435 on Build GRCh38
rs769009711 Chr.12: 13089207-13089207 on Build GRCh38
rs999762659 Chr.12: 13089200-13089200 on Build GRCh38
rs546700279 Chr.12: 13089355-13089355 on Build GRCh38
rs3736208 Chr.12: 13089437-13089437 on Build GRCh38
rs942693969 Chr.12: 13089341-13089341 on Build GRCh38
rs946925122 Chr.12: 13089385-13089385 on Build GRCh38
rs750895885 Chr.12: 13089185-13089185 on Build GRCh38
rs142432981 Chr.12: 13089424-13089424 on Build GRCh38
rs756078323 Chr.12: 13089378-13089378 on Build GRCh38
rs893623224 Chr.12: 13089453-13089453 on Build GRCh38
rs1015209746 Chr.12: 13089253-13089253 on Build GRCh38
rs186108110 Chr.12: 13089169-13089169 on Build GRCh38
rs373191738 Chr.12: 13089232-13089232 on Build GRCh38
rs779795056 Chr.12: 13089386-13089386 on Build GRCh38
rs1056902449 Chr.12: 13089467-13089469 on Build GRCh38
rs745798976 Chr.12: 13089168-13089168 on Build GRCh38
rs757856021 Chr.12: 13089322-13089322 on Build GRCh38
rs911334283 Chr.12: 13089333-13089333 on Build GRCh38
rs901121282 Chr.12: 13089193-13089193 on Build GRCh38
rs935270204 Chr.12: 13089436-13089436 on Build GRCh38
rs777702033 Chr.12: 13089541-13089541 on Build GRCh38
rs1023413065 Chr.12: 13089235-13089235 on Build GRCh38
rs79656918 Chr.12: 13089506-13089506 on Build GRCh38
rs756039401 Chr.12: 13089536-13089536 on Build GRCh38
rs1044022540 Chr.12: 13089393-13089393 on Build GRCh38
rs1027528614 Chr.12: 13089557-13089557 on Build GRCh38
rs994450378 Chr.12: 13089522-13089522 on Build GRCh38
rs201120155 Chr.12: 13089216-13089216 on Build GRCh38

View Full Product Details