Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13211388-13211892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP1
Entrez Gene ID
2012
Gene Name
epithelial membrane protein 1
Gene Aliases
CL-20, EMP-1, TMP
UniGene
Hs.719042
Gene Chromosome Location
Chr.12: 13196668-13216774 on Build GRCh38
Transcripts
NM_001423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6242400 c.6G>T p.L2F Chr.12: 13211516-13211516 on Build GRCh38
1676739 c.32T>G p.V11G Chr.12: 13211542-13211542 on Build GRCh38
6242337 c.7G>T p.V3L Chr.12: 13211517-13211517 on Build GRCh38
6653881 c.78+1G>A p.? Chr.12: 13211589-13211589 on Build GRCh38
6653880 c.47_49delTTA p.I17delI Chr.12: 13211557-13211559 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904110269 Chr.12: 13211859-13211859 on Build GRCh38
rs1041621738 Chr.12: 13211585-13211585 on Build GRCh38
rs529522945 Chr.12: 13211857-13211857 on Build GRCh38
rs894156133 Chr.12: 13211680-13211680 on Build GRCh38
rs529583201 Chr.12: 13211441-13211441 on Build GRCh38
rs770981492 Chr.12: 13211578-13211578 on Build GRCh38
rs760701186 Chr.12: 13211484-13211484 on Build GRCh38
rs147814045 Chr.12: 13211428-13211428 on Build GRCh38
rs2291061 Chr.12: 13211543-13211543 on Build GRCh38
rs758150356 Chr.12: 13211556-13211556 on Build GRCh38
rs543176001 Chr.12: 13211838-13211838 on Build GRCh38
rs952373434 Chr.12: 13211821-13211821 on Build GRCh38
rs777736024 Chr.12: 13211561-13211561 on Build GRCh38
rs756198161 Chr.12: 13211491-13211491 on Build GRCh38
rs571697910 Chr.12: 13211744-13211744 on Build GRCh38
rs771020753 Chr.12: 13211500-13211500 on Build GRCh38
rs147023177 Chr.12: 13211550-13211550 on Build GRCh38
rs537630456 Chr.12: 13211552-13211552 on Build GRCh38
rs778481106 Chr.12: 13211553-13211553 on Build GRCh38
rs776776515 Chr.12: 13211617-13211617 on Build GRCh38
rs774231293 Chr.12: 13211451-13211454 on Build GRCh38
rs747959879 Chr.12: 13211554-13211554 on Build GRCh38
rs755632922 Chr.12: 13211548-13211548 on Build GRCh38
rs191886304 Chr.12: 13211796-13211796 on Build GRCh38
rs553657382 Chr.12: 13211729-13211729 on Build GRCh38
rs1012672713 Chr.12: 13211414-13211414 on Build GRCh38
rs1026499119 Chr.12: 13211808-13211808 on Build GRCh38
rs71530953 Chr.12: 13211501-13211501 on Build GRCh38
rs576598748 Chr.12: 13211866-13211866 on Build GRCh38
rs909148736 Chr.12: 13211679-13211679 on Build GRCh38
rs772013058 Chr.12: 13211466-13211466 on Build GRCh38
rs147282605 Chr.12: 13211542-13211542 on Build GRCh38
rs34264613 Chr.12: 13211666-13211667 on Build GRCh38
rs759797796 Chr.12: 13211506-13211506 on Build GRCh38
rs1034343398 Chr.12: 13211871-13211871 on Build GRCh38
rs746951833 Chr.12: 13211567-13211567 on Build GRCh38
rs557969847 Chr.12: 13211549-13211549 on Build GRCh38
rs745874252 Chr.12: 13211619-13211619 on Build GRCh38
rs972007785 Chr.12: 13211417-13211417 on Build GRCh38
rs147635640 Chr.12: 13211792-13211792 on Build GRCh38
rs73282992 Chr.12: 13211684-13211684 on Build GRCh38
rs561332299 Chr.12: 13211492-13211493 on Build GRCh38
rs770160792 Chr.12: 13211630-13211630 on Build GRCh38
rs746233006 Chr.12: 13211575-13211575 on Build GRCh38
rs765487297 Chr.12: 13211530-13211530 on Build GRCh38
rs1006433225 Chr.12: 13211839-13211839 on Build GRCh38
rs562217820 Chr.12: 13211842-13211842 on Build GRCh38
rs773084460 Chr.12: 13211472-13211472 on Build GRCh38
rs891519954 Chr.12: 13211811-13211811 on Build GRCh38
rs188160823 Chr.12: 13211646-13211646 on Build GRCh38
rs541673675 Chr.12: 13211727-13211727 on Build GRCh38
rs774783232 Chr.12: 13211635-13211635 on Build GRCh38
rs753122550 Chr.12: 13211531-13211531 on Build GRCh38
rs531469403 Chr.12: 13211825-13211825 on Build GRCh38
rs1011233284 Chr.12: 13211416-13211416 on Build GRCh38
rs747873381 Chr.12: 13211461-13211461 on Build GRCh38
rs879177159 Chr.12: 13211471-13211471 on Build GRCh38
rs779051423 Chr.12: 13211493-13211494 on Build GRCh38
rs764207614 Chr.12: 13211493-13211493 on Build GRCh38
rs373481556 Chr.12: 13211632-13211632 on Build GRCh38

View Full Product Details