Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13213591-13214098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP1
Entrez Gene ID
2012
Gene Name
epithelial membrane protein 1
Gene Aliases
CL-20, EMP-1, TMP
UniGene
Hs.719042
Gene Chromosome Location
Chr.12: 13196668-13216774 on Build GRCh38
Transcripts
NM_001423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653879 c.186G>T p.K62N Chr.12: 13213691-13213691 on Build GRCh38
3458426 c.259T>G p.F87V Chr.12: 13213764-13213764 on Build GRCh38
6040921 c.282G>T p.R94R Chr.12: 13213787-13213787 on Build GRCh38
5970617 c.203T>C p.M68T Chr.12: 13213708-13213708 on Build GRCh38
3458425 c.219C>T p.I73I Chr.12: 13213724-13213724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768027737 Chr.12: 13213812-13213812 on Build GRCh38
rs530800191 Chr.12: 13213889-13213889 on Build GRCh38
rs533130206 Chr.12: 13213799-13213799 on Build GRCh38
rs764712996 Chr.12: 13213623-13213623 on Build GRCh38
rs199851602 Chr.12: 13213710-13213710 on Build GRCh38
rs747638498 Chr.12: 13213954-13213954 on Build GRCh38
rs753888611 Chr.12: 13214062-13214062 on Build GRCh38
rs906592749 Chr.12: 13213634-13213634 on Build GRCh38
rs774107526 Chr.12: 13213813-13213813 on Build GRCh38
rs750260641 Chr.12: 13213835-13213835 on Build GRCh38
rs927962043 Chr.12: 13213955-13213955 on Build GRCh38
rs553028446 Chr.12: 13214029-13214029 on Build GRCh38
rs74690704 Chr.12: 13213947-13213947 on Build GRCh38
rs898822877 Chr.12: 13213764-13213764 on Build GRCh38
rs761713014 Chr.12: 13213733-13213733 on Build GRCh38
rs747317366 Chr.12: 13213791-13213791 on Build GRCh38
rs763594070 Chr.12: 13213635-13213635 on Build GRCh38
rs780281141 Chr.12: 13213666-13213666 on Build GRCh38
rs2277395 Chr.12: 13213665-13213665 on Build GRCh38
rs761164918 Chr.12: 13213707-13213707 on Build GRCh38
rs769015847 Chr.12: 13213675-13213675 on Build GRCh38
rs760587318 Chr.12: 13213996-13213996 on Build GRCh38
rs759510100 Chr.12: 13213931-13213931 on Build GRCh38
rs201405991 Chr.12: 13213752-13213752 on Build GRCh38
rs761370248 Chr.12: 13213815-13213815 on Build GRCh38
rs781697269 Chr.12: 13213943-13213943 on Build GRCh38
rs753796132 Chr.12: 13213879-13213879 on Build GRCh38
rs569851146 Chr.12: 13213872-13213872 on Build GRCh38
rs567228642 Chr.12: 13213964-13213964 on Build GRCh38
rs752753834 Chr.12: 13213907-13213907 on Build GRCh38
rs866257560 Chr.12: 13213866-13213866 on Build GRCh38
rs890784702 Chr.12: 13214042-13214042 on Build GRCh38
rs747528195 Chr.12: 13213969-13213969 on Build GRCh38
rs534642100 Chr.12: 13213995-13213995 on Build GRCh38
rs1029676776 Chr.12: 13213649-13213649 on Build GRCh38
rs773741839 Chr.12: 13213950-13213950 on Build GRCh38
rs370911293 Chr.12: 13213734-13213734 on Build GRCh38
rs1019604221 Chr.12: 13214071-13214071 on Build GRCh38
rs551322995 Chr.12: 13213828-13213828 on Build GRCh38
rs781225244 Chr.12: 13213648-13213648 on Build GRCh38
rs554148363 Chr.12: 13213795-13213795 on Build GRCh38
rs777922457 Chr.12: 13213912-13213912 on Build GRCh38
rs762485781 Chr.12: 13213807-13213807 on Build GRCh38
rs139944204 Chr.12: 13213768-13213768 on Build GRCh38
rs755137293 Chr.12: 13213887-13213887 on Build GRCh38
rs749316611 Chr.12: 13213668-13213668 on Build GRCh38
rs751139207 Chr.12: 13213722-13213722 on Build GRCh38
rs780084917 Chr.12: 13213751-13213751 on Build GRCh38
rs779235341 Chr.12: 13213767-13213767 on Build GRCh38
rs752412944 Chr.12: 13213625-13213625 on Build GRCh38
rs374294823 Chr.12: 13213785-13213785 on Build GRCh38
rs543426840 Chr.12: 13213914-13213914 on Build GRCh38
rs759087942 Chr.12: 13213617-13213617 on Build GRCh38
rs201135816 Chr.12: 13213708-13213708 on Build GRCh38
rs748578842 Chr.12: 13213684-13213684 on Build GRCh38
rs565284070 Chr.12: 13213784-13213784 on Build GRCh38
rs776769907 Chr.12: 13214027-13214027 on Build GRCh38
rs758048273 Chr.12: 13213633-13213633 on Build GRCh38
rs528476175 Chr.12: 13213717-13213717 on Build GRCh38
rs967565094 Chr.12: 13214073-13214073 on Build GRCh38
rs766452292 Chr.12: 13213873-13213873 on Build GRCh38
rs886304299 Chr.12: 13213720-13213720 on Build GRCh38
rs751390049 Chr.12: 13213639-13213639 on Build GRCh38
rs1047629512 Chr.12: 13214001-13214001 on Build GRCh38
rs764118134 Chr.12: 13213944-13213944 on Build GRCh38
rs1009187722 Chr.12: 13214070-13214070 on Build GRCh38
rs780443287 Chr.12: 13213903-13213903 on Build GRCh38
rs772293765 Chr.12: 13213695-13213695 on Build GRCh38
rs773819918 Chr.12: 13213699-13213699 on Build GRCh38
rs771562237 Chr.12: 13213736-13213736 on Build GRCh38
rs771449249 Chr.12: 13213796-13213796 on Build GRCh38
rs915820160 Chr.12: 13213896-13213896 on Build GRCh38
rs531986239 Chr.12: 13213662-13213662 on Build GRCh38
rs763836564 Chr.12: 13213719-13213719 on Build GRCh38
rs771649784 Chr.12: 13213961-13213961 on Build GRCh38
rs773551996 Chr.12: 13213614-13213614 on Build GRCh38
rs756960046 Chr.12: 13213643-13213643 on Build GRCh38
rs758560645 Chr.12: 13213908-13213908 on Build GRCh38
rs765163037 Chr.12: 13214030-13214030 on Build GRCh38
rs949957632 Chr.12: 13213909-13213909 on Build GRCh38
rs766254422 Chr.12: 13214000-13214000 on Build GRCh38
rs778955534 Chr.12: 13213678-13213678 on Build GRCh38
rs756129778 Chr.12: 13213742-13213742 on Build GRCh38
rs200946352 Chr.12: 13213655-13213655 on Build GRCh38

View Full Product Details