Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13214816-13215321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP1
Entrez Gene ID
2012
Gene Name
epithelial membrane protein 1
Gene Aliases
CL-20, EMP-1, TMP
UniGene
Hs.719042
Gene Chromosome Location
Chr.12: 13196668-13216774 on Build GRCh38
Transcripts
NM_001423.2

SNP Detail

SNP ID Chromosome Location
rs117660498 Chr.12: 13215090-13215090 on Build GRCh38
rs893354750 Chr.12: 13214906-13214906 on Build GRCh38
rs375858526 Chr.12: 13214917-13214917 on Build GRCh38
rs954066941 Chr.12: 13214997-13214997 on Build GRCh38
rs1002948507 Chr.12: 13214941-13214941 on Build GRCh38
rs936305916 Chr.12: 13214901-13214901 on Build GRCh38
rs1042155529 Chr.12: 13214861-13214861 on Build GRCh38
rs1004871270 Chr.12: 13215075-13215075 on Build GRCh38
rs923665861 Chr.12: 13215068-13215068 on Build GRCh38
rs574475522 Chr.12: 13215258-13215258 on Build GRCh38
rs912422836 Chr.12: 13215023-13215023 on Build GRCh38
rs1052390080 Chr.12: 13214947-13214947 on Build GRCh38
rs961382543 Chr.12: 13214964-13214964 on Build GRCh38
rs1015619626 Chr.12: 13214958-13214958 on Build GRCh38
rs937779216 Chr.12: 13215048-13215048 on Build GRCh38
rs577174368 Chr.12: 13215099-13215099 on Build GRCh38
rs1015824063 Chr.12: 13215084-13215084 on Build GRCh38
rs954234272 Chr.12: 13215136-13215136 on Build GRCh38
rs1028280927 Chr.12: 13215114-13215114 on Build GRCh38
rs1054734401 Chr.12: 13214904-13214904 on Build GRCh38
rs904917446 Chr.12: 13215022-13215022 on Build GRCh38
rs948945713 Chr.12: 13215182-13215182 on Build GRCh38
rs544343002 Chr.12: 13215156-13215156 on Build GRCh38
rs913551089 Chr.12: 13214977-13214980 on Build GRCh38
rs995441718 Chr.12: 13215113-13215113 on Build GRCh38
rs988043766 Chr.12: 13215235-13215235 on Build GRCh38
rs911735189 Chr.12: 13214908-13214908 on Build GRCh38
rs147272464 Chr.12: 13214970-13214970 on Build GRCh38
rs1053419307 Chr.12: 13215093-13215093 on Build GRCh38
rs57151560 Chr.12: 13214979-13214979 on Build GRCh38
rs985315593 Chr.12: 13215004-13215004 on Build GRCh38
rs759438938 Chr.12: 13215042-13215042 on Build GRCh38
rs907393657 Chr.12: 13215213-13215213 on Build GRCh38
rs866849734 Chr.12: 13215237-13215237 on Build GRCh38
rs1043424994 Chr.12: 13215007-13215007 on Build GRCh38
rs979053227 Chr.12: 13215260-13215260 on Build GRCh38
rs977863797 Chr.12: 13215065-13215065 on Build GRCh38
rs371899137 Chr.12: 13215011-13215011 on Build GRCh38
rs771395242 Chr.12: 13214881-13214881 on Build GRCh38
rs965197482 Chr.12: 13215050-13215050 on Build GRCh38
rs78423139 Chr.12: 13215256-13215256 on Build GRCh38
rs1044645162 Chr.12: 13215184-13215184 on Build GRCh38
rs907287167 Chr.12: 13214937-13214937 on Build GRCh38
rs370504746 Chr.12: 13215293-13215293 on Build GRCh38
rs965928312 Chr.12: 13215205-13215205 on Build GRCh38
rs1044924252 Chr.12: 13214931-13214931 on Build GRCh38
rs1057001368 Chr.12: 13215054-13215054 on Build GRCh38
rs748933752 Chr.12: 13214877-13214877 on Build GRCh38

View Full Product Details