Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13215605-13216109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP1
Entrez Gene ID
2012
Gene Name
epithelial membrane protein 1
Gene Aliases
CL-20, EMP-1, TMP
UniGene
Hs.719042
Gene Chromosome Location
Chr.12: 13196668-13216774 on Build GRCh38
Transcripts
NM_001423.2

SNP Detail

SNP ID Chromosome Location
rs981772672 Chr.12: 13216007-13216007 on Build GRCh38
rs372242453 Chr.12: 13215944-13215944 on Build GRCh38
rs1010333183 Chr.12: 13215872-13215872 on Build GRCh38
rs777416820 Chr.12: 13215937-13215937 on Build GRCh38
rs891846602 Chr.12: 13215843-13215843 on Build GRCh38
rs967913777 Chr.12: 13215918-13215918 on Build GRCh38
rs559209377 Chr.12: 13215979-13215979 on Build GRCh38
rs944290462 Chr.12: 13216066-13216066 on Build GRCh38
rs1050032036 Chr.12: 13215786-13215786 on Build GRCh38
rs182583968 Chr.12: 13215900-13215900 on Build GRCh38
rs889770223 Chr.12: 13215789-13215789 on Build GRCh38
rs568638752 Chr.12: 13215650-13215650 on Build GRCh38
rs1030800712 Chr.12: 13215909-13215909 on Build GRCh38
rs561037465 Chr.12: 13216051-13216051 on Build GRCh38
rs538286964 Chr.12: 13215986-13215986 on Build GRCh38
rs1020792599 Chr.12: 13215891-13215891 on Build GRCh38
rs999365960 Chr.12: 13215889-13215889 on Build GRCh38
rs548339571 Chr.12: 13215816-13215816 on Build GRCh38
rs897250859 Chr.12: 13215636-13215636 on Build GRCh38
rs939794468 Chr.12: 13215671-13215671 on Build GRCh38
rs991817950 Chr.12: 13215990-13215990 on Build GRCh38
rs577162250 Chr.12: 13215982-13215982 on Build GRCh38
rs191016373 Chr.12: 13215696-13215696 on Build GRCh38
rs959566934 Chr.12: 13215972-13215972 on Build GRCh38
rs1008199399 Chr.12: 13215802-13215802 on Build GRCh38
rs931560301 Chr.12: 13215729-13215729 on Build GRCh38
rs961102433 Chr.12: 13216017-13216017 on Build GRCh38
rs918791902 Chr.12: 13216037-13216037 on Build GRCh38
rs3741814 Chr.12: 13216013-13216013 on Build GRCh38
rs56315027 Chr.12: 13216052-13216052 on Build GRCh38
rs755741828 Chr.12: 13215742-13215742 on Build GRCh38
rs931439060 Chr.12: 13215667-13215667 on Build GRCh38
rs1048463915 Chr.12: 13215669-13215669 on Build GRCh38
rs1041460624 Chr.12: 13215831-13215831 on Build GRCh38
rs7966078 Chr.12: 13216036-13216036 on Build GRCh38
rs1037124715 Chr.12: 13215681-13215681 on Build GRCh38
rs901060350 Chr.12: 13215826-13215826 on Build GRCh38
rs969480780 Chr.12: 13215983-13215983 on Build GRCh38
rs992307071 Chr.12: 13215633-13215633 on Build GRCh38
rs1019965896 Chr.12: 13215745-13215745 on Build GRCh38
rs754348960 Chr.12: 13215873-13215873 on Build GRCh38
rs938898533 Chr.12: 13216030-13216030 on Build GRCh38
rs147645678 Chr.12: 13215938-13215938 on Build GRCh38
rs1006873181 Chr.12: 13215712-13215712 on Build GRCh38

View Full Product Details