Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 28257036-28257552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC91
Entrez Gene ID
55297
Gene Name
coiled-coil domain containing 91
Gene Aliases
HSD8, p56
UniGene
Hs.653125
Gene Chromosome Location
Chr.12: 28190427-28550166 on Build GRCh38
Transcripts
NM_018318.3 XM_005253413.1 XM_005253414.1 XM_006719104.2 XM_006719105.1 XM_011520729.2 XM_011520734.1 XM_011520735.1 XM_017019569.1 XM_017019570.1 XM_017019571.1 XM_017019572.1 XM_017019574.1 XM_017019575.1 XM_017019576.1 XM_017019577.1

SNP Detail

SNP ID Chromosome Location
rs985122775 Chr.12: 28257394-28257395 on Build GRCh38
rs557308944 Chr.12: 28257400-28257400 on Build GRCh38
rs915164946 Chr.12: 28257090-28257090 on Build GRCh38
rs755754836 Chr.12: 28257317-28257317 on Build GRCh38
rs765261582 Chr.12: 28257084-28257084 on Build GRCh38
rs749207099 Chr.12: 28257369-28257369 on Build GRCh38
rs182331354 Chr.12: 28257174-28257174 on Build GRCh38
rs773085183 Chr.12: 28257220-28257220 on Build GRCh38
rs770027833 Chr.12: 28257190-28257190 on Build GRCh38
rs376740214 Chr.12: 28257164-28257164 on Build GRCh38
rs771939694 Chr.12: 28257213-28257213 on Build GRCh38
rs533029143 Chr.12: 28257121-28257121 on Build GRCh38
rs35669227 Chr.12: 28257239-28257240 on Build GRCh38
rs953707368 Chr.12: 28257085-28257085 on Build GRCh38
rs760232977 Chr.12: 28257261-28257261 on Build GRCh38
rs201689336 Chr.12: 28257171-28257171 on Build GRCh38
rs569197899 Chr.12: 28257409-28257409 on Build GRCh38
rs985341325 Chr.12: 28257086-28257086 on Build GRCh38
rs765358410 Chr.12: 28257260-28257261 on Build GRCh38
rs755341866 Chr.12: 28257168-28257168 on Build GRCh38
rs547777639 Chr.12: 28257399-28257400 on Build GRCh38
rs756947120 Chr.12: 28257294-28257294 on Build GRCh38
rs1021998340 Chr.12: 28257400-28257403 on Build GRCh38
rs761875975 Chr.12: 28257189-28257190 on Build GRCh38
rs537781392 Chr.12: 28257301-28257301 on Build GRCh38
rs778525105 Chr.12: 28257265-28257266 on Build GRCh38
rs751186808 Chr.12: 28257193-28257193 on Build GRCh38
rs539919862 Chr.12: 28257445-28257445 on Build GRCh38
rs923819525 Chr.12: 28257129-28257129 on Build GRCh38
rs76960133 Chr.12: 28257128-28257128 on Build GRCh38
rs142339420 Chr.12: 28257172-28257172 on Build GRCh38
rs978051957 Chr.12: 28257431-28257431 on Build GRCh38
rs781471141 Chr.12: 28257183-28257183 on Build GRCh38
rs1029427261 Chr.12: 28257366-28257366 on Build GRCh38
rs747941577 Chr.12: 28257204-28257204 on Build GRCh38
rs186880281 Chr.12: 28257496-28257496 on Build GRCh38
rs185398626 Chr.12: 28257118-28257118 on Build GRCh38
rs529627588 Chr.12: 28257066-28257066 on Build GRCh38
rs998316120 Chr.12: 28257333-28257333 on Build GRCh38
rs773570236 Chr.12: 28257262-28257263 on Build GRCh38
rs751973321 Chr.12: 28257263-28257264 on Build GRCh38
rs775608124 Chr.12: 28257328-28257328 on Build GRCh38
rs747052379 Chr.12: 28257173-28257173 on Build GRCh38
rs759390052 Chr.12: 28257070-28257070 on Build GRCh38
rs34981517 Chr.12: 28257351-28257352 on Build GRCh38
rs201007722 Chr.12: 28257184-28257184 on Build GRCh38
rs190601871 Chr.12: 28257170-28257170 on Build GRCh38
rs147677606 Chr.12: 28257094-28257094 on Build GRCh38
rs765849230 Chr.12: 28257287-28257287 on Build GRCh38
rs780267626 Chr.12: 28257390-28257390 on Build GRCh38
rs774094072 Chr.12: 28257202-28257202 on Build GRCh38

View Full Product Details