Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 29349830-29350244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC2
Entrez Gene ID
51290
Gene Name
ERGIC and golgi 2
Gene Aliases
CDA14, Erv41, PTX1, cd002
UniGene
Hs.339453
Gene Chromosome Location
Chr.12: 29336544-29381234 on Build GRCh38
Transcripts
NM_016570.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979169 c.622C>G p.H208D Chr.12: 29350019-29350019 on Build GRCh38
3980989 c.578T>A p.I193N Chr.12: 29350063-29350063 on Build GRCh38
938551 c.590G>A p.R197H Chr.12: 29350051-29350051 on Build GRCh38
6421642 c.574G>T p.A192S Chr.12: 29350067-29350067 on Build GRCh38
2204764 c.623A>G p.H208R Chr.12: 29350018-29350018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753844760 Chr.12: 29350086-29350086 on Build GRCh38
rs374870823 Chr.12: 29350084-29350085 on Build GRCh38
rs989257985 Chr.12: 29349990-29349990 on Build GRCh38
rs12580072 Chr.12: 29349919-29349919 on Build GRCh38
rs567720959 Chr.12: 29350219-29350219 on Build GRCh38
rs756543949 Chr.12: 29350070-29350070 on Build GRCh38
rs928254533 Chr.12: 29350050-29350050 on Build GRCh38
rs759911340 Chr.12: 29349943-29349943 on Build GRCh38
rs768751762 Chr.12: 29349985-29349985 on Build GRCh38
rs549511163 Chr.12: 29350180-29350180 on Build GRCh38
rs917152866 Chr.12: 29349979-29349979 on Build GRCh38
rs769812628 Chr.12: 29350021-29350021 on Build GRCh38
rs1047808243 Chr.12: 29350141-29350141 on Build GRCh38
rs139010014 Chr.12: 29350094-29350094 on Build GRCh38
rs762583077 Chr.12: 29350037-29350037 on Build GRCh38
rs748390176 Chr.12: 29349995-29349995 on Build GRCh38
rs749422425 Chr.12: 29349983-29349983 on Build GRCh38
rs373369156 Chr.12: 29349990-29349990 on Build GRCh38
rs560615928 Chr.12: 29349972-29349972 on Build GRCh38
rs977348823 Chr.12: 29350029-29350029 on Build GRCh38
rs770812888 Chr.12: 29350001-29350001 on Build GRCh38
rs761483514 Chr.12: 29350051-29350051 on Build GRCh38
rs924405076 Chr.12: 29350074-29350074 on Build GRCh38
rs772473372 Chr.12: 29350109-29350109 on Build GRCh38
rs531219614 Chr.12: 29350061-29350061 on Build GRCh38
rs868648895 Chr.12: 29350112-29350112 on Build GRCh38
rs529884139 Chr.12: 29350173-29350173 on Build GRCh38
rs34762815 Chr.12: 29349896-29349897 on Build GRCh38
rs887238523 Chr.12: 29350206-29350206 on Build GRCh38
rs1025861960 Chr.12: 29350203-29350203 on Build GRCh38
rs965278937 Chr.12: 29349917-29349917 on Build GRCh38
rs182909183 Chr.12: 29350136-29350136 on Build GRCh38
rs779554834 Chr.12: 29349966-29349967 on Build GRCh38
rs763525481 Chr.12: 29350044-29350044 on Build GRCh38
rs746500925 Chr.12: 29349990-29349993 on Build GRCh38
rs983854202 Chr.12: 29349858-29349858 on Build GRCh38
rs942700959 Chr.12: 29350143-29350143 on Build GRCh38
rs865954419 Chr.12: 29350042-29350042 on Build GRCh38
rs751144640 Chr.12: 29350045-29350045 on Build GRCh38
rs549322300 Chr.12: 29350075-29350075 on Build GRCh38
rs1018940499 Chr.12: 29350205-29350205 on Build GRCh38
rs767819328 Chr.12: 29350052-29350052 on Build GRCh38
rs958396825 Chr.12: 29349888-29349888 on Build GRCh38
rs909626939 Chr.12: 29349879-29349879 on Build GRCh38
rs921079124 Chr.12: 29349956-29349956 on Build GRCh38
rs998365908 Chr.12: 29350197-29350197 on Build GRCh38
rs759526033 Chr.12: 29350019-29350019 on Build GRCh38
rs141169455 Chr.12: 29349852-29349852 on Build GRCh38
rs376348079 Chr.12: 29350004-29350004 on Build GRCh38
rs768272936 Chr.12: 29350078-29350079 on Build GRCh38
rs1040135623 Chr.12: 29350146-29350146 on Build GRCh38
rs778894342 Chr.12: 29350093-29350093 on Build GRCh38
rs1008930639 Chr.12: 29350182-29350182 on Build GRCh38
rs781093765 Chr.12: 29349908-29349909 on Build GRCh38
rs565082336 Chr.12: 29350091-29350091 on Build GRCh38
rs774859065 Chr.12: 29350024-29350024 on Build GRCh38
rs372031276 Chr.12: 29349994-29349994 on Build GRCh38
rs868023274 Chr.12: 29350060-29350060 on Build GRCh38
rs547309543 Chr.12: 29350119-29350119 on Build GRCh38

View Full Product Details