Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 29356368-29356877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC2
Entrez Gene ID
51290
Gene Name
ERGIC and golgi 2
Gene Aliases
CDA14, Erv41, PTX1, cd002
UniGene
Hs.339453
Gene Chromosome Location
Chr.12: 29336544-29381234 on Build GRCh38
Transcripts
NM_016570.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
938552 c.546A>G p.A182A Chr.12: 29356408-29356408 on Build GRCh38
4922059 c.535A>G p.N179D Chr.12: 29356419-29356419 on Build GRCh38
468248 c.482A>C p.D161A Chr.12: 29356472-29356472 on Build GRCh38
7086864 c.563C>T p.T188I Chr.12: 29356391-29356391 on Build GRCh38
7086737 c.564delA p.V189fs*29 Chr.12: 29356390-29356390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559976107 Chr.12: 29356763-29356763 on Build GRCh38
rs1032595639 Chr.12: 29356743-29356743 on Build GRCh38
rs958854756 Chr.12: 29356615-29356615 on Build GRCh38
rs1018012234 Chr.12: 29356547-29356547 on Build GRCh38
rs751935202 Chr.12: 29356625-29356625 on Build GRCh38
rs543011698 Chr.12: 29356619-29356619 on Build GRCh38
rs144781775 Chr.12: 29356759-29356763 on Build GRCh38
rs766720902 Chr.12: 29356512-29356512 on Build GRCh38
rs893966176 Chr.12: 29356767-29356767 on Build GRCh38
rs755870893 Chr.12: 29356470-29356470 on Build GRCh38
rs116024345 Chr.12: 29356774-29356774 on Build GRCh38
rs892639790 Chr.12: 29356780-29356780 on Build GRCh38
rs1034300131 Chr.12: 29356596-29356596 on Build GRCh38
rs779744074 Chr.12: 29356480-29356480 on Build GRCh38
rs753247020 Chr.12: 29356791-29356791 on Build GRCh38
rs951159167 Chr.12: 29356630-29356630 on Build GRCh38
rs377371261 Chr.12: 29356426-29356426 on Build GRCh38
rs769670164 Chr.12: 29356516-29356517 on Build GRCh38
rs573611030 Chr.12: 29356566-29356566 on Build GRCh38
rs371361855 Chr.12: 29356460-29356460 on Build GRCh38
rs1035606 Chr.12: 29356517-29356517 on Build GRCh38
rs768522622 Chr.12: 29356482-29356482 on Build GRCh38
rs760280959 Chr.12: 29356500-29356500 on Build GRCh38
rs771696212 Chr.12: 29356494-29356494 on Build GRCh38
rs765388570 Chr.12: 29356435-29356435 on Build GRCh38
rs1035607 Chr.12: 29356580-29356580 on Build GRCh38
rs754333757 Chr.12: 29356418-29356418 on Build GRCh38
rs766662864 Chr.12: 29356413-29356413 on Build GRCh38
rs558118548 Chr.12: 29356535-29356535 on Build GRCh38
rs912239078 Chr.12: 29356461-29356461 on Build GRCh38
rs1007041467 Chr.12: 29356771-29356771 on Build GRCh38
rs975469693 Chr.12: 29356744-29356744 on Build GRCh38
rs773816720 Chr.12: 29356487-29356487 on Build GRCh38
rs900052096 Chr.12: 29356544-29356544 on Build GRCh38
rs1002234493 Chr.12: 29356819-29356819 on Build GRCh38
rs752866903 Chr.12: 29356443-29356443 on Build GRCh38
rs1046806809 Chr.12: 29356685-29356685 on Build GRCh38
rs936881304 Chr.12: 29356551-29356552 on Build GRCh38
rs761879507 Chr.12: 29356447-29356459 on Build GRCh38
rs370651745 Chr.12: 29356597-29356597 on Build GRCh38
rs573645691 Chr.12: 29356474-29356474 on Build GRCh38
rs976846498 Chr.12: 29356562-29356562 on Build GRCh38
rs949396132 Chr.12: 29356666-29356666 on Build GRCh38
rs758690086 Chr.12: 29356459-29356459 on Build GRCh38
rs1054362189 Chr.12: 29356552-29356552 on Build GRCh38
rs909679112 Chr.12: 29356583-29356583 on Build GRCh38
rs142711950 Chr.12: 29356706-29356706 on Build GRCh38
rs1046479558 Chr.12: 29356783-29356784 on Build GRCh38
rs906562692 Chr.12: 29356577-29356577 on Build GRCh38
rs906598207 Chr.12: 29356783-29356783 on Build GRCh38
rs895261737 Chr.12: 29356649-29356649 on Build GRCh38
rs965058172 Chr.12: 29356560-29356560 on Build GRCh38
rs999801300 Chr.12: 29356447-29356447 on Build GRCh38
rs749198696 Chr.12: 29356481-29356481 on Build GRCh38
rs912289649 Chr.12: 29356714-29356714 on Build GRCh38
rs759728554 Chr.12: 29356647-29356647 on Build GRCh38
rs562488553 Chr.12: 29356719-29356719 on Build GRCh38
rs945524907 Chr.12: 29356783-29356783 on Build GRCh38
rs965476844 Chr.12: 29356838-29356838 on Build GRCh38
rs1000145425 Chr.12: 29356730-29356730 on Build GRCh38
rs750473699 Chr.12: 29356395-29356395 on Build GRCh38
rs1034093121 Chr.12: 29356842-29356842 on Build GRCh38
rs1024977965 Chr.12: 29356567-29356567 on Build GRCh38
rs376107477 Chr.12: 29356498-29356498 on Build GRCh38
rs182849581 Chr.12: 29356628-29356628 on Build GRCh38
rs747720252 Chr.12: 29356490-29356490 on Build GRCh38
rs1032437601 Chr.12: 29356457-29356457 on Build GRCh38
rs138714900 Chr.12: 29356572-29356572 on Build GRCh38
rs982529246 Chr.12: 29356707-29356707 on Build GRCh38
rs760059564 Chr.12: 29356524-29356524 on Build GRCh38
rs937199709 Chr.12: 29356618-29356618 on Build GRCh38
rs200612847 Chr.12: 29356468-29356468 on Build GRCh38

View Full Product Details