Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47977465-47977962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL2A1
Entrez Gene ID
1280
Gene Name
collagen type II alpha 1 chain
Gene Aliases
ANFH, AOM, COL11A3, SEDC, STL1
UniGene
Hs.408182
Gene Chromosome Location
Chr.12: 47972965-48006212 on Build GRCh38
Transcripts
NM_001844.4 NM_033150.2 XM_017018828.1 XM_017018829.1 XM_017018830.1 XM_017018831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4967487 c.3145G>A p.D1049N Chr.12: 47977620-47977620 on Build GRCh38
4575422 c.3119C>T p.P1040L Chr.12: 47977646-47977646 on Build GRCh38
7233282 c.3112-1G>T p.? Chr.12: 47977654-47977654 on Build GRCh38
6632994 c.2946T>G p.A982A Chr.12: 47977612-47977612 on Build GRCh38
4575423 c.2912C>T p.P971L Chr.12: 47977646-47977646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749864741 Chr.12: 47977918-47977918 on Build GRCh38
rs562501356 Chr.12: 47977732-47977732 on Build GRCh38
rs748369672 Chr.12: 47977703-47977703 on Build GRCh38
rs376722458 Chr.12: 47977615-47977615 on Build GRCh38
rs949630325 Chr.12: 47977514-47977514 on Build GRCh38
rs774693733 Chr.12: 47977579-47977579 on Build GRCh38
rs41272745 Chr.12: 47977557-47977557 on Build GRCh38
rs756676390 Chr.12: 47977647-47977647 on Build GRCh38
rs1046621638 Chr.12: 47977516-47977516 on Build GRCh38
rs773348184 Chr.12: 47977705-47977705 on Build GRCh38
rs916906232 Chr.12: 47977496-47977496 on Build GRCh38
rs372190657 Chr.12: 47977683-47977683 on Build GRCh38
rs772366228 Chr.12: 47977704-47977704 on Build GRCh38
rs918882075 Chr.12: 47977755-47977755 on Build GRCh38
rs576884461 Chr.12: 47977849-47977849 on Build GRCh38
rs917303430 Chr.12: 47977894-47977894 on Build GRCh38
rs769023701 Chr.12: 47977576-47977576 on Build GRCh38
rs780375782 Chr.12: 47977650-47977650 on Build GRCh38
rs542335888 Chr.12: 47977497-47977497 on Build GRCh38
rs555530274 Chr.12: 47977799-47977799 on Build GRCh38
rs781510115 Chr.12: 47977834-47977834 on Build GRCh38
rs756815438 Chr.12: 47977632-47977632 on Build GRCh38
rs540124604 Chr.12: 47977892-47977892 on Build GRCh38
rs767701661 Chr.12: 47977599-47977599 on Build GRCh38
rs56265470 Chr.12: 47977937-47977937 on Build GRCh38
rs1048913517 Chr.12: 47977506-47977506 on Build GRCh38
rs551840502 Chr.12: 47977706-47977706 on Build GRCh38
rs751205887 Chr.12: 47977629-47977629 on Build GRCh38
rs775927882 Chr.12: 47977568-47977568 on Build GRCh38
rs758162798 Chr.12: 47977621-47977621 on Build GRCh38
rs41272041 Chr.12: 47977614-47977614 on Build GRCh38
rs562049171 Chr.12: 47977887-47977887 on Build GRCh38
rs901151450 Chr.12: 47977580-47977580 on Build GRCh38
rs756202836 Chr.12: 47977891-47977891 on Build GRCh38
rs573313146 Chr.12: 47977740-47977740 on Build GRCh38
rs371635111 Chr.12: 47977605-47977605 on Build GRCh38
rs773138674 Chr.12: 47977593-47977596 on Build GRCh38
rs749834758 Chr.12: 47977642-47977642 on Build GRCh38
rs949708401 Chr.12: 47977719-47977719 on Build GRCh38
rs769943171 Chr.12: 47977563-47977563 on Build GRCh38
rs41272043 Chr.12: 47977561-47977561 on Build GRCh38
rs1793949 Chr.12: 47977812-47977812 on Build GRCh38
rs757301359 Chr.12: 47977800-47977802 on Build GRCh38
rs1028577544 Chr.12: 47977847-47977847 on Build GRCh38
rs752571750 Chr.12: 47977620-47977620 on Build GRCh38
rs756720243 Chr.12: 47977551-47977551 on Build GRCh38
rs41272747 Chr.12: 47977520-47977520 on Build GRCh38
rs878872514 Chr.12: 47977877-47977877 on Build GRCh38
rs984376025 Chr.12: 47977878-47977878 on Build GRCh38
rs746371369 Chr.12: 47977560-47977560 on Build GRCh38
rs939978406 Chr.12: 47977888-47977888 on Build GRCh38
rs776493223 Chr.12: 47977606-47977606 on Build GRCh38
rs932677268 Chr.12: 47977500-47977504 on Build GRCh38
rs749711774 Chr.12: 47977677-47977677 on Build GRCh38
rs954474471 Chr.12: 47977864-47977864 on Build GRCh38
rs121912873 Chr.12: 47977627-47977627 on Build GRCh38
rs547693952 Chr.12: 47977865-47977865 on Build GRCh38
rs1635558 Chr.12: 47977700-47977700 on Build GRCh38
rs1039739197 Chr.12: 47977850-47977850 on Build GRCh38
rs749574390 Chr.12: 47977573-47977573 on Build GRCh38
rs57550917 Chr.12: 47977896-47977896 on Build GRCh38
rs746042284 Chr.12: 47977838-47977838 on Build GRCh38
rs763772608 Chr.12: 47977628-47977628 on Build GRCh38
rs568338857 Chr.12: 47977822-47977822 on Build GRCh38
rs371697518 Chr.12: 47977586-47977586 on Build GRCh38
rs121912868 Chr.12: 47977607-47977607 on Build GRCh38
rs367982631 Chr.12: 47977645-47977645 on Build GRCh38
rs901184048 Chr.12: 47977853-47977853 on Build GRCh38
rs998163211 Chr.12: 47977862-47977862 on Build GRCh38
rs891451358 Chr.12: 47977810-47977810 on Build GRCh38
rs780693364 Chr.12: 47977633-47977633 on Build GRCh38
rs1018257494 Chr.12: 47977567-47977567 on Build GRCh38
rs368647010 Chr.12: 47977549-47977549 on Build GRCh38
rs958473926 Chr.12: 47977695-47977695 on Build GRCh38
rs532142721 Chr.12: 47977678-47977678 on Build GRCh38
rs888600001 Chr.12: 47977825-47977825 on Build GRCh38
rs905690054 Chr.12: 47977911-47977911 on Build GRCh38
rs1057518157 Chr.12: 47977608-47977609 on Build GRCh38
rs1044662808 Chr.12: 47977902-47977902 on Build GRCh38
rs978657987 Chr.12: 47977646-47977646 on Build GRCh38
rs61554705 Chr.12: 47977842-47977842 on Build GRCh38
rs1032827467 Chr.12: 47977649-47977649 on Build GRCh38
rs930128565 Chr.12: 47977540-47977540 on Build GRCh38

View Full Product Details