Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3762490-3762985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
UniGene
Hs.348700
Gene Chromosome Location
Chr.1: 3752398-3771645 on Build GRCh38
Transcripts
NM_152492.2 XM_011540780.1 XM_017000402.1 XM_017000403.1 XM_017000404.1 XM_017000405.1 XM_017000406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886382 c.942G>T p.Q314H Chr.1: 3762700-3762700 on Build GRCh38
6420489 c.884T>C p.L295P Chr.1: 3762642-3762642 on Build GRCh38
5528243 c.870C>T p.L290L Chr.1: 3762628-3762628 on Build GRCh38
5949556 c.908T>A p.L303Q Chr.1: 3762666-3762666 on Build GRCh38
1602308 c.914C>T p.A305V Chr.1: 3762672-3762672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889788508 Chr.1: 3762611-3762611 on Build GRCh38
rs139134726 Chr.1: 3762688-3762688 on Build GRCh38
rs746214337 Chr.1: 3762652-3762652 on Build GRCh38
rs1010097912 Chr.1: 3762642-3762642 on Build GRCh38
rs931421607 Chr.1: 3762773-3762773 on Build GRCh38
rs752721684 Chr.1: 3762938-3762938 on Build GRCh38
rs75571263 Chr.1: 3762639-3762639 on Build GRCh38
rs1041429278 Chr.1: 3762866-3762866 on Build GRCh38
rs375200194 Chr.1: 3762634-3762634 on Build GRCh38
rs775826360 Chr.1: 3762696-3762696 on Build GRCh38
rs968489071 Chr.1: 3762898-3762898 on Build GRCh38
rs961693496 Chr.1: 3762964-3762964 on Build GRCh38
rs150485951 Chr.1: 3762635-3762635 on Build GRCh38
rs376514114 Chr.1: 3762948-3762948 on Build GRCh38
rs559509348 Chr.1: 3762890-3762890 on Build GRCh38
rs918082761 Chr.1: 3762571-3762571 on Build GRCh38
rs889801090 Chr.1: 3762831-3762831 on Build GRCh38
rs1010105394 Chr.1: 3762891-3762891 on Build GRCh38
rs918147923 Chr.1: 3762739-3762739 on Build GRCh38
rs968840829 Chr.1: 3762925-3762925 on Build GRCh38
rs995574982 Chr.1: 3762584-3762584 on Build GRCh38
rs4648556 Chr.1: 3762582-3762582 on Build GRCh38
rs991637678 Chr.1: 3762712-3762712 on Build GRCh38
rs750301491 Chr.1: 3762748-3762748 on Build GRCh38
rs1049808762 Chr.1: 3762789-3762789 on Build GRCh38
rs369668046 Chr.1: 3762709-3762709 on Build GRCh38
rs780127978 Chr.1: 3762579-3762579 on Build GRCh38
rs192243643 Chr.1: 3762671-3762671 on Build GRCh38
rs138952248 Chr.1: 3762640-3762640 on Build GRCh38
rs192646560 Chr.1: 3762746-3762746 on Build GRCh38
rs563488648 Chr.1: 3762920-3762920 on Build GRCh38
rs6699377 Chr.1: 3762913-3762913 on Build GRCh38
rs373335569 Chr.1: 3762752-3762752 on Build GRCh38
rs376711772 Chr.1: 3762583-3762583 on Build GRCh38
rs4648558 Chr.1: 3762897-3762897 on Build GRCh38
rs1181879 Chr.1: 3762575-3762575 on Build GRCh38
rs1006096220 Chr.1: 3762875-3762875 on Build GRCh38
rs188919320 Chr.1: 3762738-3762738 on Build GRCh38
rs947636741 Chr.1: 3762722-3762722 on Build GRCh38
rs1008197705 Chr.1: 3762861-3762861 on Build GRCh38
rs561016317 Chr.1: 3762956-3762956 on Build GRCh38
rs552792476 Chr.1: 3762816-3762816 on Build GRCh38
rs888923349 Chr.1: 3762871-3762871 on Build GRCh38
rs766731177 Chr.1: 3762858-3762858 on Build GRCh38
rs1049798888 Chr.1: 3762610-3762610 on Build GRCh38
rs901237674 Chr.1: 3762827-3762827 on Build GRCh38
rs376119454 Chr.1: 3762828-3762828 on Build GRCh38
rs1016559626 Chr.1: 3762887-3762887 on Build GRCh38
rs774192225 Chr.1: 3762824-3762824 on Build GRCh38
rs960274088 Chr.1: 3762710-3762710 on Build GRCh38
rs545038171 Chr.1: 3762533-3762533 on Build GRCh38
rs979761227 Chr.1: 3762523-3762523 on Build GRCh38
rs571586425 Chr.1: 3762699-3762699 on Build GRCh38
rs4648557 Chr.1: 3762743-3762743 on Build GRCh38
rs926959899 Chr.1: 3762528-3762528 on Build GRCh38
rs530863043 Chr.1: 3762576-3762576 on Build GRCh38
rs959694711 Chr.1: 3762723-3762723 on Build GRCh38
rs183267905 Chr.1: 3762717-3762717 on Build GRCh38
rs993565900 Chr.1: 3762894-3762894 on Build GRCh38
rs947392505 Chr.1: 3762530-3762530 on Build GRCh38
rs1021395552 Chr.1: 3762651-3762651 on Build GRCh38
rs938296341 Chr.1: 3762532-3762532 on Build GRCh38
rs6699385 Chr.1: 3762950-3762950 on Build GRCh38
rs779589191 Chr.1: 3762716-3762716 on Build GRCh38
rs959988834 Chr.1: 3762914-3762914 on Build GRCh38
rs760696931 Chr.1: 3762943-3762943 on Build GRCh38
rs1001242119 Chr.1: 3762900-3762900 on Build GRCh38
rs767384553 Chr.1: 3762569-3762569 on Build GRCh38
rs891732956 Chr.1: 3762888-3762888 on Build GRCh38

View Full Product Details