Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6248931-6249422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR153
Entrez Gene ID
387509
Gene Name
G protein-coupled receptor 153
Gene Aliases
PGR1
UniGene
Hs.531581
Gene Chromosome Location
Chr.1: 6247346-6260975 on Build GRCh38
Transcripts
NM_207370.2 XM_011541434.2 XM_017001250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4688034 c.1777G>A p.E593K Chr.1: 6249391-6249391 on Build GRCh38
5511451 c.1768T>C p.S590P Chr.1: 6249400-6249400 on Build GRCh38
6314278 c.1813C>A p.L605M Chr.1: 6249355-6249355 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs532166529 Chr.1: 6249165-6249165 on Build GRCh38
rs548260316 Chr.1: 6249261-6249261 on Build GRCh38
rs552692977 Chr.1: 6249304-6249304 on Build GRCh38
rs906689892 Chr.1: 6249344-6249344 on Build GRCh38
rs959768811 Chr.1: 6249175-6249175 on Build GRCh38
rs1003825293 Chr.1: 6249007-6249007 on Build GRCh38
rs560667124 Chr.1: 6249050-6249050 on Build GRCh38
rs979676960 Chr.1: 6249208-6249208 on Build GRCh38
rs1033963149 Chr.1: 6249236-6249236 on Build GRCh38
rs541363987 Chr.1: 6249226-6249226 on Build GRCh38
rs564773977 Chr.1: 6249157-6249157 on Build GRCh38
rs865806928 Chr.1: 6249375-6249375 on Build GRCh38
rs878992300 Chr.1: 6249161-6249161 on Build GRCh38
rs535366767 Chr.1: 6248976-6248976 on Build GRCh38
rs968540883 Chr.1: 6249108-6249108 on Build GRCh38
rs759056403 Chr.1: 6249366-6249366 on Build GRCh38
rs948316949 Chr.1: 6248989-6248989 on Build GRCh38
rs923471690 Chr.1: 6249233-6249233 on Build GRCh38
rs924338803 Chr.1: 6249392-6249392 on Build GRCh38
rs1050374249 Chr.1: 6249096-6249096 on Build GRCh38
rs375076370 Chr.1: 6249064-6249064 on Build GRCh38
rs746518498 Chr.1: 6249342-6249342 on Build GRCh38
rs762920453 Chr.1: 6249396-6249396 on Build GRCh38
rs575705908 Chr.1: 6249079-6249079 on Build GRCh38
rs866705177 Chr.1: 6249357-6249357 on Build GRCh38
rs1040967050 Chr.1: 6249152-6249152 on Build GRCh38
rs376754227 Chr.1: 6249115-6249115 on Build GRCh38
rs962205149 Chr.1: 6248953-6248953 on Build GRCh38
rs548324750 Chr.1: 6248963-6248963 on Build GRCh38
rs543093185 Chr.1: 6249129-6249129 on Build GRCh38
rs770558616 Chr.1: 6249354-6249354 on Build GRCh38
rs939732814 Chr.1: 6249305-6249305 on Build GRCh38
rs115543665 Chr.1: 6249039-6249039 on Build GRCh38
rs776354779 Chr.1: 6249356-6249356 on Build GRCh38
rs779957206 Chr.1: 6248958-6248958 on Build GRCh38
rs772665154 Chr.1: 6249297-6249297 on Build GRCh38
rs1022354032 Chr.1: 6249244-6249244 on Build GRCh38
rs889142288 Chr.1: 6249122-6249122 on Build GRCh38
rs775225929 Chr.1: 6249395-6249395 on Build GRCh38
rs371142322 Chr.1: 6249241-6249241 on Build GRCh38
rs754449548 Chr.1: 6249070-6249070 on Build GRCh38
rs113086922 Chr.1: 6249035-6249035 on Build GRCh38
rs866300956 Chr.1: 6249119-6249119 on Build GRCh38
rs868692439 Chr.1: 6249361-6249361 on Build GRCh38
rs769654419 Chr.1: 6249372-6249372 on Build GRCh38
rs935508398 Chr.1: 6249045-6249045 on Build GRCh38
rs925532714 Chr.1: 6249216-6249216 on Build GRCh38
rs970980141 Chr.1: 6249000-6249000 on Build GRCh38
rs968376956 Chr.1: 6249343-6249343 on Build GRCh38
rs915561936 Chr.1: 6249192-6249192 on Build GRCh38
rs943947163 Chr.1: 6249150-6249150 on Build GRCh38
rs113362011 Chr.1: 6248969-6248969 on Build GRCh38
rs571099896 Chr.1: 6249316-6249316 on Build GRCh38
rs570043772 Chr.1: 6249289-6249289 on Build GRCh38
rs968805472 Chr.1: 6249196-6249196 on Build GRCh38
rs1044392577 Chr.1: 6249010-6249010 on Build GRCh38
rs957167009 Chr.1: 6249393-6249393 on Build GRCh38
rs553385679 Chr.1: 6249016-6249016 on Build GRCh38
rs1883608 Chr.1: 6249259-6249259 on Build GRCh38
rs935471552 Chr.1: 6249217-6249217 on Build GRCh38
rs1075751 Chr.1: 6249257-6249257 on Build GRCh38
rs991149760 Chr.1: 6248972-6248972 on Build GRCh38
rs1048110450 Chr.1: 6249166-6249166 on Build GRCh38
rs924504442 Chr.1: 6249146-6249146 on Build GRCh38
rs991265847 Chr.1: 6249187-6249187 on Build GRCh38
rs1016608714 Chr.1: 6249131-6249131 on Build GRCh38
rs978835931 Chr.1: 6249380-6249380 on Build GRCh38
rs535074819 Chr.1: 6249166-6249166 on Build GRCh38
rs889094099 Chr.1: 6249256-6249256 on Build GRCh38
rs897735272 Chr.1: 6249281-6249281 on Build GRCh38
rs933391254 Chr.1: 6249235-6249235 on Build GRCh38
rs993447103 Chr.1: 6249288-6249288 on Build GRCh38
rs531058786 Chr.1: 6249291-6249291 on Build GRCh38
rs866919918 Chr.1: 6249402-6249402 on Build GRCh38
rs572013806 Chr.1: 6249018-6249018 on Build GRCh38
rs996590079 Chr.1: 6248965-6248965 on Build GRCh38
rs915317810 Chr.1: 6249090-6249090 on Build GRCh38
rs961871374 Chr.1: 6249144-6249144 on Build GRCh38
rs1048658856 Chr.1: 6248966-6248966 on Build GRCh38

View Full Product Details