Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49636624-49637124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7449196 c.1427A>G p.E476G Chr.12: 49636782-49636782 on Build GRCh38
6874752 c.1464C>T p.R488R Chr.12: 49636819-49636819 on Build GRCh38
3398785 c.1471C>T p.R491C Chr.12: 49636826-49636826 on Build GRCh38
5479517 c.1481G>A p.R494Q Chr.12: 49636836-49636836 on Build GRCh38
5900252 c.1420G>A p.E474K Chr.12: 49636775-49636775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761025843 Chr.12: 49636782-49636782 on Build GRCh38
rs200339348 Chr.12: 49636765-49636765 on Build GRCh38
rs1032010843 Chr.12: 49636984-49636984 on Build GRCh38
rs115064935 Chr.12: 49636924-49636924 on Build GRCh38
rs771169576 Chr.12: 49636873-49636873 on Build GRCh38
rs548343320 Chr.12: 49637010-49637010 on Build GRCh38
rs974916085 Chr.12: 49637093-49637094 on Build GRCh38
rs560424980 Chr.12: 49636923-49636923 on Build GRCh38
rs1007789153 Chr.12: 49636975-49636975 on Build GRCh38
rs550550875 Chr.12: 49637059-49637059 on Build GRCh38
rs781235317 Chr.12: 49636817-49636817 on Build GRCh38
rs555779705 Chr.12: 49637054-49637054 on Build GRCh38
rs757263724 Chr.12: 49636758-49636758 on Build GRCh38
rs373033025 Chr.12: 49636854-49636854 on Build GRCh38
rs749260225 Chr.12: 49636692-49636692 on Build GRCh38
rs201178215 Chr.12: 49636775-49636775 on Build GRCh38
rs570783662 Chr.12: 49637072-49637072 on Build GRCh38
rs202143759 Chr.12: 49636803-49636803 on Build GRCh38
rs748346733 Chr.12: 49636776-49636776 on Build GRCh38
rs565351071 Chr.12: 49636693-49636693 on Build GRCh38
rs200375855 Chr.12: 49636761-49636761 on Build GRCh38
rs754436465 Chr.12: 49636719-49636719 on Build GRCh38
rs188148516 Chr.12: 49636946-49636946 on Build GRCh38
rs374081725 Chr.12: 49636790-49636790 on Build GRCh38
rs368203210 Chr.12: 49637061-49637061 on Build GRCh38
rs992806129 Chr.12: 49636815-49636815 on Build GRCh38
rs185094759 Chr.12: 49637034-49637034 on Build GRCh38
rs1017789212 Chr.12: 49636997-49636997 on Build GRCh38
rs907044841 Chr.12: 49636952-49636952 on Build GRCh38
rs770279133 Chr.12: 49636897-49636897 on Build GRCh38
rs753458912 Chr.12: 49636718-49636718 on Build GRCh38
rs145198596 Chr.12: 49636886-49636886 on Build GRCh38
rs756035810 Chr.12: 49636826-49636826 on Build GRCh38
rs561353855 Chr.12: 49636835-49636835 on Build GRCh38
rs767834251 Chr.12: 49636703-49636703 on Build GRCh38
rs369226181 Chr.12: 49636894-49636894 on Build GRCh38
rs768592072 Chr.12: 49636767-49636767 on Build GRCh38
rs535964869 Chr.12: 49636888-49636888 on Build GRCh38
rs772230038 Chr.12: 49636778-49636778 on Build GRCh38
rs899343571 Chr.12: 49637027-49637027 on Build GRCh38
rs746740779 Chr.12: 49636742-49636742 on Build GRCh38
rs372672216 Chr.12: 49636687-49636687 on Build GRCh38
rs769706835 Chr.12: 49636883-49636883 on Build GRCh38
rs890112431 Chr.12: 49636954-49636954 on Build GRCh38
rs748093490 Chr.12: 49636676-49636686 on Build GRCh38
rs752204001 Chr.12: 49636797-49636797 on Build GRCh38
rs546835771 Chr.12: 49636721-49636721 on Build GRCh38
rs565902316 Chr.12: 49637066-49637067 on Build GRCh38
rs747427823 Chr.12: 49636852-49636852 on Build GRCh38
rs981786077 Chr.12: 49636785-49636785 on Build GRCh38
rs943156553 Chr.12: 49636813-49636813 on Build GRCh38
rs148704115 Chr.12: 49636939-49636939 on Build GRCh38
rs895137519 Chr.12: 49636935-49636935 on Build GRCh38
rs148146841 Chr.12: 49636818-49636818 on Build GRCh38
rs778879798 Chr.12: 49636827-49636827 on Build GRCh38
rs1025037127 Chr.12: 49636943-49636943 on Build GRCh38
rs766932461 Chr.12: 49636789-49636789 on Build GRCh38
rs749402466 Chr.12: 49636747-49636747 on Build GRCh38
rs774865121 Chr.12: 49636898-49636898 on Build GRCh38
rs773522286 Chr.12: 49636704-49636704 on Build GRCh38
rs372164491 Chr.12: 49636705-49636705 on Build GRCh38
rs765190928 Chr.12: 49637039-49637039 on Build GRCh38
rs762282816 Chr.12: 49636695-49636695 on Build GRCh38
rs758085600 Chr.12: 49636735-49636735 on Build GRCh38
rs752260668 Chr.12: 49636722-49636722 on Build GRCh38
rs773573283 Chr.12: 49636780-49636780 on Build GRCh38
rs759127357 Chr.12: 49636791-49636791 on Build GRCh38
rs145385705 Chr.12: 49636694-49636694 on Build GRCh38
rs368904714 Chr.12: 49636808-49636808 on Build GRCh38
rs545428941 Chr.12: 49636691-49636691 on Build GRCh38
rs192922894 Chr.12: 49637031-49637031 on Build GRCh38
rs769745722 Chr.12: 49636658-49636658 on Build GRCh38
rs771971581 Chr.12: 49636686-49636686 on Build GRCh38
rs114081796 Chr.12: 49636846-49636846 on Build GRCh38
rs769423273 Chr.12: 49636711-49636711 on Build GRCh38
rs766807655 Chr.12: 49636710-49636710 on Build GRCh38
rs955996605 Chr.12: 49637002-49637002 on Build GRCh38
rs753684139 Chr.12: 49636802-49636802 on Build GRCh38
rs775846220 Chr.12: 49636666-49636666 on Build GRCh38
rs936492590 Chr.12: 49636901-49636901 on Build GRCh38
rs772834181 Chr.12: 49637082-49637082 on Build GRCh38
rs202075390 Chr.12: 49636809-49636809 on Build GRCh38
rs746975774 Chr.12: 49636690-49636690 on Build GRCh38
rs764686125 Chr.12: 49636796-49636796 on Build GRCh38
rs759984772 Chr.12: 49636890-49636890 on Build GRCh38
rs182897573 Chr.12: 49636936-49636936 on Build GRCh38
rs777735176 Chr.12: 49636741-49636741 on Build GRCh38

View Full Product Details