Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57611144-57611648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF25
Entrez Gene ID
115557
Gene Name
Rho guanine nucleotide exchange factor 25
Gene Aliases
GEFT, p63RhoGEF
UniGene
Hs.61581
Gene Chromosome Location
Chr.12: 57610180-57617245 on Build GRCh38
Transcripts
NM_001111270.2 NM_182947.3 NR_046223.1

SNP Detail

SNP ID Chromosome Location
rs937345283 Chr.12: 57611223-57611223 on Build GRCh38
rs1041349071 Chr.12: 57611483-57611483 on Build GRCh38
rs933171932 Chr.12: 57611451-57611451 on Build GRCh38
rs1006411024 Chr.12: 57611510-57611510 on Build GRCh38
rs889036074 Chr.12: 57611466-57611466 on Build GRCh38
rs913300066 Chr.12: 57611368-57611368 on Build GRCh38
rs570956692 Chr.12: 57611626-57611626 on Build GRCh38
rs981667674 Chr.12: 57611331-57611331 on Build GRCh38
rs34181394 Chr.12: 57611490-57611490 on Build GRCh38
rs57989706 Chr.12: 57611245-57611245 on Build GRCh38
rs1050273739 Chr.12: 57611461-57611461 on Build GRCh38
rs1003050932 Chr.12: 57611608-57611608 on Build GRCh38
rs940798917 Chr.12: 57611492-57611492 on Build GRCh38
rs942594278 Chr.12: 57611481-57611481 on Build GRCh38
rs1020482520 Chr.12: 57611329-57611329 on Build GRCh38
rs1001808862 Chr.12: 57611575-57611575 on Build GRCh38
rs947207580 Chr.12: 57611241-57611241 on Build GRCh38
rs562357564 Chr.12: 57611294-57611294 on Build GRCh38
rs1025398294 Chr.12: 57611493-57611493 on Build GRCh38
rs1051781265 Chr.12: 57611485-57611485 on Build GRCh38
rs1010815466 Chr.12: 57611319-57611319 on Build GRCh38
rs147163070 Chr.12: 57611209-57611209 on Build GRCh38
rs901759948 Chr.12: 57611252-57611252 on Build GRCh38
rs1019218571 Chr.12: 57611256-57611256 on Build GRCh38
rs993962059 Chr.12: 57611265-57611265 on Build GRCh38
rs559870039 Chr.12: 57611253-57611254 on Build GRCh38
rs537809438 Chr.12: 57611218-57611218 on Build GRCh38
rs532931573 Chr.12: 57611450-57611450 on Build GRCh38
rs1006514981 Chr.12: 57611249-57611249 on Build GRCh38
rs148974792 Chr.12: 57611577-57611577 on Build GRCh38
rs988951491 Chr.12: 57611366-57611366 on Build GRCh38
rs889666017 Chr.12: 57611248-57611248 on Build GRCh38
rs928729536 Chr.12: 57611171-57611171 on Build GRCh38
rs970274718 Chr.12: 57611295-57611295 on Build GRCh38
rs888152107 Chr.12: 57611504-57611504 on Build GRCh38
rs980111021 Chr.12: 57611338-57611338 on Build GRCh38
rs889721370 Chr.12: 57611472-57611472 on Build GRCh38
rs1053599184 Chr.12: 57611198-57611198 on Build GRCh38
rs1036378175 Chr.12: 57611505-57611505 on Build GRCh38
rs947231202 Chr.12: 57611403-57611403 on Build GRCh38
rs985252137 Chr.12: 57611235-57611235 on Build GRCh38
rs928680552 Chr.12: 57611345-57611345 on Build GRCh38
rs1016411800 Chr.12: 57611538-57611538 on Build GRCh38
rs972292916 Chr.12: 57611424-57611424 on Build GRCh38
rs540264849 Chr.12: 57611179-57611179 on Build GRCh38
rs970199758 Chr.12: 57611560-57611560 on Build GRCh38
rs938750438 Chr.12: 57611173-57611173 on Build GRCh38
rs938662698 Chr.12: 57611360-57611360 on Build GRCh38
rs933266682 Chr.12: 57611253-57611253 on Build GRCh38
rs933890745 Chr.12: 57611480-57611480 on Build GRCh38
rs892199294 Chr.12: 57611525-57611525 on Build GRCh38
rs900933760 Chr.12: 57611257-57611257 on Build GRCh38
rs1025163856 Chr.12: 57611552-57611552 on Build GRCh38
rs900861434 Chr.12: 57611486-57611486 on Build GRCh38
rs984921377 Chr.12: 57611291-57611291 on Build GRCh38
rs950945581 Chr.12: 57611286-57611286 on Build GRCh38
rs1032469754 Chr.12: 57611289-57611289 on Build GRCh38
rs940819511 Chr.12: 57611421-57611421 on Build GRCh38
rs866812717 Chr.12: 57611312-57611312 on Build GRCh38
rs529151926 Chr.12: 57611242-57611242 on Build GRCh38
rs984809966 Chr.12: 57611400-57611400 on Build GRCh38
rs1025812352 Chr.12: 57611278-57611278 on Build GRCh38

View Full Product Details