Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57613924-57614414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF25
Entrez Gene ID
115557
Gene Name
Rho guanine nucleotide exchange factor 25
Gene Aliases
GEFT, p63RhoGEF
UniGene
Hs.61581
Gene Chromosome Location
Chr.12: 57610180-57617245 on Build GRCh38
Transcripts
NM_001111270.2 NM_182947.3 NR_046223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7304595 c.774-8C>A p.? Chr.12: 57614323-57614323 on Build GRCh38
1178591 c.722G>A p.R241H Chr.12: 57614068-57614068 on Build GRCh38
3359871 c.686T>A p.L229Q Chr.12: 57614360-57614360 on Build GRCh38
431597 c.707C>T p.A236V Chr.12: 57614381-57614381 on Build GRCh38
549364 c.598C>T p.R200* Chr.12: 57614061-57614061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762708060 Chr.12: 57614145-57614145 on Build GRCh38
rs150532634 Chr.12: 57614351-57614351 on Build GRCh38
rs531817541 Chr.12: 57614293-57614293 on Build GRCh38
rs987342721 Chr.12: 57614253-57614253 on Build GRCh38
rs78827077 Chr.12: 57614154-57614154 on Build GRCh38
rs756197273 Chr.12: 57614062-57614062 on Build GRCh38
rs766160305 Chr.12: 57614034-57614034 on Build GRCh38
rs111730781 Chr.12: 57614274-57614274 on Build GRCh38
rs141643619 Chr.12: 57614118-57614118 on Build GRCh38
rs752715781 Chr.12: 57614386-57614386 on Build GRCh38
rs780550596 Chr.12: 57614310-57614310 on Build GRCh38
rs749619877 Chr.12: 57613974-57613974 on Build GRCh38
rs770199613 Chr.12: 57613967-57613967 on Build GRCh38
rs765926337 Chr.12: 57614153-57614153 on Build GRCh38
rs773813272 Chr.12: 57614355-57614355 on Build GRCh38
rs748306195 Chr.12: 57614057-57614057 on Build GRCh38
rs758610687 Chr.12: 57614155-57614156 on Build GRCh38
rs201534150 Chr.12: 57614294-57614294 on Build GRCh38
rs902398268 Chr.12: 57614052-57614052 on Build GRCh38
rs758139060 Chr.12: 57614033-57614033 on Build GRCh38
rs369082821 Chr.12: 57613976-57613976 on Build GRCh38
rs1030492082 Chr.12: 57614201-57614201 on Build GRCh38
rs376746783 Chr.12: 57614114-57614114 on Build GRCh38
rs369771487 Chr.12: 57614142-57614142 on Build GRCh38
rs761295474 Chr.12: 57613994-57613994 on Build GRCh38
rs900165265 Chr.12: 57614158-57614158 on Build GRCh38
rs746298203 Chr.12: 57614304-57614304 on Build GRCh38
rs919745548 Chr.12: 57613960-57613960 on Build GRCh38
rs974296248 Chr.12: 57613950-57613950 on Build GRCh38
rs750212387 Chr.12: 57614060-57614068 on Build GRCh38
rs777873296 Chr.12: 57614074-57614074 on Build GRCh38
rs771272053 Chr.12: 57613975-57613975 on Build GRCh38
rs951176649 Chr.12: 57614376-57614376 on Build GRCh38
rs780460921 Chr.12: 57614322-57614323 on Build GRCh38
rs759421652 Chr.12: 57614368-57614368 on Build GRCh38
rs750021947 Chr.12: 57614005-57614005 on Build GRCh38
rs149566238 Chr.12: 57614167-57614167 on Build GRCh38
rs200046593 Chr.12: 57613981-57613981 on Build GRCh38
rs4760307 Chr.12: 57614231-57614231 on Build GRCh38
rs571298065 Chr.12: 57614325-57614325 on Build GRCh38
rs377474244 Chr.12: 57614353-57614353 on Build GRCh38
rs774815529 Chr.12: 57614104-57614104 on Build GRCh38
rs772354760 Chr.12: 57614305-57614305 on Build GRCh38
rs896356663 Chr.12: 57614003-57614003 on Build GRCh38
rs746206067 Chr.12: 57614108-57614108 on Build GRCh38
rs974306796 Chr.12: 57614380-57614380 on Build GRCh38
rs760047079 Chr.12: 57613978-57613978 on Build GRCh38
rs542870649 Chr.12: 57614263-57614263 on Build GRCh38
rs538464790 Chr.12: 57614315-57614315 on Build GRCh38
rs764668464 Chr.12: 57614141-57614141 on Build GRCh38
rs751663570 Chr.12: 57614352-57614353 on Build GRCh38
rs916646345 Chr.12: 57613945-57613945 on Build GRCh38
rs532231407 Chr.12: 57614333-57614333 on Build GRCh38
rs202200912 Chr.12: 57614164-57614164 on Build GRCh38
rs767570617 Chr.12: 57614159-57614159 on Build GRCh38
rs776166768 Chr.12: 57613983-57613983 on Build GRCh38
rs771228784 Chr.12: 57614096-57614096 on Build GRCh38
rs373514799 Chr.12: 57614050-57614050 on Build GRCh38
rs969530436 Chr.12: 57613948-57613949 on Build GRCh38
rs764932430 Chr.12: 57613998-57613998 on Build GRCh38
rs752627724 Chr.12: 57614160-57614160 on Build GRCh38
rs755018413 Chr.12: 57614043-57614043 on Build GRCh38
rs754821004 Chr.12: 57614156-57614156 on Build GRCh38
rs781266800 Chr.12: 57614048-57614048 on Build GRCh38
rs762759558 Chr.12: 57614243-57614243 on Build GRCh38
rs974155001 Chr.12: 57614352-57614352 on Build GRCh38
rs1015857968 Chr.12: 57614257-57614257 on Build GRCh38
rs761316002 Chr.12: 57614127-57614127 on Build GRCh38
rs369247314 Chr.12: 57614038-57614038 on Build GRCh38
rs772829192 Chr.12: 57614346-57614346 on Build GRCh38
rs547082119 Chr.12: 57614354-57614354 on Build GRCh38
rs749479774 Chr.12: 57614081-57614081 on Build GRCh38
rs773565464 Chr.12: 57613973-57613973 on Build GRCh38
rs193921117 Chr.12: 57614324-57614324 on Build GRCh38
rs767374507 Chr.12: 57614381-57614381 on Build GRCh38

View Full Product Details