Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57763505-57764012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27B1
Entrez Gene ID
1594
Gene Name
cytochrome P450 family 27 subfamily B member 1
Gene Aliases
CP2B, CYP1, CYP1alpha, CYP27B, P450c1, PDDR, VDD1, VDDR, VDDRI, VDR
UniGene
Hs.524528
Gene Chromosome Location
Chr.12: 57762334-57767193 on Build GRCh38
Transcripts
NM_000785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7315714 c.1375C>T p.R459C Chr.12: 57763649-57763649 on Build GRCh38
1363434 c.1295G>A p.R432H Chr.12: 57763729-57763729 on Build GRCh38
1284389 c.1313C>A p.P438H Chr.12: 57763711-57763711 on Build GRCh38
6220822 c.1413+1G>A p.? Chr.12: 57763610-57763610 on Build GRCh38
1989859 c.1322A>C p.H441P Chr.12: 57763702-57763702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949265477 Chr.12: 57763952-57763952 on Build GRCh38
rs1039653245 Chr.12: 57763590-57763590 on Build GRCh38
rs754559171 Chr.12: 57763650-57763651 on Build GRCh38
rs926566523 Chr.12: 57763966-57763966 on Build GRCh38
rs557461860 Chr.12: 57763564-57763564 on Build GRCh38
rs150025067 Chr.12: 57763714-57763714 on Build GRCh38
rs866796725 Chr.12: 57763711-57763711 on Build GRCh38
rs1029699819 Chr.12: 57763730-57763730 on Build GRCh38
rs753678470 Chr.12: 57763704-57763704 on Build GRCh38
rs762550487 Chr.12: 57763648-57763648 on Build GRCh38
rs756848076 Chr.12: 57763946-57763946 on Build GRCh38
rs780440003 Chr.12: 57763719-57763719 on Build GRCh38
rs199923824 Chr.12: 57763644-57763644 on Build GRCh38
rs780280592 Chr.12: 57763810-57763810 on Build GRCh38
rs958837367 Chr.12: 57763892-57763892 on Build GRCh38
rs377154743 Chr.12: 57763802-57763802 on Build GRCh38
rs1048050999 Chr.12: 57763989-57763989 on Build GRCh38
rs118204008 Chr.12: 57763798-57763798 on Build GRCh38
rs781353865 Chr.12: 57763840-57763840 on Build GRCh38
rs772650262 Chr.12: 57763647-57763647 on Build GRCh38
rs925235815 Chr.12: 57763869-57763869 on Build GRCh38
rs752504237 Chr.12: 57763671-57763671 on Build GRCh38
rs374454037 Chr.12: 57763776-57763776 on Build GRCh38
rs762701380 Chr.12: 57763744-57763744 on Build GRCh38
rs767480544 Chr.12: 57763667-57763667 on Build GRCh38
rs760548148 Chr.12: 57763773-57763773 on Build GRCh38
rs1003845117 Chr.12: 57763733-57763733 on Build GRCh38
rs751262543 Chr.12: 57763662-57763662 on Build GRCh38
rs372020956 Chr.12: 57763589-57763589 on Build GRCh38
rs768857789 Chr.12: 57763729-57763729 on Build GRCh38
rs769534483 Chr.12: 57763770-57763770 on Build GRCh38
rs935422610 Chr.12: 57763923-57763923 on Build GRCh38
rs556493554 Chr.12: 57763732-57763732 on Build GRCh38
rs778903870 Chr.12: 57763707-57763707 on Build GRCh38
rs756054548 Chr.12: 57763692-57763692 on Build GRCh38
rs183213602 Chr.12: 57763565-57763565 on Build GRCh38
rs746144650 Chr.12: 57763713-57763713 on Build GRCh38
rs775815261 Chr.12: 57763609-57763609 on Build GRCh38
rs925934052 Chr.12: 57763932-57763932 on Build GRCh38
rs544219775 Chr.12: 57763721-57763721 on Build GRCh38
rs188819808 Chr.12: 57763826-57763826 on Build GRCh38
rs917303574 Chr.12: 57763914-57763914 on Build GRCh38
rs757358360 Chr.12: 57763705-57763705 on Build GRCh38
rs749221712 Chr.12: 57763866-57763866 on Build GRCh38
rs761149528 Chr.12: 57763621-57763621 on Build GRCh38
rs931210968 Chr.12: 57763968-57763968 on Build GRCh38
rs190694526 Chr.12: 57763544-57763544 on Build GRCh38
rs748748564 Chr.12: 57763848-57763848 on Build GRCh38
rs1039572705 Chr.12: 57763961-57763961 on Build GRCh38
rs750468088 Chr.12: 57763806-57763806 on Build GRCh38
rs780950819 Chr.12: 57763698-57763699 on Build GRCh38
rs550565740 Chr.12: 57763880-57763880 on Build GRCh38
rs112144903 Chr.12: 57763929-57763929 on Build GRCh38
rs753823609 Chr.12: 57763780-57763780 on Build GRCh38
rs773986669 Chr.12: 57763756-57763756 on Build GRCh38
rs549873942 Chr.12: 57763619-57763619 on Build GRCh38
rs555068245 Chr.12: 57763649-57763649 on Build GRCh38
rs962437616 Chr.12: 57763751-57763751 on Build GRCh38
rs568165874 Chr.12: 57763738-57763738 on Build GRCh38
rs573538895 Chr.12: 57763703-57763703 on Build GRCh38
rs759208930 Chr.12: 57763666-57763666 on Build GRCh38
rs946039130 Chr.12: 57763547-57763547 on Build GRCh38
rs886049725 Chr.12: 57763639-57763639 on Build GRCh38
rs759331498 Chr.12: 57763758-57763758 on Build GRCh38
rs78250220 Chr.12: 57763788-57763788 on Build GRCh38
rs904563253 Chr.12: 57763575-57763575 on Build GRCh38
rs531984918 Chr.12: 57763835-57763835 on Build GRCh38
rs765035904 Chr.12: 57763660-57763660 on Build GRCh38
rs746019534 Chr.12: 57763578-57763578 on Build GRCh38
rs758671027 Chr.12: 57763807-57763807 on Build GRCh38
rs767253913 Chr.12: 57763769-57763769 on Build GRCh38
rs993009102 Chr.12: 57763901-57763901 on Build GRCh38
rs1029340989 Chr.12: 57763698-57763698 on Build GRCh38
rs946125321 Chr.12: 57763919-57763919 on Build GRCh38
rs111235166 Chr.12: 57763849-57763849 on Build GRCh38

View Full Product Details