Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89588274-89588777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

SNP Detail

SNP ID Chromosome Location
rs901111937 Chr.12: 89588348-89588348 on Build GRCh38
rs951147205 Chr.12: 89588524-89588524 on Build GRCh38
rs982431833 Chr.12: 89588364-89588364 on Build GRCh38
rs560640090 Chr.12: 89588357-89588357 on Build GRCh38
rs1051875385 Chr.12: 89588402-89588402 on Build GRCh38
rs368205120 Chr.12: 89588480-89588480 on Build GRCh38
rs1034595129 Chr.12: 89588489-89588489 on Build GRCh38
rs996611340 Chr.12: 89588389-89588389 on Build GRCh38
rs926016583 Chr.12: 89588732-89588732 on Build GRCh38
rs890445399 Chr.12: 89588414-89588414 on Build GRCh38
rs531877745 Chr.12: 89588560-89588560 on Build GRCh38
rs550755291 Chr.12: 89588311-89588311 on Build GRCh38
rs904385521 Chr.12: 89588422-89588422 on Build GRCh38
rs540947852 Chr.12: 89588299-89588299 on Build GRCh38
rs904406582 Chr.12: 89588462-89588462 on Build GRCh38
rs942303820 Chr.12: 89588378-89588378 on Build GRCh38
rs370780260 Chr.12: 89588666-89588666 on Build GRCh38
rs1013946959 Chr.12: 89588515-89588515 on Build GRCh38
rs894343418 Chr.12: 89588496-89588496 on Build GRCh38
rs182188093 Chr.12: 89588531-89588531 on Build GRCh38
rs529527096 Chr.12: 89588398-89588398 on Build GRCh38
rs970950347 Chr.12: 89588588-89588588 on Build GRCh38
rs1032880666 Chr.12: 89588517-89588517 on Build GRCh38
rs939693274 Chr.12: 89588339-89588339 on Build GRCh38
rs946123083 Chr.12: 89588673-89588673 on Build GRCh38
rs953030620 Chr.12: 89588671-89588671 on Build GRCh38
rs79224935 Chr.12: 89588634-89588634 on Build GRCh38
rs973938372 Chr.12: 89588392-89588392 on Build GRCh38
rs767033371 Chr.12: 89588396-89588398 on Build GRCh38
rs571648330 Chr.12: 89588674-89588674 on Build GRCh38
rs776976693 Chr.12: 89588584-89588584 on Build GRCh38
rs749181051 Chr.12: 89588578-89588578 on Build GRCh38
rs908090388 Chr.12: 89588365-89588365 on Build GRCh38
rs764660742 Chr.12: 89588556-89588556 on Build GRCh38
rs1040906783 Chr.12: 89588342-89588342 on Build GRCh38
rs932164149 Chr.12: 89588616-89588619 on Build GRCh38
rs911951892 Chr.12: 89588652-89588652 on Build GRCh38
rs951074384 Chr.12: 89588317-89588317 on Build GRCh38
rs983849193 Chr.12: 89588675-89588675 on Build GRCh38
rs1000951855 Chr.12: 89588460-89588460 on Build GRCh38
rs207473236 Chr.12: 89588490-89588490 on Build GRCh38
rs1000120518 Chr.12: 89588485-89588485 on Build GRCh38
rs1016601143 Chr.12: 89588545-89588545 on Build GRCh38
rs908308903 Chr.12: 89588716-89588716 on Build GRCh38
rs982378023 Chr.12: 89588544-89588544 on Build GRCh38
rs1041754882 Chr.12: 89588443-89588443 on Build GRCh38
rs770761133 Chr.12: 89588661-89588661 on Build GRCh38
rs145069315 Chr.12: 89588537-89588537 on Build GRCh38

View Full Product Details