Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89589658-89590207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

SNP Detail

SNP ID Chromosome Location
rs1015552427 Chr.12: 89590122-89590122 on Build GRCh38
rs1005752765 Chr.12: 89589695-89589695 on Build GRCh38
rs1033081401 Chr.12: 89589791-89589791 on Build GRCh38
rs953909098 Chr.12: 89589778-89589778 on Build GRCh38
rs957099678 Chr.12: 89589805-89589805 on Build GRCh38
rs981292396 Chr.12: 89589879-89589879 on Build GRCh38
rs144498091 Chr.12: 89589845-89589845 on Build GRCh38
rs577775232 Chr.12: 89590030-89590032 on Build GRCh38
rs548760600 Chr.12: 89589736-89589736 on Build GRCh38
rs1015840386 Chr.12: 89589716-89589716 on Build GRCh38
rs769621257 Chr.12: 89589831-89589832 on Build GRCh38
rs988686776 Chr.12: 89589808-89589808 on Build GRCh38
rs897428633 Chr.12: 89590116-89590116 on Build GRCh38
rs869048491 Chr.12: 89590175-89590178 on Build GRCh38
rs869061067 Chr.12: 89590168-89590168 on Build GRCh38
rs973895014 Chr.12: 89589759-89589759 on Build GRCh38
rs941369834 Chr.12: 89590082-89590082 on Build GRCh38
rs1037212729 Chr.12: 89590102-89590102 on Build GRCh38
rs938082293 Chr.12: 89589952-89589952 on Build GRCh38
rs147827425 Chr.12: 89590071-89590071 on Build GRCh38
rs985253388 Chr.12: 89589780-89589780 on Build GRCh38
rs537514187 Chr.12: 89589967-89589967 on Build GRCh38
rs927172774 Chr.12: 89589884-89589884 on Build GRCh38
rs890778741 Chr.12: 89589801-89589801 on Build GRCh38
rs745549634 Chr.12: 89590126-89590126 on Build GRCh38
rs182085535 Chr.12: 89589972-89589972 on Build GRCh38
rs183753564 Chr.12: 89590118-89590118 on Build GRCh38
rs932411891 Chr.12: 89589784-89589784 on Build GRCh38
rs762169278 Chr.12: 89589973-89589973 on Build GRCh38
rs998318456 Chr.12: 89589743-89589743 on Build GRCh38
rs1012981568 Chr.12: 89590073-89590073 on Build GRCh38
rs949836722 Chr.12: 89589842-89589842 on Build GRCh38
rs1042082931 Chr.12: 89589822-89589823 on Build GRCh38
rs974433551 Chr.12: 89590128-89590128 on Build GRCh38
rs889992679 Chr.12: 89590137-89590137 on Build GRCh38
rs538876125 Chr.12: 89590095-89590095 on Build GRCh38
rs1049535150 Chr.12: 89589794-89589794 on Build GRCh38
rs971465473 Chr.12: 89590100-89590100 on Build GRCh38
rs897291669 Chr.12: 89589728-89589728 on Build GRCh38
rs903394784 Chr.12: 89589823-89589826 on Build GRCh38
rs1007048705 Chr.12: 89590162-89590162 on Build GRCh38
rs1029388464 Chr.12: 89589754-89589754 on Build GRCh38
rs913159002 Chr.12: 89589821-89589821 on Build GRCh38

View Full Product Details