Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89590488-89591000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399475 c.3660C>G p.L1220L Chr.12: 89590987-89590987 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs989514046 Chr.12: 89590825-89590828 on Build GRCh38
rs191454131 Chr.12: 89590962-89590962 on Build GRCh38
rs991274672 Chr.12: 89590550-89590550 on Build GRCh38
rs965230330 Chr.12: 89590811-89590811 on Build GRCh38
rs957818392 Chr.12: 89590825-89590825 on Build GRCh38
rs971086069 Chr.12: 89590791-89590791 on Build GRCh38
rs1054584074 Chr.12: 89590728-89590728 on Build GRCh38
rs1026995530 Chr.12: 89590786-89590786 on Build GRCh38
rs541026380 Chr.12: 89590724-89590724 on Build GRCh38
rs775990335 Chr.12: 89590939-89590939 on Build GRCh38
rs1037547186 Chr.12: 89590706-89590706 on Build GRCh38
rs769927537 Chr.12: 89590808-89590808 on Build GRCh38
rs768192439 Chr.12: 89590937-89590937 on Build GRCh38
rs199802801 Chr.12: 89590558-89590558 on Build GRCh38
rs78076751 Chr.12: 89590825-89590825 on Build GRCh38
rs371640664 Chr.12: 89590824-89590825 on Build GRCh38
rs977935555 Chr.12: 89590821-89590821 on Build GRCh38
rs962753584 Chr.12: 89590927-89590927 on Build GRCh38
rs1051388369 Chr.12: 89590659-89590659 on Build GRCh38
rs914773055 Chr.12: 89590621-89590621 on Build GRCh38
rs1037216277 Chr.12: 89590883-89590883 on Build GRCh38
rs1031137246 Chr.12: 89590824-89590827 on Build GRCh38
rs182144764 Chr.12: 89590569-89590569 on Build GRCh38
rs909977361 Chr.12: 89590554-89590554 on Build GRCh38
rs181112736 Chr.12: 89590530-89590530 on Build GRCh38
rs531539802 Chr.12: 89590918-89590918 on Build GRCh38
rs148868798 Chr.12: 89590644-89590644 on Build GRCh38
rs1044541533 Chr.12: 89590655-89590655 on Build GRCh38
rs866234045 Chr.12: 89590560-89590560 on Build GRCh38
rs928789187 Chr.12: 89590836-89590837 on Build GRCh38
rs1053468923 Chr.12: 89590613-89590613 on Build GRCh38
rs892910160 Chr.12: 89590748-89590748 on Build GRCh38
rs918615092 Chr.12: 89590628-89590628 on Build GRCh38
rs995886625 Chr.12: 89590750-89590750 on Build GRCh38
rs931453054 Chr.12: 89590895-89590895 on Build GRCh38
rs897198104 Chr.12: 89590886-89590886 on Build GRCh38
rs773115766 Chr.12: 89590963-89590963 on Build GRCh38
rs143710306 Chr.12: 89590702-89590702 on Build GRCh38
rs1036974005 Chr.12: 89590601-89590601 on Build GRCh38
rs1010013944 Chr.12: 89590775-89590775 on Build GRCh38
rs114701386 Chr.12: 89590590-89590590 on Build GRCh38
rs562508539 Chr.12: 89590836-89590836 on Build GRCh38
rs889912756 Chr.12: 89590700-89590700 on Build GRCh38
rs368755572 Chr.12: 89590873-89590873 on Build GRCh38
rs551140844 Chr.12: 89590953-89590953 on Build GRCh38
rs879404210 Chr.12: 89590837-89590837 on Build GRCh38
rs186661759 Chr.12: 89590790-89590790 on Build GRCh38
rs948968233 Chr.12: 89590835-89590835 on Build GRCh38
rs909072014 Chr.12: 89590857-89590857 on Build GRCh38

View Full Product Details