Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89598450-89598989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6553588 c.3458C>A p.P1153H Chr.12: 89598637-89598637 on Build GRCh38
3813167 c.3415T>C p.S1139P Chr.12: 89598680-89598680 on Build GRCh38
4622748 c.3488C>T p.A1163V Chr.12: 89598607-89598607 on Build GRCh38
1586796 c.3452C>A p.S1151Y Chr.12: 89598643-89598643 on Build GRCh38
1995152 c.3408G>A p.R1136R Chr.12: 89598687-89598687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879090481 Chr.12: 89598899-89598899 on Build GRCh38
rs776656075 Chr.12: 89598792-89598792 on Build GRCh38
rs754471105 Chr.12: 89598680-89598680 on Build GRCh38
rs1042059276 Chr.12: 89598819-89598819 on Build GRCh38
rs763168352 Chr.12: 89598767-89598775 on Build GRCh38
rs910951645 Chr.12: 89598689-89598689 on Build GRCh38
rs796600203 Chr.12: 89598780-89598781 on Build GRCh38
rs773354657 Chr.12: 89598541-89598541 on Build GRCh38
rs911136744 Chr.12: 89598526-89598526 on Build GRCh38
rs781105389 Chr.12: 89598781-89598784 on Build GRCh38
rs893565372 Chr.12: 89598784-89598784 on Build GRCh38
rs765072481 Chr.12: 89598559-89598561 on Build GRCh38
rs909897284 Chr.12: 89598503-89598503 on Build GRCh38
rs942645517 Chr.12: 89598534-89598534 on Build GRCh38
rs779049824 Chr.12: 89598763-89598763 on Build GRCh38
rs765987662 Chr.12: 89598643-89598643 on Build GRCh38
rs78003840 Chr.12: 89598711-89598711 on Build GRCh38
rs776729297 Chr.12: 89598625-89598625 on Build GRCh38
rs750305317 Chr.12: 89598640-89598640 on Build GRCh38
rs748657084 Chr.12: 89598700-89598700 on Build GRCh38
rs1023762184 Chr.12: 89598792-89598793 on Build GRCh38
rs907022198 Chr.12: 89598841-89598841 on Build GRCh38
rs984129288 Chr.12: 89598954-89598954 on Build GRCh38
rs529097497 Chr.12: 89598556-89598556 on Build GRCh38
rs892797175 Chr.12: 89598795-89598795 on Build GRCh38
rs756411545 Chr.12: 89598703-89598703 on Build GRCh38
rs984122467 Chr.12: 89598492-89598492 on Build GRCh38
rs796193186 Chr.12: 89598629-89598629 on Build GRCh38
rs901024971 Chr.12: 89598630-89598630 on Build GRCh38
rs773599230 Chr.12: 89598805-89598805 on Build GRCh38
rs761636320 Chr.12: 89598628-89598628 on Build GRCh38
rs184935550 Chr.12: 89598735-89598735 on Build GRCh38
rs751059113 Chr.12: 89598668-89598668 on Build GRCh38
rs771149758 Chr.12: 89598750-89598750 on Build GRCh38
rs143275091 Chr.12: 89598781-89598782 on Build GRCh38
rs566194725 Chr.12: 89598784-89598785 on Build GRCh38
rs1035847184 Chr.12: 89598813-89598813 on Build GRCh38
rs764067538 Chr.12: 89598616-89598616 on Build GRCh38
rs1002223993 Chr.12: 89598806-89598806 on Build GRCh38
rs1016895385 Chr.12: 89598954-89598954 on Build GRCh38
rs778096901 Chr.12: 89598719-89598719 on Build GRCh38
rs568933475 Chr.12: 89598866-89598866 on Build GRCh38
rs771116355 Chr.12: 89598552-89598552 on Build GRCh38
rs781693758 Chr.12: 89598684-89598684 on Build GRCh38
rs766548128 Chr.12: 89598780-89598781 on Build GRCh38
rs952810483 Chr.12: 89598939-89598939 on Build GRCh38
rs918756481 Chr.12: 89598575-89598575 on Build GRCh38
rs911241124 Chr.12: 89598959-89598959 on Build GRCh38
rs772052453 Chr.12: 89598780-89598780 on Build GRCh38
rs959417270 Chr.12: 89598895-89598895 on Build GRCh38
rs774480914 Chr.12: 89598555-89598555 on Build GRCh38
rs765141104 Chr.12: 89598638-89598638 on Build GRCh38
rs749524221 Chr.12: 89598549-89598549 on Build GRCh38
rs763190035 Chr.12: 89598843-89598843 on Build GRCh38
rs765643194 Chr.12: 89598770-89598770 on Build GRCh38
rs750159750 Chr.12: 89598559-89598560 on Build GRCh38
rs1034162285 Chr.12: 89598846-89598846 on Build GRCh38
rs189067131 Chr.12: 89598912-89598912 on Build GRCh38
rs962957087 Chr.12: 89598533-89598533 on Build GRCh38
rs866065156 Chr.12: 89598679-89598679 on Build GRCh38
rs748860987 Chr.12: 89598649-89598649 on Build GRCh38
rs374894314 Chr.12: 89598929-89598929 on Build GRCh38
rs764171024 Chr.12: 89598868-89598868 on Build GRCh38

View Full Product Details