Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89599002-89599501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1628915 c.3326G>T p.R1109I Chr.12: 89599142-89599142 on Build GRCh38
6607579 c.3346A>G p.T1116A Chr.12: 89599122-89599122 on Build GRCh38
6607581 c.3184C>T p.P1062S Chr.12: 89599284-89599284 on Build GRCh38
6607585 c.3305G>A p.R1102H Chr.12: 89599163-89599163 on Build GRCh38
1628914 c.3326G>T p.R1109I Chr.12: 89599142-89599142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017464543 Chr.12: 89599374-89599374 on Build GRCh38
rs576466764 Chr.12: 89599306-89599306 on Build GRCh38
rs762706417 Chr.12: 89599079-89599079 on Build GRCh38
rs567705698 Chr.12: 89599049-89599049 on Build GRCh38
rs756792747 Chr.12: 89599028-89599028 on Build GRCh38
rs767026355 Chr.12: 89599097-89599097 on Build GRCh38
rs534457946 Chr.12: 89599039-89599039 on Build GRCh38
rs755114579 Chr.12: 89599048-89599048 on Build GRCh38
rs778284136 Chr.12: 89599087-89599088 on Build GRCh38
rs746094247 Chr.12: 89599167-89599167 on Build GRCh38
rs35974419 Chr.12: 89599292-89599293 on Build GRCh38
rs945731279 Chr.12: 89599206-89599206 on Build GRCh38
rs758667242 Chr.12: 89599334-89599334 on Build GRCh38
rs757581271 Chr.12: 89599156-89599156 on Build GRCh38
rs769922943 Chr.12: 89599075-89599075 on Build GRCh38
rs756609401 Chr.12: 89599104-89599104 on Build GRCh38
rs572124292 Chr.12: 89599368-89599368 on Build GRCh38
rs773111312 Chr.12: 89599077-89599077 on Build GRCh38
rs1041616775 Chr.12: 89599230-89599230 on Build GRCh38
rs753026592 Chr.12: 89599275-89599275 on Build GRCh38
rs556520609 Chr.12: 89599290-89599290 on Build GRCh38
rs757585812 Chr.12: 89599313-89599313 on Build GRCh38
rs745704815 Chr.12: 89599470-89599470 on Build GRCh38
rs1018378027 Chr.12: 89599347-89599347 on Build GRCh38
rs762771112 Chr.12: 89599201-89599201 on Build GRCh38
rs922484397 Chr.12: 89599424-89599424 on Build GRCh38
rs754204853 Chr.12: 89599153-89599153 on Build GRCh38
rs747129280 Chr.12: 89599185-89599185 on Build GRCh38
rs772398709 Chr.12: 89599171-89599171 on Build GRCh38
rs199955362 Chr.12: 89599261-89599261 on Build GRCh38
rs199565655 Chr.12: 89599317-89599317 on Build GRCh38
rs766008672 Chr.12: 89599087-89599087 on Build GRCh38
rs752167578 Chr.12: 89599299-89599299 on Build GRCh38
rs377632094 Chr.12: 89599183-89599183 on Build GRCh38
rs759017096 Chr.12: 89599093-89599093 on Build GRCh38
rs780473210 Chr.12: 89599337-89599337 on Build GRCh38
rs540097524 Chr.12: 89599397-89599397 on Build GRCh38
rs752261513 Chr.12: 89599098-89599098 on Build GRCh38
rs865869268 Chr.12: 89599160-89599160 on Build GRCh38
rs769832772 Chr.12: 89599195-89599195 on Build GRCh38
rs73443080 Chr.12: 89599088-89599088 on Build GRCh38
rs761832655 Chr.12: 89599074-89599074 on Build GRCh38
rs372936177 Chr.12: 89599163-89599163 on Build GRCh38
rs184499616 Chr.12: 89599310-89599310 on Build GRCh38
rs986938936 Chr.12: 89599059-89599059 on Build GRCh38
rs976787044 Chr.12: 89599394-89599394 on Build GRCh38
rs554514036 Chr.12: 89599040-89599040 on Build GRCh38
rs759097062 Chr.12: 89599287-89599287 on Build GRCh38
rs1012280325 Chr.12: 89599346-89599346 on Build GRCh38
rs759304954 Chr.12: 89599312-89599312 on Build GRCh38
rs773996677 Chr.12: 89599274-89599274 on Build GRCh38
rs773305924 Chr.12: 89599199-89599199 on Build GRCh38
rs756568339 Chr.12: 89599083-89599085 on Build GRCh38
rs376299523 Chr.12: 89599084-89599084 on Build GRCh38

View Full Product Details