Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 90953208-90953705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCER1
Entrez Gene ID
196477
Gene Name
coiled-coil glutamate rich protein 1
Gene Aliases
C12orf12
UniGene
Hs.128574
Gene Chromosome Location
Chr.12: 90952215-90955176 on Build GRCh38
Transcripts
NM_152638.3 NR_130711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4393859 c.1139G>A p.S380N Chr.12: 90953604-90953604 on Build GRCh38
6882786 c.1124G>A p.R375K Chr.12: 90953619-90953619 on Build GRCh38
431975 c.1144A>T p.T382S Chr.12: 90953599-90953599 on Build GRCh38
6622747 c.1105G>A p.V369I Chr.12: 90953638-90953638 on Build GRCh38
6213088 c.1179G>T p.V393V Chr.12: 90953564-90953564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200956633 Chr.12: 90953545-90953545 on Build GRCh38
rs764042876 Chr.12: 90953612-90953612 on Build GRCh38
rs779769895 Chr.12: 90953530-90953530 on Build GRCh38
rs1016171508 Chr.12: 90953555-90953555 on Build GRCh38
rs998998111 Chr.12: 90953654-90953654 on Build GRCh38
rs917682706 Chr.12: 90953306-90953306 on Build GRCh38
rs780961194 Chr.12: 90953633-90953633 on Build GRCh38
rs199852069 Chr.12: 90953631-90953631 on Build GRCh38
rs755752784 Chr.12: 90953652-90953652 on Build GRCh38
rs781765606 Chr.12: 90953665-90953665 on Build GRCh38
rs145405070 Chr.12: 90953601-90953601 on Build GRCh38
rs774451656 Chr.12: 90953598-90953598 on Build GRCh38
rs528219668 Chr.12: 90953615-90953615 on Build GRCh38
rs781647163 Chr.12: 90953548-90953548 on Build GRCh38
rs956375546 Chr.12: 90953263-90953263 on Build GRCh38
rs931136418 Chr.12: 90953416-90953416 on Build GRCh38
rs913492328 Chr.12: 90953304-90953304 on Build GRCh38
rs1004729720 Chr.12: 90953540-90953540 on Build GRCh38
rs987628764 Chr.12: 90953284-90953288 on Build GRCh38
rs750033012 Chr.12: 90953639-90953639 on Build GRCh38
rs764352304 Chr.12: 90953588-90953588 on Build GRCh38
rs1008243570 Chr.12: 90953485-90953485 on Build GRCh38
rs758137703 Chr.12: 90953640-90953640 on Build GRCh38
rs369040462 Chr.12: 90953325-90953325 on Build GRCh38
rs992205759 Chr.12: 90953293-90953293 on Build GRCh38
rs895892662 Chr.12: 90953398-90953400 on Build GRCh38
rs978774761 Chr.12: 90953333-90953333 on Build GRCh38
rs748484594 Chr.12: 90953678-90953678 on Build GRCh38
rs760998750 Chr.12: 90953467-90953467 on Build GRCh38
rs770038565 Chr.12: 90953684-90953684 on Build GRCh38
rs770908113 Chr.12: 90953593-90953593 on Build GRCh38
rs769990759 Chr.12: 90953573-90953573 on Build GRCh38
rs774775189 Chr.12: 90953250-90953250 on Build GRCh38
rs1046990314 Chr.12: 90953423-90953423 on Build GRCh38
rs904714602 Chr.12: 90953638-90953638 on Build GRCh38
rs779687869 Chr.12: 90953642-90953642 on Build GRCh38
rs373391977 Chr.12: 90953472-90953472 on Build GRCh38
rs543716523 Chr.12: 90953316-90953316 on Build GRCh38
rs574656484 Chr.12: 90953268-90953268 on Build GRCh38
rs778465131 Chr.12: 90953478-90953478 on Build GRCh38
rs140346503 Chr.12: 90953385-90953385 on Build GRCh38
rs749882809 Chr.12: 90953491-90953491 on Build GRCh38
rs757860865 Chr.12: 90953521-90953521 on Build GRCh38
rs747515353 Chr.12: 90953534-90953534 on Build GRCh38
rs376042086 Chr.12: 90953558-90953558 on Build GRCh38
rs773316610 Chr.12: 90953577-90953577 on Build GRCh38
rs937344290 Chr.12: 90953384-90953384 on Build GRCh38
rs537393521 Chr.12: 90953234-90953234 on Build GRCh38
rs539105127 Chr.12: 90953398-90953398 on Build GRCh38
rs768034864 Chr.12: 90953334-90953334 on Build GRCh38
rs563590819 Chr.12: 90953366-90953366 on Build GRCh38
rs755501697 Chr.12: 90953653-90953653 on Build GRCh38
rs17855513 Chr.12: 90953606-90953606 on Build GRCh38
rs752416827 Chr.12: 90953498-90953499 on Build GRCh38
rs1046176378 Chr.12: 90953474-90953474 on Build GRCh38
rs373737062 Chr.12: 90953429-90953429 on Build GRCh38
rs1031939041 Chr.12: 90953666-90953666 on Build GRCh38
rs761519228 Chr.12: 90953617-90953617 on Build GRCh38
rs751135601 Chr.12: 90953643-90953643 on Build GRCh38
rs760061684 Chr.12: 90953262-90953262 on Build GRCh38
rs955082591 Chr.12: 90953278-90953278 on Build GRCh38

View Full Product Details