Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 90954728-90955223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCER1
Entrez Gene ID
196477
Gene Name
coiled-coil glutamate rich protein 1
Gene Aliases
C12orf12
UniGene
Hs.128574
Gene Chromosome Location
Chr.12: 90952215-90955176 on Build GRCh38
Transcripts
NM_152638.3 NR_130711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3417256 c.15C>T p.L5L Chr.12: 90954728-90954728 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953850152 Chr.12: 90954772-90954772 on Build GRCh38
rs148304050 Chr.12: 90955141-90955141 on Build GRCh38
rs778176162 Chr.12: 90954757-90954757 on Build GRCh38
rs373595326 Chr.12: 90954852-90954852 on Build GRCh38
rs867243109 Chr.12: 90954874-90954874 on Build GRCh38
rs115628025 Chr.12: 90955061-90955061 on Build GRCh38
rs372899674 Chr.12: 90955115-90955115 on Build GRCh38
rs1018761688 Chr.12: 90955149-90955149 on Build GRCh38
rs3803100 Chr.12: 90954863-90954863 on Build GRCh38
rs757526649 Chr.12: 90954778-90954778 on Build GRCh38
rs940989801 Chr.12: 90955161-90955161 on Build GRCh38
rs550764744 Chr.12: 90955024-90955024 on Build GRCh38
rs754030690 Chr.12: 90954760-90954760 on Build GRCh38
rs957096520 Chr.12: 90955199-90955199 on Build GRCh38
rs954231084 Chr.12: 90955122-90955122 on Build GRCh38
rs773877156 Chr.12: 90954813-90954813 on Build GRCh38
rs564320171 Chr.12: 90954838-90954838 on Build GRCh38
rs902954665 Chr.12: 90954787-90954788 on Build GRCh38
rs546873295 Chr.12: 90954880-90954880 on Build GRCh38
rs762074606 Chr.12: 90955131-90955131 on Build GRCh38
rs935736369 Chr.12: 90954802-90954802 on Build GRCh38
rs143256938 Chr.12: 90954898-90954898 on Build GRCh38
rs1036658896 Chr.12: 90954909-90954909 on Build GRCh38
rs1045835219 Chr.12: 90955083-90955083 on Build GRCh38
rs889646971 Chr.12: 90955058-90955058 on Build GRCh38
rs953277139 Chr.12: 90954932-90954932 on Build GRCh38
rs370481825 Chr.12: 90954921-90954921 on Build GRCh38
rs377490423 Chr.12: 90955086-90955086 on Build GRCh38
rs769135966 Chr.12: 90954934-90954934 on Build GRCh38
rs1015524387 Chr.12: 90954864-90954864 on Build GRCh38
rs533483124 Chr.12: 90954890-90954890 on Build GRCh38
rs370211513 Chr.12: 90954806-90954806 on Build GRCh38
rs1044873585 Chr.12: 90954860-90954860 on Build GRCh38
rs911814524 Chr.12: 90954984-90954984 on Build GRCh38
rs1049326653 Chr.12: 90954870-90954870 on Build GRCh38
rs568091742 Chr.12: 90954960-90954960 on Build GRCh38
rs539599575 Chr.12: 90955063-90955063 on Build GRCh38
rs924303854 Chr.12: 90955029-90955029 on Build GRCh38
rs971798166 Chr.12: 90955088-90955088 on Build GRCh38
rs117919138 Chr.12: 90954779-90954779 on Build GRCh38
rs900830850 Chr.12: 90954995-90954995 on Build GRCh38
rs776008878 Chr.12: 90954892-90954892 on Build GRCh38
rs980241524 Chr.12: 90955092-90955092 on Build GRCh38
rs754066601 Chr.12: 90955067-90955067 on Build GRCh38
rs765273186 Chr.12: 90955146-90955146 on Build GRCh38
rs927501847 Chr.12: 90954861-90954861 on Build GRCh38
rs984887632 Chr.12: 90955137-90955137 on Build GRCh38
rs375821867 Chr.12: 90954881-90954881 on Build GRCh38
rs988799008 Chr.12: 90954827-90954827 on Build GRCh38
rs989221501 Chr.12: 90955070-90955070 on Build GRCh38
rs965856770 Chr.12: 90955158-90955158 on Build GRCh38
rs3803099 Chr.12: 90955009-90955009 on Build GRCh38
rs1019852098 Chr.12: 90955109-90955109 on Build GRCh38
rs553531489 Chr.12: 90955110-90955110 on Build GRCh38
rs924523102 Chr.12: 90955191-90955191 on Build GRCh38
rs886661399 Chr.12: 90954883-90954883 on Build GRCh38
rs750906576 Chr.12: 90954823-90954823 on Build GRCh38
rs977360510 Chr.12: 90955160-90955160 on Build GRCh38
rs963130173 Chr.12: 90954886-90954886 on Build GRCh38
rs1028807201 Chr.12: 90954891-90954891 on Build GRCh38
rs116821338 Chr.12: 90954828-90954828 on Build GRCh38
rs977098681 Chr.12: 90955022-90955022 on Build GRCh38
rs948920255 Chr.12: 90955076-90955076 on Build GRCh38
rs1015784622 Chr.12: 90955097-90955097 on Build GRCh38
rs71456517 Chr.12: 90955011-90955011 on Build GRCh38

View Full Product Details