Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95890741-95891246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC38
Entrez Gene ID
120935
Gene Name
coiled-coil domain containing 38
Gene Aliases
-
UniGene
Hs.210377
Gene Chromosome Location
Chr.12: 95867048-95943301 on Build GRCh38
Transcripts
NM_182496.2 XM_006719229.2 XM_011537883.2 XM_011537884.2 XM_011537886.2 XM_011537887.2 XM_011537888.2 XM_011537889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747307 c.811G>A p.E271K Chr.12: 95890892-95890892 on Build GRCh38
6350572 c.831T>C p.A277A Chr.12: 95890872-95890872 on Build GRCh38
3813243 c.857G>A p.G286E Chr.12: 95890846-95890846 on Build GRCh38
1299979 c.814G>C p.E272Q Chr.12: 95890889-95890889 on Build GRCh38
6622129 c.851C>A p.S284Y Chr.12: 95890852-95890852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756142478 Chr.12: 95890775-95890775 on Build GRCh38
rs983861215 Chr.12: 95890837-95890837 on Build GRCh38
rs769141643 Chr.12: 95890968-95890968 on Build GRCh38
rs1000978552 Chr.12: 95890774-95890774 on Build GRCh38
rs763514846 Chr.12: 95890819-95890819 on Build GRCh38
rs370923741 Chr.12: 95890978-95890978 on Build GRCh38
rs374889180 Chr.12: 95890816-95890816 on Build GRCh38
rs774557592 Chr.12: 95890887-95890889 on Build GRCh38
rs1017610954 Chr.12: 95890831-95890831 on Build GRCh38
rs371090001 Chr.12: 95890852-95890852 on Build GRCh38
rs774353469 Chr.12: 95890979-95890979 on Build GRCh38
rs761946685 Chr.12: 95890883-95890883 on Build GRCh38
rs772064975 Chr.12: 95890971-95890971 on Build GRCh38
rs35455788 Chr.12: 95890883-95890884 on Build GRCh38
rs1032408606 Chr.12: 95890780-95890780 on Build GRCh38
rs200906075 Chr.12: 95890786-95890786 on Build GRCh38
rs765246811 Chr.12: 95890793-95890793 on Build GRCh38
rs6538681 Chr.12: 95890872-95890872 on Build GRCh38
rs763807844 Chr.12: 95890929-95890929 on Build GRCh38
rs747484352 Chr.12: 95890964-95890964 on Build GRCh38
rs766418863 Chr.12: 95890909-95890909 on Build GRCh38
rs771693671 Chr.12: 95890848-95890848 on Build GRCh38
rs1008194900 Chr.12: 95890824-95890824 on Build GRCh38
rs139965944 Chr.12: 95890871-95890871 on Build GRCh38
rs910980289 Chr.12: 95891170-95891170 on Build GRCh38
rs537978222 Chr.12: 95891055-95891055 on Build GRCh38
rs746029921 Chr.12: 95890919-95890919 on Build GRCh38
rs866024502 Chr.12: 95890924-95890924 on Build GRCh38
rs971346684 Chr.12: 95890798-95890798 on Build GRCh38
rs933703842 Chr.12: 95891120-95891120 on Build GRCh38
rs748607370 Chr.12: 95890973-95890973 on Build GRCh38
rs763073791 Chr.12: 95890906-95890906 on Build GRCh38
rs541526866 Chr.12: 95890773-95890773 on Build GRCh38
rs150971634 Chr.12: 95891138-95891138 on Build GRCh38
rs547953917 Chr.12: 95890976-95890976 on Build GRCh38
rs140908091 Chr.12: 95891217-95891217 on Build GRCh38
rs773411975 Chr.12: 95890885-95890885 on Build GRCh38
rs529367030 Chr.12: 95890943-95890943 on Build GRCh38
rs770705010 Chr.12: 95890877-95890878 on Build GRCh38
rs994858422 Chr.12: 95891112-95891112 on Build GRCh38
rs796261736 Chr.12: 95891083-95891083 on Build GRCh38
rs558797702 Chr.12: 95891049-95891049 on Build GRCh38
rs555693056 Chr.12: 95891194-95891194 on Build GRCh38
rs183009873 Chr.12: 95891213-95891213 on Build GRCh38
rs533767797 Chr.12: 95890794-95890794 on Build GRCh38
rs752658250 Chr.12: 95890912-95890912 on Build GRCh38
rs375612160 Chr.12: 95890884-95890884 on Build GRCh38
rs566654117 Chr.12: 95890977-95890977 on Build GRCh38
rs753598852 Chr.12: 95890934-95890934 on Build GRCh38
rs775598764 Chr.12: 95890977-95890977 on Build GRCh38
rs756996860 Chr.12: 95890940-95890940 on Build GRCh38
rs142196054 Chr.12: 95890976-95890976 on Build GRCh38
rs1051950497 Chr.12: 95891184-95891184 on Build GRCh38
rs756024450 Chr.12: 95891056-95891056 on Build GRCh38
rs147138435 Chr.12: 95890925-95890925 on Build GRCh38
rs770034886 Chr.12: 95890974-95890974 on Build GRCh38
rs945988169 Chr.12: 95890919-95890919 on Build GRCh38
rs533772885 Chr.12: 95891042-95891042 on Build GRCh38
rs767502832 Chr.12: 95890799-95890799 on Build GRCh38
rs368325499 Chr.12: 95890944-95890944 on Build GRCh38
rs141952202 Chr.12: 95890832-95890832 on Build GRCh38
rs779663238 Chr.12: 95890956-95890956 on Build GRCh38
rs781547981 Chr.12: 95890970-95890970 on Build GRCh38
rs150685119 Chr.12: 95890836-95890836 on Build GRCh38
rs775717099 Chr.12: 95891084-95891084 on Build GRCh38
rs747486733 Chr.12: 95890870-95890870 on Build GRCh38
rs770162273 Chr.12: 95890805-95890805 on Build GRCh38
rs759863705 Chr.12: 95890950-95890950 on Build GRCh38
rs756873395 Chr.12: 95890822-95890822 on Build GRCh38
rs375989913 Chr.12: 95890983-95890983 on Build GRCh38
rs574166362 Chr.12: 95890770-95890770 on Build GRCh38
rs7954709 Chr.12: 95890811-95890811 on Build GRCh38
rs557081888 Chr.12: 95891145-95891145 on Build GRCh38
rs778568140 Chr.12: 95890826-95890826 on Build GRCh38
rs959545752 Chr.12: 95890788-95890788 on Build GRCh38
rs377584116 Chr.12: 95890795-95890795 on Build GRCh38
rs372764363 Chr.12: 95890792-95890792 on Build GRCh38
rs763937097 Chr.12: 95891093-95891093 on Build GRCh38
rs963726542 Chr.12: 95891176-95891176 on Build GRCh38
rs1025879111 Chr.12: 95891118-95891118 on Build GRCh38
rs537707764 Chr.12: 95890923-95890923 on Build GRCh38

View Full Product Details