Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10442126-10442624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APITD1
Entrez Gene ID
378708
Gene Name
apoptosis-inducing, TAF9-like domain 1
Gene Aliases
CENP-S, CENPS, FAAP16, MHF1
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10442815 on Build GRCh38
Transcripts
NM_199294.2 NR_036462.1 NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1

Gene Symbol
APITD1-CORT
Entrez Gene ID
100526739
Gene Name
APITD1-CORT readthrough
Gene Aliases
-
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10452003 on Build GRCh38
Transcripts
NM_199294.2 NR_036462.1 NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602176 c.348G>T p.K116N Chr.1: 10442336-10442336 on Build GRCh38
462520 c.354A>G p.S118S Chr.1: 10442342-10442342 on Build GRCh38
2155621 c.396_398delAGT p.V134delV Chr.1: 10442384-10442386 on Build GRCh38
6602179 c.277-1G>A p.? Chr.1: 10442264-10442264 on Build GRCh38
6049061 c.409G>A p.E137K Chr.1: 10442397-10442397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374660128 Chr.1: 10442416-10442416 on Build GRCh38
rs943523535 Chr.1: 10442288-10442288 on Build GRCh38
rs770975490 Chr.1: 10442428-10442428 on Build GRCh38
rs746629388 Chr.1: 10442399-10442399 on Build GRCh38
rs773520502 Chr.1: 10442320-10442320 on Build GRCh38
rs755520880 Chr.1: 10442384-10442384 on Build GRCh38
rs767016690 Chr.1: 10442247-10442247 on Build GRCh38
rs776471507 Chr.1: 10442274-10442274 on Build GRCh38
rs1007468379 Chr.1: 10442550-10442550 on Build GRCh38
rs1039460528 Chr.1: 10442209-10442209 on Build GRCh38
rs945616998 Chr.1: 10442265-10442265 on Build GRCh38
rs1033792793 Chr.1: 10442423-10442423 on Build GRCh38
rs773608700 Chr.1: 10442422-10442422 on Build GRCh38
rs899576173 Chr.1: 10442317-10442317 on Build GRCh38
rs966702596 Chr.1: 10442528-10442528 on Build GRCh38
rs962539713 Chr.1: 10442197-10442197 on Build GRCh38
rs202139467 Chr.1: 10442262-10442262 on Build GRCh38
rs921050746 Chr.1: 10442599-10442599 on Build GRCh38
rs745312672 Chr.1: 10442413-10442413 on Build GRCh38
rs964091999 Chr.1: 10442362-10442362 on Build GRCh38
rs767887699 Chr.1: 10442319-10442319 on Build GRCh38
rs980106970 Chr.1: 10442529-10442529 on Build GRCh38
rs1017179801 Chr.1: 10442191-10442191 on Build GRCh38
rs776540445 Chr.1: 10442342-10442343 on Build GRCh38
rs1043405597 Chr.1: 10442494-10442494 on Build GRCh38
rs200181245 Chr.1: 10442305-10442305 on Build GRCh38
rs779333169 Chr.1: 10442254-10442254 on Build GRCh38
rs770402378 Chr.1: 10442271-10442271 on Build GRCh38
rs750538622 Chr.1: 10442214-10442214 on Build GRCh38
rs766537484 Chr.1: 10442379-10442379 on Build GRCh38
rs760791153 Chr.1: 10442251-10442252 on Build GRCh38
rs60102421 Chr.1: 10442249-10442249 on Build GRCh38
rs960719809 Chr.1: 10442216-10442216 on Build GRCh38
rs868655415 Chr.1: 10442392-10442392 on Build GRCh38
rs376453769 Chr.1: 10442233-10442233 on Build GRCh38
rs760887605 Chr.1: 10442356-10442356 on Build GRCh38
rs976838855 Chr.1: 10442369-10442369 on Build GRCh38
rs777364388 Chr.1: 10442266-10442266 on Build GRCh38
rs538557550 Chr.1: 10442388-10442388 on Build GRCh38
rs926812845 Chr.1: 10442419-10442419 on Build GRCh38
rs748558053 Chr.1: 10442236-10442236 on Build GRCh38
rs943558242 Chr.1: 10442566-10442566 on Build GRCh38
rs759565264 Chr.1: 10442400-10442403 on Build GRCh38
rs755340738 Chr.1: 10442252-10442252 on Build GRCh38
rs748402018 Chr.1: 10442257-10442257 on Build GRCh38
rs766544904 Chr.1: 10442250-10442251 on Build GRCh38
rs756825633 Chr.1: 10442407-10442407 on Build GRCh38
rs960774826 Chr.1: 10442438-10442438 on Build GRCh38
rs182536626 Chr.1: 10442270-10442270 on Build GRCh38
rs369886397 Chr.1: 10442414-10442414 on Build GRCh38
rs760976958 Chr.1: 10442426-10442426 on Build GRCh38
rs375741629 Chr.1: 10442409-10442409 on Build GRCh38
rs115927965 Chr.1: 10442312-10442312 on Build GRCh38
rs146240548 Chr.1: 10442397-10442397 on Build GRCh38
rs33919161 Chr.1: 10442251-10442251 on Build GRCh38
rs75614849 Chr.1: 10442260-10442260 on Build GRCh38
rs758711988 Chr.1: 10442394-10442394 on Build GRCh38
rs1039448174 Chr.1: 10442302-10442302 on Build GRCh38
rs371433852 Chr.1: 10442443-10442443 on Build GRCh38
rs1039166153 Chr.1: 10442575-10442575 on Build GRCh38
rs765439195 Chr.1: 10442458-10442458 on Build GRCh38
rs1056136993 Chr.1: 10442190-10442190 on Build GRCh38
rs759492323 Chr.1: 10442444-10442444 on Build GRCh38
rs749121078 Chr.1: 10442417-10442417 on Build GRCh38
rs201200029 Chr.1: 10442235-10442235 on Build GRCh38
rs925536513 Chr.1: 10442559-10442559 on Build GRCh38
rs942411384 Chr.1: 10442188-10442188 on Build GRCh38
rs769538389 Chr.1: 10442284-10442284 on Build GRCh38
rs201711835 Chr.1: 10442316-10442316 on Build GRCh38
rs765595004 Chr.1: 10442386-10442386 on Build GRCh38
rs397979185 Chr.1: 10442260-10442260 on Build GRCh38
rs1022612045 Chr.1: 10442219-10442219 on Build GRCh38
rs772501815 Chr.1: 10442248-10442249 on Build GRCh38
rs377631941 Chr.1: 10442448-10442448 on Build GRCh38
rs1009619999 Chr.1: 10442464-10442464 on Build GRCh38
rs35796534 Chr.1: 10442455-10442456 on Build GRCh38
rs992147125 Chr.1: 10442226-10442226 on Build GRCh38
rs553647550 Chr.1: 10442559-10442560 on Build GRCh38
rs776774961 Chr.1: 10442442-10442442 on Build GRCh38
rs981277158 Chr.1: 10442208-10442208 on Build GRCh38
rs745811069 Chr.1: 10442278-10442278 on Build GRCh38
rs368912564 Chr.1: 10442382-10442382 on Build GRCh38

View Full Product Details