Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10460860-10461366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFA
Entrez Gene ID
1676
Gene Name
DNA fragmentation factor subunit alpha
Gene Aliases
DFF-45, DFF1, ICAD
UniGene
Hs.484782
Gene Chromosome Location
Chr.1: 10460531-10472556 on Build GRCh38
Transcripts
NM_004401.2 NM_213566.1

SNP Detail

SNP ID Chromosome Location
rs533569552 Chr.1: 10461117-10461117 on Build GRCh38
rs557534118 Chr.1: 10461159-10461159 on Build GRCh38
rs754025390 Chr.1: 10461187-10461187 on Build GRCh38
rs562124006 Chr.1: 10461200-10461200 on Build GRCh38
rs937416023 Chr.1: 10461059-10461059 on Build GRCh38
rs532325691 Chr.1: 10461235-10461235 on Build GRCh38
rs1051822512 Chr.1: 10461121-10461121 on Build GRCh38
rs528535323 Chr.1: 10460978-10460978 on Build GRCh38
rs529412377 Chr.1: 10461333-10461333 on Build GRCh38
rs967483860 Chr.1: 10461323-10461323 on Build GRCh38
rs747635394 Chr.1: 10461125-10461125 on Build GRCh38
rs749888591 Chr.1: 10460910-10460910 on Build GRCh38
rs34862260 Chr.1: 10460926-10460926 on Build GRCh38
rs898176360 Chr.1: 10461243-10461243 on Build GRCh38
rs571516497 Chr.1: 10461192-10461192 on Build GRCh38
rs1002089062 Chr.1: 10461260-10461260 on Build GRCh38
rs994646746 Chr.1: 10461053-10461053 on Build GRCh38
rs867915966 Chr.1: 10461044-10461044 on Build GRCh38
rs376299380 Chr.1: 10460887-10460887 on Build GRCh38
rs998566415 Chr.1: 10460985-10460985 on Build GRCh38
rs867427994 Chr.1: 10461102-10461102 on Build GRCh38
rs1014161891 Chr.1: 10461152-10461152 on Build GRCh38
rs755559011 Chr.1: 10460909-10460910 on Build GRCh38
rs936132634 Chr.1: 10461110-10461110 on Build GRCh38
rs967067202 Chr.1: 10461096-10461096 on Build GRCh38
rs980667385 Chr.1: 10461034-10461034 on Build GRCh38
rs756474659 Chr.1: 10461132-10461132 on Build GRCh38
rs1017002686 Chr.1: 10461043-10461043 on Build GRCh38
rs1039901078 Chr.1: 10461114-10461114 on Build GRCh38
rs546709707 Chr.1: 10461178-10461178 on Build GRCh38
rs1032735553 Chr.1: 10461166-10461166 on Build GRCh38
rs962852192 Chr.1: 10461023-10461023 on Build GRCh38
rs975948572 Chr.1: 10461179-10461179 on Build GRCh38
rs936237521 Chr.1: 10461176-10461176 on Build GRCh38
rs189424960 Chr.1: 10461054-10461054 on Build GRCh38
rs765490469 Chr.1: 10460908-10460910 on Build GRCh38
rs568592153 Chr.1: 10461046-10461046 on Build GRCh38
rs575793601 Chr.1: 10461161-10461161 on Build GRCh38
rs1024377705 Chr.1: 10461308-10461308 on Build GRCh38
rs201652956 Chr.1: 10460976-10460976 on Build GRCh38
rs664947 Chr.1: 10461208-10461208 on Build GRCh38
rs111443910 Chr.1: 10460906-10460906 on Build GRCh38
rs770774428 Chr.1: 10461134-10461134 on Build GRCh38
rs774097968 Chr.1: 10461183-10461183 on Build GRCh38
rs995043656 Chr.1: 10461138-10461138 on Build GRCh38
rs186083478 Chr.1: 10461100-10461100 on Build GRCh38
rs1004251701 Chr.1: 10461029-10461029 on Build GRCh38
rs866451291 Chr.1: 10461065-10461065 on Build GRCh38
rs960522071 Chr.1: 10461275-10461275 on Build GRCh38
rs569013535 Chr.1: 10461066-10461066 on Build GRCh38
rs566113039 Chr.1: 10461112-10461112 on Build GRCh38
rs1013464196 Chr.1: 10461296-10461296 on Build GRCh38
rs917216227 Chr.1: 10461079-10461079 on Build GRCh38
rs767375414 Chr.1: 10461203-10461203 on Build GRCh38
rs934167444 Chr.1: 10461185-10461185 on Build GRCh38
rs3829642 Chr.1: 10461342-10461342 on Build GRCh38
rs977106678 Chr.1: 10461162-10461162 on Build GRCh38
rs745828600 Chr.1: 10461169-10461170 on Build GRCh38
rs182037207 Chr.1: 10461058-10461058 on Build GRCh38
rs977443811 Chr.1: 10461153-10461154 on Build GRCh38
rs894337574 Chr.1: 10461147-10461147 on Build GRCh38
rs74834298 Chr.1: 10460927-10460927 on Build GRCh38
rs531244397 Chr.1: 10460886-10460886 on Build GRCh38
rs769292761 Chr.1: 10461128-10461128 on Build GRCh38
rs569152049 Chr.1: 10461343-10461343 on Build GRCh38
rs547095340 Chr.1: 10460994-10460994 on Build GRCh38
rs372395272 Chr.1: 10461157-10461157 on Build GRCh38
rs982621864 Chr.1: 10460888-10460888 on Build GRCh38
rs573414959 Chr.1: 10461150-10461150 on Build GRCh38
rs992863748 Chr.1: 10461064-10461064 on Build GRCh38
rs1016801570 Chr.1: 10461127-10461127 on Build GRCh38
rs142383097 Chr.1: 10461101-10461101 on Build GRCh38
rs1051110526 Chr.1: 10460990-10460990 on Build GRCh38
rs374537999 Chr.1: 10460885-10460885 on Build GRCh38
rs1135131 Chr.1: 10461143-10461143 on Build GRCh38
rs371959339 Chr.1: 10461199-10461199 on Build GRCh38
rs1057026 Chr.1: 10461197-10461197 on Build GRCh38
rs887102351 Chr.1: 10461123-10461123 on Build GRCh38
rs934264334 Chr.1: 10461119-10461119 on Build GRCh38
rs564655542 Chr.1: 10461188-10461188 on Build GRCh38
rs1038341001 Chr.1: 10461206-10461206 on Build GRCh38
rs898545037 Chr.1: 10461315-10461315 on Build GRCh38
rs1038171010 Chr.1: 10461004-10461004 on Build GRCh38
rs554529439 Chr.1: 10460959-10460959 on Build GRCh38
rs199811240 Chr.1: 10461024-10461024 on Build GRCh38
rs375616513 Chr.1: 10461118-10461118 on Build GRCh38
rs564501986 Chr.1: 10460969-10460969 on Build GRCh38

View Full Product Details