Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10466999-10467505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFA
Entrez Gene ID
1676
Gene Name
DNA fragmentation factor subunit alpha
Gene Aliases
DFF-45, DFF1, ICAD
UniGene
Hs.484782
Gene Chromosome Location
Chr.1: 10460531-10472556 on Build GRCh38
Transcripts
NM_004401.2 NM_213566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2116179 c.389A>G p.Q130R Chr.1: 10467242-10467242 on Build GRCh38
4020076 c.396A>T p.K132N Chr.1: 10467235-10467235 on Build GRCh38
6041803 c.393G>T p.L131L Chr.1: 10467238-10467238 on Build GRCh38
6041804 c.393G>T p.L131L Chr.1: 10467238-10467238 on Build GRCh38
3801109 c.425C>T p.S142L Chr.1: 10467206-10467206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201753478 Chr.1: 10467337-10467337 on Build GRCh38
rs990052146 Chr.1: 10467402-10467402 on Build GRCh38
rs944163755 Chr.1: 10467442-10467442 on Build GRCh38
rs973823200 Chr.1: 10467121-10467121 on Build GRCh38
rs891268785 Chr.1: 10467127-10467127 on Build GRCh38
rs934704663 Chr.1: 10467332-10467332 on Build GRCh38
rs754147021 Chr.1: 10467220-10467220 on Build GRCh38
rs748300602 Chr.1: 10467293-10467293 on Build GRCh38
rs1049437784 Chr.1: 10467424-10467424 on Build GRCh38
rs757159787 Chr.1: 10467272-10467272 on Build GRCh38
rs971611679 Chr.1: 10467278-10467278 on Build GRCh38
rs991140182 Chr.1: 10467240-10467240 on Build GRCh38
rs1037654956 Chr.1: 10467165-10467165 on Build GRCh38
rs757811839 Chr.1: 10467256-10467256 on Build GRCh38
rs143293907 Chr.1: 10467213-10467213 on Build GRCh38
rs1037199732 Chr.1: 10467478-10467478 on Build GRCh38
rs1050418019 Chr.1: 10467375-10467375 on Build GRCh38
rs370273583 Chr.1: 10467120-10467120 on Build GRCh38
rs879761585 Chr.1: 10467418-10467418 on Build GRCh38
rs768119414 Chr.1: 10467184-10467184 on Build GRCh38
rs867937510 Chr.1: 10467249-10467249 on Build GRCh38
rs763280107 Chr.1: 10467144-10467144 on Build GRCh38
rs774307153 Chr.1: 10467203-10467205 on Build GRCh38
rs200190552 Chr.1: 10467374-10467374 on Build GRCh38
rs766642706 Chr.1: 10467218-10467218 on Build GRCh38
rs777830922 Chr.1: 10467261-10467261 on Build GRCh38
rs71643094 Chr.1: 10467193-10467193 on Build GRCh38
rs768932062 Chr.1: 10467159-10467159 on Build GRCh38
rs531303673 Chr.1: 10467401-10467401 on Build GRCh38
rs745707454 Chr.1: 10467384-10467384 on Build GRCh38
rs3215947 Chr.1: 10467051-10467051 on Build GRCh38
rs371199069 Chr.1: 10467208-10467208 on Build GRCh38
rs891350816 Chr.1: 10467397-10467397 on Build GRCh38
rs763523699 Chr.1: 10467371-10467371 on Build GRCh38
rs750866837 Chr.1: 10467194-10467194 on Build GRCh38
rs779973233 Chr.1: 10467138-10467138 on Build GRCh38
rs919561139 Chr.1: 10467140-10467140 on Build GRCh38
rs181774839 Chr.1: 10467242-10467242 on Build GRCh38
rs932387941 Chr.1: 10467112-10467112 on Build GRCh38
rs766807668 Chr.1: 10467329-10467329 on Build GRCh38
rs766768174 Chr.1: 10467136-10467136 on Build GRCh38
rs569451729 Chr.1: 10467440-10467440 on Build GRCh38
rs865947278 Chr.1: 10467474-10467474 on Build GRCh38
rs914270629 Chr.1: 10467073-10467073 on Build GRCh38
rs923787980 Chr.1: 10467306-10467306 on Build GRCh38
rs879085482 Chr.1: 10467219-10467219 on Build GRCh38
rs138579895 Chr.1: 10467305-10467305 on Build GRCh38
rs747270629 Chr.1: 10467139-10467139 on Build GRCh38
rs147033969 Chr.1: 10467277-10467277 on Build GRCh38
rs981307570 Chr.1: 10467286-10467286 on Build GRCh38
rs897337873 Chr.1: 10467427-10467427 on Build GRCh38
rs548713735 Chr.1: 10467288-10467288 on Build GRCh38
rs764905737 Chr.1: 10467334-10467334 on Build GRCh38
rs2274986 Chr.1: 10467404-10467404 on Build GRCh38
rs757172761 Chr.1: 10467372-10467372 on Build GRCh38
rs1026979935 Chr.1: 10467214-10467214 on Build GRCh38
rs752249326 Chr.1: 10467251-10467251 on Build GRCh38
rs1054476997 Chr.1: 10467340-10467340 on Build GRCh38
rs757982041 Chr.1: 10467342-10467342 on Build GRCh38
rs540059103 Chr.1: 10467445-10467445 on Build GRCh38
rs759848494 Chr.1: 10467330-10467330 on Build GRCh38
rs746903840 Chr.1: 10467267-10467267 on Build GRCh38
rs71643093 Chr.1: 10467177-10467177 on Build GRCh38
rs754634163 Chr.1: 10467238-10467238 on Build GRCh38
rs761102520 Chr.1: 10467321-10467321 on Build GRCh38
rs781137474 Chr.1: 10467276-10467276 on Build GRCh38
rs761521224 Chr.1: 10467209-10467211 on Build GRCh38
rs774603856 Chr.1: 10467163-10467163 on Build GRCh38

View Full Product Details