Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98733828-98734338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
XM_006719507.3 XM_006719514.3 XM_017019653.1 XM_017019654.1 XM_017019658.1 NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
XM_006719507.3 XM_006719514.3 XM_017019653.1 XM_017019654.1 XM_017019658.1 NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1

SNP Detail

SNP ID Chromosome Location
rs374060808 Chr.12: 98734223-98734224 on Build GRCh38
rs564727447 Chr.12: 98734273-98734273 on Build GRCh38
rs773089739 Chr.12: 98734100-98734100 on Build GRCh38
rs879179549 Chr.12: 98734188-98734188 on Build GRCh38
rs11296996 Chr.12: 98733918-98733918 on Build GRCh38
rs916808744 Chr.12: 98734197-98734197 on Build GRCh38
rs539913464 Chr.12: 98733875-98733875 on Build GRCh38
rs890444437 Chr.12: 98734223-98734223 on Build GRCh38
rs888450118 Chr.12: 98733997-98733997 on Build GRCh38
rs35746294 Chr.12: 98734295-98734296 on Build GRCh38
rs998911538 Chr.12: 98734185-98734185 on Build GRCh38
rs939286488 Chr.12: 98733956-98733956 on Build GRCh38
rs1030094657 Chr.12: 98734077-98734077 on Build GRCh38
rs748981340 Chr.12: 98734102-98734102 on Build GRCh38
rs6538882 Chr.12: 98734224-98734224 on Build GRCh38
rs949570240 Chr.12: 98734199-98734199 on Build GRCh38
rs778619034 Chr.12: 98734300-98734300 on Build GRCh38
rs386765656 Chr.12: 98734223-98734224 on Build GRCh38
rs574225129 Chr.12: 98734045-98734045 on Build GRCh38
rs187010424 Chr.12: 98734250-98734250 on Build GRCh38
rs1027748990 Chr.12: 98734253-98734257 on Build GRCh38
rs950435537 Chr.12: 98733866-98733866 on Build GRCh38
rs900711296 Chr.12: 98734230-98734230 on Build GRCh38
rs191552272 Chr.12: 98734280-98734280 on Build GRCh38
rs747950449 Chr.12: 98734310-98734310 on Build GRCh38
rs752094345 Chr.12: 98734014-98734014 on Build GRCh38
rs145416277 Chr.12: 98734036-98734036 on Build GRCh38
rs1006875885 Chr.12: 98734288-98734288 on Build GRCh38
rs754516281 Chr.12: 98734222-98734222 on Build GRCh38
rs562294713 Chr.12: 98734041-98734041 on Build GRCh38
rs983090674 Chr.12: 98733916-98733916 on Build GRCh38
rs755727824 Chr.12: 98734029-98734029 on Build GRCh38
rs183510672 Chr.12: 98734054-98734054 on Build GRCh38
rs6538881 Chr.12: 98734223-98734223 on Build GRCh38
rs994191785 Chr.12: 98733946-98733946 on Build GRCh38
rs139964896 Chr.12: 98733981-98733981 on Build GRCh38
rs758831215 Chr.12: 98733859-98733859 on Build GRCh38
rs1006175154 Chr.12: 98734011-98734011 on Build GRCh38
rs977866014 Chr.12: 98734101-98734101 on Build GRCh38
rs398020798 Chr.12: 98733920-98733920 on Build GRCh38
rs558264298 Chr.12: 98733968-98733968 on Build GRCh38
rs936328032 Chr.12: 98734132-98734132 on Build GRCh38
rs991055898 Chr.12: 98734162-98734162 on Build GRCh38
rs559484281 Chr.12: 98734139-98734139 on Build GRCh38

View Full Product Details